Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57532852-57533345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN2
Entrez Gene ID
10540
Gene Name
dynactin subunit 2
Gene Aliases
DCTN50, DYNAMITIN, HEL-S-77, RBP50
UniGene
Hs.289123
Gene Chromosome Location
Chr.12: 57530050-57547331 on Build GRCh38
Transcripts
NM_001261412.1 NM_001261413.1 NM_006400.4 XM_011537761.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6378693 c.685G>T p.V229F Chr.12: 57533303-57533303 on Build GRCh38
7261927 c.688G>A p.A230T Chr.12: 57533300-57533300 on Build GRCh38
4811834 c.776G>A p.G259E Chr.12: 57532997-57532997 on Build GRCh38
6314085 c.730C>A p.R244S Chr.12: 57533258-57533258 on Build GRCh38
5482549 c.778G>T p.A260S Chr.12: 57532995-57532995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772045825 Chr.12: 57533203-57533203 on Build GRCh38
rs374990638 Chr.12: 57533269-57533269 on Build GRCh38
rs374915852 Chr.12: 57532941-57532941 on Build GRCh38
rs756899121 Chr.12: 57533280-57533280 on Build GRCh38
rs780910525 Chr.12: 57532938-57532938 on Build GRCh38
rs555604231 Chr.12: 57533163-57533163 on Build GRCh38
rs749772648 Chr.12: 57533088-57533088 on Build GRCh38
rs987269523 Chr.12: 57533076-57533076 on Build GRCh38
rs377537975 Chr.12: 57533221-57533221 on Build GRCh38
rs368867965 Chr.12: 57532918-57532918 on Build GRCh38
rs762243869 Chr.12: 57533000-57533000 on Build GRCh38
rs768553333 Chr.12: 57533227-57533227 on Build GRCh38
rs955366653 Chr.12: 57533258-57533258 on Build GRCh38
rs1028187808 Chr.12: 57533230-57533230 on Build GRCh38
rs998781140 Chr.12: 57533127-57533127 on Build GRCh38
rs763358233 Chr.12: 57533252-57533252 on Build GRCh38
rs374635042 Chr.12: 57533257-57533257 on Build GRCh38
rs760723090 Chr.12: 57532986-57532986 on Build GRCh38
rs772319650 Chr.12: 57533284-57533284 on Build GRCh38
rs748231491 Chr.12: 57533317-57533317 on Build GRCh38
rs747393007 Chr.12: 57532949-57532950 on Build GRCh38
rs2292656 Chr.12: 57533008-57533008 on Build GRCh38
rs939644012 Chr.12: 57532875-57532875 on Build GRCh38
rs768628340 Chr.12: 57533302-57533302 on Build GRCh38
rs755269025 Chr.12: 57533118-57533118 on Build GRCh38
rs748831125 Chr.12: 57533063-57533063 on Build GRCh38
rs780566233 Chr.12: 57533062-57533062 on Build GRCh38
rs748809587 Chr.12: 57533232-57533232 on Build GRCh38
rs777240098 Chr.12: 57532944-57532944 on Build GRCh38
rs770560656 Chr.12: 57532965-57532965 on Build GRCh38
rs200168725 Chr.12: 57533300-57533300 on Build GRCh38
rs755661027 Chr.12: 57533261-57533261 on Build GRCh38
rs573841285 Chr.12: 57533178-57533178 on Build GRCh38
rs995005444 Chr.12: 57532955-57532955 on Build GRCh38
rs760840844 Chr.12: 57533285-57533285 on Build GRCh38
rs111434135 Chr.12: 57533041-57533041 on Build GRCh38
rs866559621 Chr.12: 57533250-57533250 on Build GRCh38
rs776926470 Chr.12: 57533231-57533231 on Build GRCh38
rs758150188 Chr.12: 57533298-57533298 on Build GRCh38
rs372605251 Chr.12: 57532940-57532940 on Build GRCh38
rs373346070 Chr.12: 57533213-57533213 on Build GRCh38
rs371249543 Chr.12: 57533059-57533059 on Build GRCh38
rs200604619 Chr.12: 57532985-57532985 on Build GRCh38
rs200846627 Chr.12: 57533301-57533301 on Build GRCh38
rs767687338 Chr.12: 57533053-57533053 on Build GRCh38
rs770193570 Chr.12: 57533318-57533318 on Build GRCh38
rs778746712 Chr.12: 57533188-57533188 on Build GRCh38
rs773492177 Chr.12: 57533251-57533251 on Build GRCh38
rs762001763 Chr.12: 57533240-57533240 on Build GRCh38
rs1042499851 Chr.12: 57533283-57533283 on Build GRCh38
rs1038138891 Chr.12: 57532889-57532889 on Build GRCh38
rs534051294 Chr.12: 57533145-57533145 on Build GRCh38
rs774358145 Chr.12: 57532974-57532974 on Build GRCh38
rs146253880 Chr.12: 57533116-57533116 on Build GRCh38
rs779319249 Chr.12: 57533256-57533256 on Build GRCh38
rs756920328 Chr.12: 57533072-57533072 on Build GRCh38
rs776644821 Chr.12: 57533307-57533307 on Build GRCh38
rs370235568 Chr.12: 57533244-57533244 on Build GRCh38
rs566669140 Chr.12: 57533136-57533136 on Build GRCh38
rs745630176 Chr.12: 57533197-57533197 on Build GRCh38
rs759527435 Chr.12: 57532977-57532977 on Build GRCh38
rs73346014 Chr.12: 57533132-57533132 on Build GRCh38
rs911776493 Chr.12: 57533105-57533105 on Build GRCh38
rs780679600 Chr.12: 57533047-57533047 on Build GRCh38
rs771990781 Chr.12: 57532978-57532978 on Build GRCh38
rs753967632 Chr.12: 57532997-57532997 on Build GRCh38
rs753382974 Chr.12: 57533271-57533271 on Build GRCh38
rs759044305 Chr.12: 57533042-57533042 on Build GRCh38
rs777330105 Chr.12: 57533223-57533225 on Build GRCh38
rs769531856 Chr.12: 57532943-57532943 on Build GRCh38
rs755748899 Chr.12: 57533010-57533010 on Build GRCh38
rs369355032 Chr.12: 57532962-57532962 on Build GRCh38
rs764401715 Chr.12: 57532987-57532987 on Build GRCh38
rs539305059 Chr.12: 57532930-57532930 on Build GRCh38
rs750808338 Chr.12: 57533019-57533019 on Build GRCh38

View Full Product Details