Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57797258-57797763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVIL
Entrez Gene ID
10677
Gene Name
advillin
Gene Aliases
ADVIL, DOC6, p92
UniGene
Hs.584854
Gene Chromosome Location
Chr.12: 57797372-57818704 on Build GRCh38
Transcripts
NM_006576.3 XM_017018710.1 XM_017018711.1 XM_017018712.1 XM_017018713.1 XM_017018714.1 XM_017018715.1 NM_001172695.1 NM_001172696.1 NM_001172697.1 NM_005726.5

Gene Symbol
TSFM
Entrez Gene ID
10102
Gene Name
Ts translation elongation factor, mitochondrial
Gene Aliases
EFTS, EFTSMT
UniGene
Hs.632704
Gene Chromosome Location
Chr.12: 57782745-57802856 on Build GRCh38
Transcripts
NM_006576.3 XM_017018710.1 XM_017018711.1 XM_017018712.1 XM_017018713.1 XM_017018714.1 XM_017018715.1 NM_001172695.1 NM_001172696.1 NM_001172697.1 NM_005726.5

SNP Detail

SNP ID Chromosome Location
rs1007444971 Chr.12: 57797675-57797675 on Build GRCh38
rs58949411 Chr.12: 57797478-57797478 on Build GRCh38
rs953769659 Chr.12: 57797531-57797531 on Build GRCh38
rs1002689831 Chr.12: 57797421-57797421 on Build GRCh38
rs946197001 Chr.12: 57797382-57797382 on Build GRCh38
rs761171357 Chr.12: 57797386-57797386 on Build GRCh38
rs535671956 Chr.12: 57797735-57797735 on Build GRCh38
rs878971554 Chr.12: 57797301-57797301 on Build GRCh38
rs1049024144 Chr.12: 57797614-57797614 on Build GRCh38
rs530629327 Chr.12: 57797681-57797681 on Build GRCh38
rs563065302 Chr.12: 57797464-57797464 on Build GRCh38
rs145448503 Chr.12: 57797560-57797560 on Build GRCh38
rs568300779 Chr.12: 57797594-57797594 on Build GRCh38
rs887663754 Chr.12: 57797342-57797342 on Build GRCh38
rs910705851 Chr.12: 57797316-57797316 on Build GRCh38
rs993360328 Chr.12: 57797620-57797620 on Build GRCh38
rs530240073 Chr.12: 57797397-57797397 on Build GRCh38
rs568113109 Chr.12: 57797383-57797383 on Build GRCh38
rs766644978 Chr.12: 57797595-57797595 on Build GRCh38
rs890419020 Chr.12: 57797643-57797643 on Build GRCh38
rs74096250 Chr.12: 57797590-57797590 on Build GRCh38
rs766642456 Chr.12: 57797398-57797423 on Build GRCh38
rs748499502 Chr.12: 57797306-57797306 on Build GRCh38
rs1044691898 Chr.12: 57797694-57797694 on Build GRCh38
rs971185521 Chr.12: 57797390-57797390 on Build GRCh38
rs57561819 Chr.12: 57797541-57797541 on Build GRCh38
rs957816721 Chr.12: 57797313-57797313 on Build GRCh38
rs773569362 Chr.12: 57797340-57797340 on Build GRCh38
rs976261154 Chr.12: 57797575-57797575 on Build GRCh38
rs1022920049 Chr.12: 57797374-57797374 on Build GRCh38
rs946958990 Chr.12: 57797288-57797288 on Build GRCh38
rs548327552 Chr.12: 57797438-57797438 on Build GRCh38
rs146488830 Chr.12: 57797313-57797313 on Build GRCh38
rs993468550 Chr.12: 57797339-57797339 on Build GRCh38
rs114214463 Chr.12: 57797580-57797580 on Build GRCh38
rs887771449 Chr.12: 57797635-57797635 on Build GRCh38
rs955805754 Chr.12: 57797453-57797453 on Build GRCh38
rs944899100 Chr.12: 57797326-57797326 on Build GRCh38
rs1030162634 Chr.12: 57797623-57797623 on Build GRCh38
rs559936665 Chr.12: 57797337-57797337 on Build GRCh38
rs910831205 Chr.12: 57797548-57797548 on Build GRCh38
rs61938184 Chr.12: 57797655-57797655 on Build GRCh38
rs1021715229 Chr.12: 57797471-57797471 on Build GRCh38
rs1031354296 Chr.12: 57797422-57797422 on Build GRCh38
rs948771684 Chr.12: 57797658-57797658 on Build GRCh38
rs1040465683 Chr.12: 57797335-57797335 on Build GRCh38
rs928918291 Chr.12: 57797469-57797469 on Build GRCh38
rs1038161437 Chr.12: 57797582-57797582 on Build GRCh38
rs1013258364 Chr.12: 57797364-57797364 on Build GRCh38

View Full Product Details