Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 63142913-63143413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVPR1A
Entrez Gene ID
552
Gene Name
arginine vasopressin receptor 1A
Gene Aliases
AVPR V1a, AVPR1, V1aR
UniGene
Hs.2131
Gene Chromosome Location
Chr.12: 63142759-63152810 on Build GRCh38
Transcripts
NM_000706.4

SNP Detail

SNP ID Chromosome Location
rs1040548799 Chr.12: 63142940-63142940 on Build GRCh38
rs1007852583 Chr.12: 63142954-63142954 on Build GRCh38
rs932562814 Chr.12: 63143261-63143261 on Build GRCh38
rs960492361 Chr.12: 63143164-63143164 on Build GRCh38
rs944827114 Chr.12: 63143292-63143292 on Build GRCh38
rs1018353198 Chr.12: 63142960-63142960 on Build GRCh38
rs117611685 Chr.12: 63143222-63143222 on Build GRCh38
rs367959622 Chr.12: 63143255-63143255 on Build GRCh38
rs1000588313 Chr.12: 63143325-63143325 on Build GRCh38
rs1033731762 Chr.12: 63143155-63143157 on Build GRCh38
rs182143538 Chr.12: 63143103-63143103 on Build GRCh38
rs570804684 Chr.12: 63143174-63143174 on Build GRCh38
rs930093156 Chr.12: 63143223-63143223 on Build GRCh38
rs1000557231 Chr.12: 63142998-63142998 on Build GRCh38
rs538194964 Chr.12: 63143346-63143346 on Build GRCh38
rs1039399903 Chr.12: 63143298-63143298 on Build GRCh38
rs981499484 Chr.12: 63143140-63143140 on Build GRCh38
rs961224344 Chr.12: 63143242-63143242 on Build GRCh38
rs1043912507 Chr.12: 63143355-63143355 on Build GRCh38
rs539900266 Chr.12: 63143177-63143177 on Build GRCh38
rs140582095 Chr.12: 63143344-63143344 on Build GRCh38
rs963860087 Chr.12: 63142993-63142993 on Build GRCh38
rs1032159026 Chr.12: 63143102-63143102 on Build GRCh38
rs186786757 Chr.12: 63143195-63143195 on Build GRCh38
rs917803770 Chr.12: 63143107-63143107 on Build GRCh38
rs986155671 Chr.12: 63143273-63143273 on Build GRCh38
rs899489561 Chr.12: 63143303-63143303 on Build GRCh38
rs555543350 Chr.12: 63143259-63143259 on Build GRCh38
rs1025221210 Chr.12: 63143387-63143387 on Build GRCh38
rs945549081 Chr.12: 63143349-63143349 on Build GRCh38
rs567694265 Chr.12: 63143126-63143126 on Build GRCh38
rs910503239 Chr.12: 63143281-63143281 on Build GRCh38
rs1022581049 Chr.12: 63143041-63143041 on Build GRCh38
rs769134249 Chr.12: 63143244-63143244 on Build GRCh38
rs956541987 Chr.12: 63143347-63143347 on Build GRCh38
rs992295971 Chr.12: 63143212-63143212 on Build GRCh38
rs1774 Chr.12: 63143384-63143384 on Build GRCh38
rs1038441211 Chr.12: 63143209-63143209 on Build GRCh38
rs892613668 Chr.12: 63142991-63142991 on Build GRCh38
rs988031004 Chr.12: 63143105-63143105 on Build GRCh38

View Full Product Details