Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 63150095-63150588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVPR1A
Entrez Gene ID
552
Gene Name
arginine vasopressin receptor 1A
Gene Aliases
AVPR V1a, AVPR1, V1aR
UniGene
Hs.2131
Gene Chromosome Location
Chr.12: 63142759-63152810 on Build GRCh38
Transcripts
NM_000706.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
942399 c.515C>T p.A172V Chr.12: 63150322-63150322 on Build GRCh38
1283606 c.509T>C p.M170T Chr.12: 63150328-63150328 on Build GRCh38
3955051 c.513C>A p.I171I Chr.12: 63150324-63150324 on Build GRCh38
1990764 c.503G>A p.R168H Chr.12: 63150334-63150334 on Build GRCh38
4664792 c.454G>T p.A152S Chr.12: 63150383-63150383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747558639 Chr.12: 63150204-63150204 on Build GRCh38
rs768124807 Chr.12: 63150396-63150396 on Build GRCh38
rs181053969 Chr.12: 63150267-63150267 on Build GRCh38
rs974264460 Chr.12: 63150171-63150171 on Build GRCh38
rs773269527 Chr.12: 63150370-63150370 on Build GRCh38
rs770677863 Chr.12: 63150220-63150220 on Build GRCh38
rs1042615 Chr.12: 63150429-63150429 on Build GRCh38
rs771623728 Chr.12: 63150251-63150251 on Build GRCh38
rs762698235 Chr.12: 63150444-63150444 on Build GRCh38
rs563622430 Chr.12: 63150450-63150450 on Build GRCh38
rs751180151 Chr.12: 63150503-63150503 on Build GRCh38
rs767709671 Chr.12: 63150470-63150470 on Build GRCh38
rs371769743 Chr.12: 63150156-63150156 on Build GRCh38
rs951738536 Chr.12: 63150521-63150521 on Build GRCh38
rs146103040 Chr.12: 63150493-63150493 on Build GRCh38
rs756603967 Chr.12: 63150340-63150340 on Build GRCh38
rs748117343 Chr.12: 63150434-63150434 on Build GRCh38
rs945631623 Chr.12: 63150345-63150345 on Build GRCh38
rs180760072 Chr.12: 63150439-63150439 on Build GRCh38
rs761284955 Chr.12: 63150477-63150477 on Build GRCh38
rs748572296 Chr.12: 63150218-63150218 on Build GRCh38
rs868850167 Chr.12: 63150542-63150542 on Build GRCh38
rs537394626 Chr.12: 63150318-63150318 on Build GRCh38
rs199684240 Chr.12: 63150260-63150260 on Build GRCh38
rs775085135 Chr.12: 63150351-63150351 on Build GRCh38
rs754016556 Chr.12: 63150133-63150133 on Build GRCh38
rs780371622 Chr.12: 63150197-63150197 on Build GRCh38
rs899668332 Chr.12: 63150347-63150347 on Build GRCh38
rs769689727 Chr.12: 63150529-63150529 on Build GRCh38
rs779151917 Chr.12: 63150153-63150153 on Build GRCh38
rs770644257 Chr.12: 63150539-63150539 on Build GRCh38
rs920203508 Chr.12: 63150238-63150238 on Build GRCh38
rs753269775 Chr.12: 63150399-63150399 on Build GRCh38
rs369710823 Chr.12: 63150403-63150403 on Build GRCh38
rs546786206 Chr.12: 63150170-63150170 on Build GRCh38
rs912208925 Chr.12: 63150258-63150258 on Build GRCh38
rs775222891 Chr.12: 63150255-63150255 on Build GRCh38
rs763953060 Chr.12: 63150132-63150132 on Build GRCh38
rs748519112 Chr.12: 63150335-63150335 on Build GRCh38
rs750338761 Chr.12: 63150356-63150357 on Build GRCh38
rs976250964 Chr.12: 63150315-63150315 on Build GRCh38
rs780705756 Chr.12: 63150517-63150517 on Build GRCh38
rs112945642 Chr.12: 63150158-63150158 on Build GRCh38
rs925524625 Chr.12: 63150352-63150352 on Build GRCh38
rs1020309705 Chr.12: 63150416-63150416 on Build GRCh38
rs148278650 Chr.12: 63150285-63150285 on Build GRCh38
rs773038860 Chr.12: 63150438-63150438 on Build GRCh38
rs771180379 Chr.12: 63150446-63150446 on Build GRCh38
rs778311693 Chr.12: 63150286-63150286 on Build GRCh38
rs781774666 Chr.12: 63150334-63150334 on Build GRCh38
rs190242785 Chr.12: 63150211-63150211 on Build GRCh38
rs764555580 Chr.12: 63150490-63150490 on Build GRCh38
rs143160244 Chr.12: 63150282-63150282 on Build GRCh38
rs535731753 Chr.12: 63150241-63150241 on Build GRCh38
rs530628313 Chr.12: 63150297-63150297 on Build GRCh38
rs138241660 Chr.12: 63150369-63150369 on Build GRCh38
rs369032173 Chr.12: 63150363-63150363 on Build GRCh38
rs778650138 Chr.12: 63150341-63150341 on Build GRCh38
rs772227542 Chr.12: 63150560-63150560 on Build GRCh38
rs763936762 Chr.12: 63150307-63150307 on Build GRCh38
rs369768172 Chr.12: 63150561-63150561 on Build GRCh38
rs753214363 Chr.12: 63150474-63150474 on Build GRCh38
rs754196870 Chr.12: 63150492-63150492 on Build GRCh38
rs769533111 Chr.12: 63150437-63150437 on Build GRCh38
rs758567125 Chr.12: 63150182-63150182 on Build GRCh38
rs375549087 Chr.12: 63150410-63150410 on Build GRCh38
rs914049115 Chr.12: 63150344-63150344 on Build GRCh38
rs756736522 Chr.12: 63150197-63150198 on Build GRCh38
rs774566062 Chr.12: 63150545-63150545 on Build GRCh38
rs745458336 Chr.12: 63150418-63150418 on Build GRCh38
rs749096878 Chr.12: 63150534-63150534 on Build GRCh38
rs754449459 Chr.12: 63150513-63150513 on Build GRCh38
rs759692282 Chr.12: 63150214-63150214 on Build GRCh38
rs753439666 Chr.12: 63150167-63150168 on Build GRCh38
rs976163430 Chr.12: 63150508-63150508 on Build GRCh38
rs776384659 Chr.12: 63150265-63150265 on Build GRCh38
rs774213180 Chr.12: 63150236-63150236 on Build GRCh38
rs776488571 Chr.12: 63150430-63150430 on Build GRCh38
rs376518166 Chr.12: 63150203-63150203 on Build GRCh38
rs927452816 Chr.12: 63150122-63150122 on Build GRCh38
rs747708229 Chr.12: 63150522-63150522 on Build GRCh38
rs943668675 Chr.12: 63150565-63150565 on Build GRCh38
rs761542657 Chr.12: 63150366-63150366 on Build GRCh38
rs759630291 Chr.12: 63150452-63150452 on Build GRCh38
rs758345632 Chr.12: 63150290-63150290 on Build GRCh38
rs775685841 Chr.12: 63150566-63150566 on Build GRCh38
rs763745233 Chr.12: 63150261-63150261 on Build GRCh38
rs751963924 Chr.12: 63150300-63150300 on Build GRCh38
rs763262198 Chr.12: 63150374-63150374 on Build GRCh38
rs745537191 Chr.12: 63150348-63150348 on Build GRCh38
rs377107751 Chr.12: 63150380-63150380 on Build GRCh38
rs779608829 Chr.12: 63150422-63150422 on Build GRCh38
rs757489706 Chr.12: 63150143-63150143 on Build GRCh38
rs758045345 Chr.12: 63150420-63150420 on Build GRCh38
rs777640447 Chr.12: 63150531-63150531 on Build GRCh38
rs1037287168 Chr.12: 63150388-63150388 on Build GRCh38
rs779739647 Chr.12: 63150501-63150501 on Build GRCh38
rs758453856 Chr.12: 63150496-63150497 on Build GRCh38
rs146296879 Chr.12: 63150359-63150359 on Build GRCh38
rs755081148 Chr.12: 63150390-63150390 on Build GRCh38
rs553995625 Chr.12: 63150381-63150381 on Build GRCh38
rs966944204 Chr.12: 63150554-63150554 on Build GRCh38
rs149088415 Chr.12: 63150129-63150129 on Build GRCh38
rs2228155 Chr.12: 63150354-63150354 on Build GRCh38
rs2228153 Chr.12: 63150546-63150546 on Build GRCh38

View Full Product Details