Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64192941-64193448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf66
Entrez Gene ID
144577
Gene Name
chromosome 12 open reading frame 66
Gene Aliases
-
UniGene
Hs.444671
Gene Chromosome Location
Chr.12: 64186312-64222296 on Build GRCh38
Transcripts
NM_001300940.1 NM_001300941.1 NM_152440.4

SNP Detail

SNP ID Chromosome Location
rs1007193660 Chr.12: 64193035-64193035 on Build GRCh38
rs975228696 Chr.12: 64193081-64193081 on Build GRCh38
rs549250455 Chr.12: 64193000-64193000 on Build GRCh38
rs7969588 Chr.12: 64193186-64193186 on Build GRCh38
rs1026871032 Chr.12: 64193019-64193019 on Build GRCh38
rs762067188 Chr.12: 64193009-64193009 on Build GRCh38
rs946304572 Chr.12: 64193190-64193190 on Build GRCh38
rs1019234890 Chr.12: 64193395-64193395 on Build GRCh38
rs957468290 Chr.12: 64193204-64193204 on Build GRCh38
rs535757266 Chr.12: 64192970-64192970 on Build GRCh38
rs965089759 Chr.12: 64193074-64193074 on Build GRCh38
rs867235415 Chr.12: 64193253-64193253 on Build GRCh38
rs928654487 Chr.12: 64193239-64193239 on Build GRCh38
rs894350205 Chr.12: 64192974-64192974 on Build GRCh38
rs768076630 Chr.12: 64193338-64193338 on Build GRCh38
rs900079392 Chr.12: 64193107-64193107 on Build GRCh38
rs1048771177 Chr.12: 64193310-64193310 on Build GRCh38
rs1029795763 Chr.12: 64193176-64193176 on Build GRCh38
rs894561526 Chr.12: 64193282-64193282 on Build GRCh38
rs1003814560 Chr.12: 64193067-64193067 on Build GRCh38
rs914807059 Chr.12: 64193187-64193187 on Build GRCh38
rs887040844 Chr.12: 64193042-64193042 on Build GRCh38
rs887515105 Chr.12: 64193311-64193311 on Build GRCh38
rs949991655 Chr.12: 64193268-64193268 on Build GRCh38
rs151219869 Chr.12: 64193346-64193346 on Build GRCh38
rs371208109 Chr.12: 64193304-64193304 on Build GRCh38
rs1028492706 Chr.12: 64193100-64193100 on Build GRCh38
rs576153400 Chr.12: 64193200-64193200 on Build GRCh38
rs115391565 Chr.12: 64193408-64193408 on Build GRCh38
rs61931230 Chr.12: 64192994-64192994 on Build GRCh38
rs573146374 Chr.12: 64193231-64193231 on Build GRCh38
rs115027388 Chr.12: 64193072-64193072 on Build GRCh38
rs778328652 Chr.12: 64193280-64193280 on Build GRCh38
rs79466051 Chr.12: 64193367-64193367 on Build GRCh38
rs1056027054 Chr.12: 64192967-64192967 on Build GRCh38
rs981774838 Chr.12: 64193286-64193286 on Build GRCh38
rs568665777 Chr.12: 64193109-64193109 on Build GRCh38
rs148652510 Chr.12: 64193080-64193080 on Build GRCh38
rs998877500 Chr.12: 64193108-64193108 on Build GRCh38
rs569170576 Chr.12: 64193066-64193066 on Build GRCh38
rs575518388 Chr.12: 64193296-64193296 on Build GRCh38
rs368954919 Chr.12: 64193252-64193252 on Build GRCh38
rs919058383 Chr.12: 64193294-64193294 on Build GRCh38
rs116677548 Chr.12: 64193330-64193330 on Build GRCh38
rs79254473 Chr.12: 64193361-64193361 on Build GRCh38
rs998408666 Chr.12: 64193357-64193357 on Build GRCh38
rs185135732 Chr.12: 64193169-64193169 on Build GRCh38
rs1038667598 Chr.12: 64193341-64193341 on Build GRCh38
rs745895749 Chr.12: 64193188-64193189 on Build GRCh38
rs541820361 Chr.12: 64193246-64193246 on Build GRCh38
rs993229129 Chr.12: 64192997-64192997 on Build GRCh38
rs1048800691 Chr.12: 64193004-64193004 on Build GRCh38

View Full Product Details