Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64736922-64737424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNS
Entrez Gene ID
2799
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Gene Aliases
G6S
UniGene
Hs.334534
Gene Chromosome Location
Chr.12: 64713442-64759446 on Build GRCh38
Transcripts
NM_002076.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1991294 c.1031A>T p.Y344F Chr.12: 64737071-64737071 on Build GRCh38
4846709 c.1020G>C p.K340N Chr.12: 64737082-64737082 on Build GRCh38
3792891 c.1087C>G p.Q363E Chr.12: 64737015-64737015 on Build GRCh38
1606502 c.1040A>T p.D347V Chr.12: 64737062-64737062 on Build GRCh38
5083014 c.995-8G>A p.? Chr.12: 64737115-64737115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147537682 Chr.12: 64737096-64737096 on Build GRCh38
rs764333876 Chr.12: 64737141-64737141 on Build GRCh38
rs200521875 Chr.12: 64737122-64737122 on Build GRCh38
rs750838819 Chr.12: 64737236-64737236 on Build GRCh38
rs769308458 Chr.12: 64737054-64737054 on Build GRCh38
rs1016336468 Chr.12: 64737199-64737199 on Build GRCh38
rs759777247 Chr.12: 64737110-64737110 on Build GRCh38
rs559585451 Chr.12: 64737309-64737309 on Build GRCh38
rs531223748 Chr.12: 64737382-64737382 on Build GRCh38
rs750705389 Chr.12: 64737156-64737156 on Build GRCh38
rs369866661 Chr.12: 64737013-64737013 on Build GRCh38
rs1046086978 Chr.12: 64737018-64737018 on Build GRCh38
rs937632426 Chr.12: 64737388-64737388 on Build GRCh38
rs1004688091 Chr.12: 64737214-64737214 on Build GRCh38
rs774066835 Chr.12: 64737094-64737094 on Build GRCh38
rs945038474 Chr.12: 64736997-64736997 on Build GRCh38
rs754398113 Chr.12: 64736980-64736980 on Build GRCh38
rs954864929 Chr.12: 64737257-64737257 on Build GRCh38
rs190329056 Chr.12: 64737160-64737160 on Build GRCh38
rs542541100 Chr.12: 64737335-64737335 on Build GRCh38
rs934287662 Chr.12: 64737316-64737316 on Build GRCh38
rs762457978 Chr.12: 64737152-64737152 on Build GRCh38
rs371914257 Chr.12: 64737073-64737073 on Build GRCh38
rs184120469 Chr.12: 64737377-64737377 on Build GRCh38
rs119461974 Chr.12: 64737039-64737039 on Build GRCh38
rs780928340 Chr.12: 64737049-64737049 on Build GRCh38
rs567183752 Chr.12: 64736972-64736972 on Build GRCh38
rs374200272 Chr.12: 64736976-64736976 on Build GRCh38
rs760767113 Chr.12: 64737127-64737127 on Build GRCh38
rs114429281 Chr.12: 64737310-64737310 on Build GRCh38
rs1035579491 Chr.12: 64737159-64737159 on Build GRCh38
rs1044229045 Chr.12: 64737365-64737365 on Build GRCh38
rs575740832 Chr.12: 64737145-64737145 on Build GRCh38
rs1019641305 Chr.12: 64737222-64737222 on Build GRCh38
rs963819822 Chr.12: 64737221-64737221 on Build GRCh38
rs76432695 Chr.12: 64737392-64737392 on Build GRCh38
rs779476775 Chr.12: 64737031-64737031 on Build GRCh38
rs1029607903 Chr.12: 64737251-64737251 on Build GRCh38
rs996814528 Chr.12: 64737291-64737291 on Build GRCh38
rs191598266 Chr.12: 64737250-64737250 on Build GRCh38
rs375533636 Chr.12: 64737090-64737090 on Build GRCh38
rs914311698 Chr.12: 64737349-64737349 on Build GRCh38
rs577716013 Chr.12: 64737200-64737200 on Build GRCh38
rs115586725 Chr.12: 64737076-64737076 on Build GRCh38
rs982259364 Chr.12: 64737368-64737368 on Build GRCh38
rs1005304180 Chr.12: 64737181-64737186 on Build GRCh38
rs578022380 Chr.12: 64737150-64737150 on Build GRCh38
rs747563703 Chr.12: 64737051-64737051 on Build GRCh38
rs772132248 Chr.12: 64737121-64737121 on Build GRCh38
rs111741965 Chr.12: 64737316-64737316 on Build GRCh38
rs369864878 Chr.12: 64737155-64737155 on Build GRCh38
rs1056955091 Chr.12: 64737403-64737403 on Build GRCh38
rs751200375 Chr.12: 64737038-64737038 on Build GRCh38
rs966477922 Chr.12: 64737363-64737363 on Build GRCh38
rs913673675 Chr.12: 64737332-64737332 on Build GRCh38
rs957867415 Chr.12: 64737314-64737314 on Build GRCh38
rs754626354 Chr.12: 64737046-64737046 on Build GRCh38

View Full Product Details