Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 67656996-67657491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK2
Entrez Gene ID
8445
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 2
Gene Aliases
-
UniGene
Hs.173135
Gene Chromosome Location
Chr.12: 67648732-67665405 on Build GRCh38
Transcripts
NM_003583.3 NM_006482.2 XM_017020032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
244174 c.416C>T p.S139F Chr.12: 67657323-67657323 on Build GRCh38
5515492 c.549C>A p.F183L Chr.12: 67657456-67657456 on Build GRCh38
6553469 c.459C>T p.P153P Chr.12: 67657366-67657366 on Build GRCh38
942817 c.435C>T p.G145G Chr.12: 67657342-67657342 on Build GRCh38
4147414 c.308T>A p.V103E Chr.12: 67657215-67657215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757234288 Chr.12: 67657089-67657089 on Build GRCh38
rs768498909 Chr.12: 67657291-67657291 on Build GRCh38
rs773201871 Chr.12: 67657304-67657304 on Build GRCh38
rs138020482 Chr.12: 67657175-67657175 on Build GRCh38
rs764808820 Chr.12: 67657453-67657453 on Build GRCh38
rs923691915 Chr.12: 67657448-67657448 on Build GRCh38
rs764431941 Chr.12: 67657176-67657176 on Build GRCh38
rs549647443 Chr.12: 67657052-67657052 on Build GRCh38
rs772124910 Chr.12: 67657156-67657156 on Build GRCh38
rs767837825 Chr.12: 67657327-67657327 on Build GRCh38
rs747006331 Chr.12: 67657393-67657393 on Build GRCh38
rs530244321 Chr.12: 67657036-67657036 on Build GRCh38
rs537543305 Chr.12: 67657462-67657462 on Build GRCh38
rs373326108 Chr.12: 67657280-67657280 on Build GRCh38
rs777366599 Chr.12: 67657140-67657140 on Build GRCh38
rs528905325 Chr.12: 67657147-67657147 on Build GRCh38
rs955628835 Chr.12: 67657033-67657033 on Build GRCh38
rs754908658 Chr.12: 67657469-67657469 on Build GRCh38
rs751954468 Chr.12: 67657225-67657225 on Build GRCh38
rs933131402 Chr.12: 67657430-67657430 on Build GRCh38
rs752009659 Chr.12: 67657315-67657315 on Build GRCh38
rs753092948 Chr.12: 67657332-67657332 on Build GRCh38
rs1029808577 Chr.12: 67657031-67657031 on Build GRCh38
rs758935204 Chr.12: 67657111-67657111 on Build GRCh38
rs994324956 Chr.12: 67657023-67657023 on Build GRCh38
rs770033499 Chr.12: 67657300-67657300 on Build GRCh38
rs539367942 Chr.12: 67657394-67657394 on Build GRCh38
rs924007523 Chr.12: 67657171-67657171 on Build GRCh38
rs763038832 Chr.12: 67657414-67657414 on Build GRCh38
rs149514292 Chr.12: 67657222-67657222 on Build GRCh38
rs909089798 Chr.12: 67657024-67657024 on Build GRCh38
rs368663247 Chr.12: 67657208-67657208 on Build GRCh38
rs780144360 Chr.12: 67657378-67657378 on Build GRCh38
rs200772204 Chr.12: 67657447-67657447 on Build GRCh38
rs765295796 Chr.12: 67657198-67657198 on Build GRCh38
rs61744989 Chr.12: 67657293-67657293 on Build GRCh38
rs981967777 Chr.12: 67657290-67657291 on Build GRCh38
rs755291219 Chr.12: 67657230-67657230 on Build GRCh38
rs755807656 Chr.12: 67657133-67657133 on Build GRCh38
rs773437748 Chr.12: 67657413-67657413 on Build GRCh38
rs749974322 Chr.12: 67657367-67657367 on Build GRCh38
rs753981412 Chr.12: 67657063-67657063 on Build GRCh38
rs768047878 Chr.12: 67657423-67657423 on Build GRCh38
rs780452325 Chr.12: 67657127-67657127 on Build GRCh38
rs372026905 Chr.12: 67657257-67657257 on Build GRCh38
rs201692417 Chr.12: 67657281-67657281 on Build GRCh38
rs747348496 Chr.12: 67657132-67657132 on Build GRCh38
rs35139851 Chr.12: 67657199-67657199 on Build GRCh38
rs748856707 Chr.12: 67657144-67657144 on Build GRCh38
rs369962925 Chr.12: 67657269-67657269 on Build GRCh38
rs747940965 Chr.12: 67657399-67657399 on Build GRCh38
rs941860034 Chr.12: 67657026-67657026 on Build GRCh38
rs775477051 Chr.12: 67657157-67657157 on Build GRCh38
rs769942888 Chr.12: 67657410-67657410 on Build GRCh38
rs745859869 Chr.12: 67657153-67657153 on Build GRCh38
rs148267811 Chr.12: 67657434-67657434 on Build GRCh38
rs986078966 Chr.12: 67657048-67657048 on Build GRCh38
rs777497932 Chr.12: 67657248-67657248 on Build GRCh38
rs756064741 Chr.12: 67657045-67657045 on Build GRCh38
rs772165908 Chr.12: 67657272-67657272 on Build GRCh38
rs778616034 Chr.12: 67657260-67657262 on Build GRCh38
rs983446748 Chr.12: 67657350-67657350 on Build GRCh38
rs756911170 Chr.12: 67657334-67657334 on Build GRCh38
rs376543566 Chr.12: 67657056-67657056 on Build GRCh38
rs192078400 Chr.12: 67657395-67657395 on Build GRCh38
rs777721568 Chr.12: 67657067-67657067 on Build GRCh38
rs569784741 Chr.12: 67657415-67657415 on Build GRCh38
rs546992961 Chr.12: 67657162-67657162 on Build GRCh38
rs199671497 Chr.12: 67657148-67657148 on Build GRCh38
rs751088641 Chr.12: 67657463-67657463 on Build GRCh38
rs376315832 Chr.12: 67657258-67657258 on Build GRCh38
rs968443245 Chr.12: 67657112-67657112 on Build GRCh38
rs750825894 Chr.12: 67657097-67657097 on Build GRCh38
rs758950789 Chr.12: 67657221-67657221 on Build GRCh38
rs764378967 Chr.12: 67657061-67657061 on Build GRCh38
rs80173460 Chr.12: 67657372-67657372 on Build GRCh38
rs757060907 Chr.12: 67657244-67657245 on Build GRCh38
rs750029345 Chr.12: 67657459-67657459 on Build GRCh38
rs200418247 Chr.12: 67657131-67657131 on Build GRCh38
rs893362378 Chr.12: 67657242-67657242 on Build GRCh38
rs761954038 Chr.12: 67657188-67657188 on Build GRCh38
rs376836219 Chr.12: 67657163-67657163 on Build GRCh38
rs774410869 Chr.12: 67657417-67657417 on Build GRCh38
rs776478421 Chr.12: 67657292-67657292 on Build GRCh38
rs765786973 Chr.12: 67657091-67657091 on Build GRCh38
rs749177348 Chr.12: 67657079-67657084 on Build GRCh38
rs991964702 Chr.12: 67657236-67657236 on Build GRCh38
rs567711118 Chr.12: 67657054-67657054 on Build GRCh38
rs778346638 Chr.12: 67657266-67657266 on Build GRCh38
rs759901724 Chr.12: 67657318-67657318 on Build GRCh38
rs900322867 Chr.12: 67657039-67657039 on Build GRCh38
rs778598481 Chr.12: 67657366-67657366 on Build GRCh38
rs780963887 Chr.12: 67657180-67657180 on Build GRCh38

View Full Product Details