Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 68852880-68853429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPM
Entrez Gene ID
1368
Gene Name
carboxypeptidase M
Gene Aliases
-
UniGene
Hs.654387
Gene Chromosome Location
Chr.12: 68842200-68963240 on Build GRCh38
Transcripts
NM_001005502.2 NM_001874.4 NM_198320.3

SNP Detail

SNP ID Chromosome Location
rs530530602 Chr.12: 68853187-68853187 on Build GRCh38
rs1009558844 Chr.12: 68853381-68853381 on Build GRCh38
rs752836014 Chr.12: 68853010-68853010 on Build GRCh38
rs190409295 Chr.12: 68853125-68853125 on Build GRCh38
rs369916260 Chr.12: 68852956-68852956 on Build GRCh38
rs1019177169 Chr.12: 68852945-68852945 on Build GRCh38
rs1022586205 Chr.12: 68853038-68853038 on Build GRCh38
rs968698814 Chr.12: 68853097-68853097 on Build GRCh38
rs1040246362 Chr.12: 68853206-68853207 on Build GRCh38
rs944108631 Chr.12: 68853202-68853202 on Build GRCh38
rs892475389 Chr.12: 68853012-68853012 on Build GRCh38
rs1027540755 Chr.12: 68853246-68853246 on Build GRCh38
rs941241838 Chr.12: 68852948-68852948 on Build GRCh38
rs909749745 Chr.12: 68852921-68852921 on Build GRCh38
rs998246196 Chr.12: 68853009-68853009 on Build GRCh38
rs951655655 Chr.12: 68853008-68853008 on Build GRCh38
rs547014942 Chr.12: 68853275-68853275 on Build GRCh38
rs183801591 Chr.12: 68853209-68853209 on Build GRCh38
rs1051102519 Chr.12: 68853011-68853011 on Build GRCh38
rs77971375 Chr.12: 68853189-68853189 on Build GRCh38
rs552856584 Chr.12: 68852942-68852943 on Build GRCh38
rs1027343415 Chr.12: 68852949-68852949 on Build GRCh38
rs538914320 Chr.12: 68853400-68853400 on Build GRCh38
rs971533476 Chr.12: 68853091-68853091 on Build GRCh38
rs375736747 Chr.12: 68853279-68853279 on Build GRCh38
rs751363741 Chr.12: 68852994-68852994 on Build GRCh38
rs573875697 Chr.12: 68852912-68852912 on Build GRCh38
rs571927268 Chr.12: 68853282-68853282 on Build GRCh38
rs982850842 Chr.12: 68853193-68853193 on Build GRCh38
rs555638988 Chr.12: 68852907-68852907 on Build GRCh38
rs1009485413 Chr.12: 68853019-68853019 on Build GRCh38
rs1034245334 Chr.12: 68853112-68853112 on Build GRCh38
rs577825566 Chr.12: 68853105-68853105 on Build GRCh38
rs563092081 Chr.12: 68853140-68853140 on Build GRCh38
rs868395193 Chr.12: 68853159-68853159 on Build GRCh38
rs909832583 Chr.12: 68853204-68853204 on Build GRCh38
rs933794142 Chr.12: 68853272-68853272 on Build GRCh38
rs964653474 Chr.12: 68852947-68852947 on Build GRCh38
rs921111501 Chr.12: 68853232-68853232 on Build GRCh38
rs764259309 Chr.12: 68853384-68853384 on Build GRCh38
rs887688956 Chr.12: 68853328-68853328 on Build GRCh38
rs892549347 Chr.12: 68853339-68853339 on Build GRCh38
rs1039560423 Chr.12: 68853212-68853212 on Build GRCh38
rs757058077 Chr.12: 68853399-68853400 on Build GRCh38
rs796246543 Chr.12: 68853332-68853332 on Build GRCh38
rs566385119 Chr.12: 68853129-68853130 on Build GRCh38
rs879512042 Chr.12: 68853395-68853396 on Build GRCh38
rs914582493 Chr.12: 68853153-68853153 on Build GRCh38
rs960841561 Chr.12: 68853151-68853151 on Build GRCh38
rs373314883 Chr.12: 68853044-68853044 on Build GRCh38
rs891234726 Chr.12: 68853190-68853191 on Build GRCh38
rs899940856 Chr.12: 68853005-68853005 on Build GRCh38
rs561196262 Chr.12: 68853201-68853201 on Build GRCh38
rs767582591 Chr.12: 68852995-68852995 on Build GRCh38
rs551758892 Chr.12: 68853014-68853014 on Build GRCh38
rs143238687 Chr.12: 68853204-68853205 on Build GRCh38

View Full Product Details