Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 68855699-68856221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPM
Entrez Gene ID
1368
Gene Name
carboxypeptidase M
Gene Aliases
-
UniGene
Hs.654387
Gene Chromosome Location
Chr.12: 68842200-68963240 on Build GRCh38
Transcripts
NM_001005502.2 NM_001874.4 NM_198320.3

SNP Detail

SNP ID Chromosome Location
rs912078204 Chr.12: 68855729-68855729 on Build GRCh38
rs899271070 Chr.12: 68856149-68856149 on Build GRCh38
rs927864348 Chr.12: 68856122-68856122 on Build GRCh38
rs529820820 Chr.12: 68855873-68855873 on Build GRCh38
rs188236523 Chr.12: 68855763-68855763 on Build GRCh38
rs544211977 Chr.12: 68856155-68856155 on Build GRCh38
rs995348942 Chr.12: 68856178-68856178 on Build GRCh38
rs936386270 Chr.12: 68855751-68855751 on Build GRCh38
rs141805531 Chr.12: 68856069-68856069 on Build GRCh38
rs939587313 Chr.12: 68855739-68855740 on Build GRCh38
rs772460467 Chr.12: 68856096-68856096 on Build GRCh38
rs531716452 Chr.12: 68855765-68855765 on Build GRCh38
rs536074041 Chr.12: 68855998-68855998 on Build GRCh38
rs574866635 Chr.12: 68855743-68855744 on Build GRCh38
rs985111182 Chr.12: 68855860-68855860 on Build GRCh38
rs1012291807 Chr.12: 68855748-68855749 on Build GRCh38
rs1006598961 Chr.12: 68856127-68856127 on Build GRCh38
rs1144947 Chr.12: 68855959-68855959 on Build GRCh38
rs903972215 Chr.12: 68856010-68856010 on Build GRCh38
rs143234420 Chr.12: 68855800-68855800 on Build GRCh38
rs947972496 Chr.12: 68855835-68855835 on Build GRCh38
rs1036488641 Chr.12: 68855875-68855875 on Build GRCh38
rs115882008 Chr.12: 68855901-68855901 on Build GRCh38
rs1046319290 Chr.12: 68855841-68855841 on Build GRCh38
rs371786488 Chr.12: 68856087-68856087 on Build GRCh38
rs151329296 Chr.12: 68855989-68855989 on Build GRCh38
rs1129786 Chr.12: 68855966-68855966 on Build GRCh38
rs895108883 Chr.12: 68855743-68855743 on Build GRCh38
rs576004228 Chr.12: 68856109-68856109 on Build GRCh38
rs545900954 Chr.12: 68855824-68855824 on Build GRCh38
rs549856138 Chr.12: 68855832-68855832 on Build GRCh38
rs899237164 Chr.12: 68855878-68855878 on Build GRCh38
rs775929736 Chr.12: 68856133-68856133 on Build GRCh38
rs188704455 Chr.12: 68856091-68856091 on Build GRCh38
rs181714783 Chr.12: 68856134-68856134 on Build GRCh38
rs994876287 Chr.12: 68855879-68855879 on Build GRCh38
rs537898707 Chr.12: 68855748-68855748 on Build GRCh38
rs1050439885 Chr.12: 68856181-68856181 on Build GRCh38
rs547944925 Chr.12: 68855874-68855874 on Build GRCh38
rs57330829 Chr.12: 68855750-68855751 on Build GRCh38
rs564460939 Chr.12: 68855830-68855830 on Build GRCh38
rs531704564 Chr.12: 68855831-68855831 on Build GRCh38
rs948081565 Chr.12: 68855974-68855974 on Build GRCh38
rs946187336 Chr.12: 68855741-68855741 on Build GRCh38
rs562367791 Chr.12: 68855842-68855842 on Build GRCh38
rs376542148 Chr.12: 68855937-68855937 on Build GRCh38
rs551895112 Chr.12: 68855936-68855936 on Build GRCh38
rs1053851989 Chr.12: 68855796-68855796 on Build GRCh38
rs566673485 Chr.12: 68855900-68855900 on Build GRCh38
rs977987220 Chr.12: 68855981-68855981 on Build GRCh38
rs758423925 Chr.12: 68855887-68855887 on Build GRCh38
rs397938437 Chr.12: 68855756-68855757 on Build GRCh38
rs987621941 Chr.12: 68855725-68855725 on Build GRCh38
rs572655721 Chr.12: 68856071-68856071 on Build GRCh38
rs923645097 Chr.12: 68855749-68855749 on Build GRCh38
rs552534824 Chr.12: 68855882-68855882 on Build GRCh38
rs147093263 Chr.12: 68856095-68856095 on Build GRCh38
rs113194580 Chr.12: 68856147-68856147 on Build GRCh38
rs1000055740 Chr.12: 68855769-68855769 on Build GRCh38
rs150990177 Chr.12: 68856093-68856093 on Build GRCh38
rs1029335617 Chr.12: 68855808-68855808 on Build GRCh38
rs1031908265 Chr.12: 68855774-68855774 on Build GRCh38
rs572666666 Chr.12: 68855802-68855802 on Build GRCh38
rs866585562 Chr.12: 68855850-68855850 on Build GRCh38
rs180943143 Chr.12: 68855793-68855793 on Build GRCh38

View Full Product Details