Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69530883-69531423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRS2
Entrez Gene ID
10818
Gene Name
fibroblast growth factor receptor substrate 2
Gene Aliases
FRS1A, FRS2A, FRS2alpha, SNT, SNT-1, SNT1
UniGene
Hs.593446
Gene Chromosome Location
Chr.12: 69470349-69579793 on Build GRCh38
Transcripts
NM_001042555.2 NM_001278351.1 NM_001278353.1 NM_001278354.1 NM_001278355.1 NM_001278356.1 NM_001278357.1 NM_006654.4 XM_017018717.1 XM_017018718.1

SNP Detail

SNP ID Chromosome Location
rs78851242 Chr.12: 69530977-69530977 on Build GRCh38
rs967825319 Chr.12: 69531343-69531343 on Build GRCh38
rs575280179 Chr.12: 69531198-69531198 on Build GRCh38
rs200147478 Chr.12: 69530978-69530979 on Build GRCh38
rs1046273654 Chr.12: 69531389-69531389 on Build GRCh38
rs544722521 Chr.12: 69531158-69531158 on Build GRCh38
rs371893005 Chr.12: 69531036-69531037 on Build GRCh38
rs1003290195 Chr.12: 69531043-69531046 on Build GRCh38
rs1033321606 Chr.12: 69531355-69531355 on Build GRCh38
rs975081105 Chr.12: 69531258-69531258 on Build GRCh38
rs59475670 Chr.12: 69530978-69530978 on Build GRCh38
rs892281051 Chr.12: 69531297-69531297 on Build GRCh38
rs929519611 Chr.12: 69531155-69531155 on Build GRCh38
rs747906448 Chr.12: 69530923-69530923 on Build GRCh38
rs866422113 Chr.12: 69531391-69531391 on Build GRCh38
rs1009427084 Chr.12: 69531298-69531298 on Build GRCh38
rs562945098 Chr.12: 69531175-69531175 on Build GRCh38
rs11380565 Chr.12: 69531335-69531336 on Build GRCh38
rs757859406 Chr.12: 69531090-69531090 on Build GRCh38
rs868357196 Chr.12: 69531225-69531225 on Build GRCh38
rs765208454 Chr.12: 69530915-69530915 on Build GRCh38
rs397969975 Chr.12: 69531347-69531348 on Build GRCh38
rs992440568 Chr.12: 69531102-69531102 on Build GRCh38
rs1001955852 Chr.12: 69531354-69531354 on Build GRCh38
rs751172949 Chr.12: 69531207-69531207 on Build GRCh38
rs919390758 Chr.12: 69531106-69531106 on Build GRCh38
rs1049591172 Chr.12: 69531156-69531156 on Build GRCh38
rs796102173 Chr.12: 69530994-69530994 on Build GRCh38
rs573509990 Chr.12: 69531052-69531052 on Build GRCh38
rs901856353 Chr.12: 69531201-69531201 on Build GRCh38
rs79668456 Chr.12: 69530981-69530981 on Build GRCh38
rs1038975803 Chr.12: 69531200-69531200 on Build GRCh38
rs1022083178 Chr.12: 69530910-69530910 on Build GRCh38
rs376187895 Chr.12: 69531096-69531096 on Build GRCh38
rs926855500 Chr.12: 69531091-69531091 on Build GRCh38
rs540567992 Chr.12: 69531053-69531053 on Build GRCh38
rs492300 Chr.12: 69531390-69531390 on Build GRCh38
rs565966014 Chr.12: 69531095-69531095 on Build GRCh38
rs1021776583 Chr.12: 69531147-69531147 on Build GRCh38
rs7299779 Chr.12: 69531335-69531335 on Build GRCh38
rs530119596 Chr.12: 69531260-69531260 on Build GRCh38
rs943714920 Chr.12: 69531254-69531254 on Build GRCh38
rs546447753 Chr.12: 69531387-69531387 on Build GRCh38
rs566746933 Chr.12: 69531312-69531312 on Build GRCh38
rs542576248 Chr.12: 69530944-69530944 on Build GRCh38
rs554253236 Chr.12: 69530949-69530949 on Build GRCh38
rs528423762 Chr.12: 69531325-69531325 on Build GRCh38
rs549623 Chr.12: 69531273-69531273 on Build GRCh38
rs79598264 Chr.12: 69531337-69531337 on Build GRCh38
rs907169570 Chr.12: 69531039-69531039 on Build GRCh38
rs749969616 Chr.12: 69531082-69531082 on Build GRCh38

View Full Product Details