Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69653367-69653864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST3
Entrez Gene ID
144453
Gene Name
bestrophin 3
Gene Aliases
VMD2L3
UniGene
Hs.280782
Gene Chromosome Location
Chr.12: 69643508-69699416 on Build GRCh38
Transcripts
NM_001282613.1 NM_001282614.1 NM_032735.2 NM_152439.3 XM_011537961.1 XM_011537962.2 XM_011537963.1 XM_011537965.2

SNP Detail

SNP ID Chromosome Location
rs879188391 Chr.12: 69653837-69653837 on Build GRCh38
rs56834803 Chr.12: 69653672-69653672 on Build GRCh38
rs542332240 Chr.12: 69653450-69653450 on Build GRCh38
rs958864239 Chr.12: 69653599-69653599 on Build GRCh38
rs767780539 Chr.12: 69653466-69653467 on Build GRCh38
rs984548403 Chr.12: 69653491-69653491 on Build GRCh38
rs993330773 Chr.12: 69653610-69653610 on Build GRCh38
rs2161438 Chr.12: 69653582-69653582 on Build GRCh38
rs114166365 Chr.12: 69653836-69653836 on Build GRCh38
rs533949475 Chr.12: 69653687-69653687 on Build GRCh38
rs1034567673 Chr.12: 69653818-69653818 on Build GRCh38
rs528081085 Chr.12: 69653574-69653574 on Build GRCh38
rs1016549709 Chr.12: 69653742-69653742 on Build GRCh38
rs368040461 Chr.12: 69653643-69653643 on Build GRCh38
rs181818149 Chr.12: 69653516-69653516 on Build GRCh38
rs878869900 Chr.12: 69653829-69653829 on Build GRCh38
rs890129995 Chr.12: 69653531-69653531 on Build GRCh38
rs1051966555 Chr.12: 69653761-69653761 on Build GRCh38
rs975831625 Chr.12: 69653467-69653467 on Build GRCh38
rs116335970 Chr.12: 69653614-69653614 on Build GRCh38
rs992120853 Chr.12: 69653828-69653828 on Build GRCh38
rs756919380 Chr.12: 69653838-69653838 on Build GRCh38
rs189480207 Chr.12: 69653730-69653730 on Build GRCh38
rs753282200 Chr.12: 69653594-69653594 on Build GRCh38
rs558898728 Chr.12: 69653710-69653710 on Build GRCh38
rs940636864 Chr.12: 69653505-69653505 on Build GRCh38
rs1044249729 Chr.12: 69653622-69653622 on Build GRCh38
rs757624209 Chr.12: 69653583-69653583 on Build GRCh38
rs1000453909 Chr.12: 69653809-69653809 on Build GRCh38
rs181902118 Chr.12: 69653807-69653807 on Build GRCh38
rs901529482 Chr.12: 69653686-69653686 on Build GRCh38
rs372474135 Chr.12: 69653711-69653711 on Build GRCh38
rs900341500 Chr.12: 69653477-69653477 on Build GRCh38
rs2547016 Chr.12: 69653595-69653595 on Build GRCh38
rs909136667 Chr.12: 69653502-69653502 on Build GRCh38
rs1019942044 Chr.12: 69653572-69653572 on Build GRCh38
rs766749465 Chr.12: 69653460-69653460 on Build GRCh38
rs575021655 Chr.12: 69653422-69653422 on Build GRCh38
rs751222619 Chr.12: 69653778-69653778 on Build GRCh38
rs996330899 Chr.12: 69653522-69653522 on Build GRCh38
rs890233859 Chr.12: 69653764-69653764 on Build GRCh38
rs754096975 Chr.12: 69653528-69653528 on Build GRCh38
rs1010386566 Chr.12: 69653765-69653765 on Build GRCh38
rs187375024 Chr.12: 69653590-69653590 on Build GRCh38
rs953232024 Chr.12: 69653478-69653478 on Build GRCh38
rs546718135 Chr.12: 69653796-69653796 on Build GRCh38
rs760870853 Chr.12: 69653410-69653410 on Build GRCh38
rs965722158 Chr.12: 69653432-69653432 on Build GRCh38
rs370232728 Chr.12: 69653628-69653628 on Build GRCh38
rs1030047187 Chr.12: 69653529-69653529 on Build GRCh38
rs904014620 Chr.12: 69653632-69653632 on Build GRCh38
rs534682899 Chr.12: 69653588-69653588 on Build GRCh38
rs11177773 Chr.12: 69653571-69653571 on Build GRCh38
rs1014402580 Chr.12: 69653841-69653841 on Build GRCh38
rs972421275 Chr.12: 69653775-69653775 on Build GRCh38
rs548413044 Chr.12: 69653640-69653640 on Build GRCh38
rs538522229 Chr.12: 69653719-69653719 on Build GRCh38
rs10906 Chr.12: 69653718-69653718 on Build GRCh38

View Full Product Details