Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69676838-69677351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST3
Entrez Gene ID
144453
Gene Name
bestrophin 3
Gene Aliases
VMD2L3
UniGene
Hs.280782
Gene Chromosome Location
Chr.12: 69643508-69699416 on Build GRCh38
Transcripts
NM_001282613.1 NM_001282614.1 NM_032735.2 NM_152439.3 XM_006719256.3 XM_011537961.1 XM_011537962.2 XM_011537963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5699739 c.830C>T p.T277I Chr.12: 69676953-69676953 on Build GRCh38
4044534 c.647G>A p.R216Q Chr.12: 69677247-69677247 on Build GRCh38
6265679 c.666C>A p.S222R Chr.12: 69677228-69677228 on Build GRCh38
3464635 c.693G>A p.G231G Chr.12: 69677201-69677201 on Build GRCh38
5613607 c.736A>T p.T246S Chr.12: 69677047-69677047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755555260 Chr.12: 69677234-69677234 on Build GRCh38
rs775589749 Chr.12: 69677045-69677045 on Build GRCh38
rs767324127 Chr.12: 69677077-69677077 on Build GRCh38
rs377359402 Chr.12: 69677252-69677252 on Build GRCh38
rs763222464 Chr.12: 69677288-69677288 on Build GRCh38
rs747344001 Chr.12: 69676900-69676900 on Build GRCh38
rs116917484 Chr.12: 69677297-69677297 on Build GRCh38
rs779109477 Chr.12: 69676978-69676978 on Build GRCh38
rs1009357030 Chr.12: 69676944-69676944 on Build GRCh38
rs753563227 Chr.12: 69677062-69677062 on Build GRCh38
rs900649520 Chr.12: 69676958-69676958 on Build GRCh38
rs710718 Chr.12: 69677162-69677162 on Build GRCh38
rs778798425 Chr.12: 69676874-69676874 on Build GRCh38
rs771720622 Chr.12: 69677166-69677166 on Build GRCh38
rs563762545 Chr.12: 69676916-69676916 on Build GRCh38
rs773849380 Chr.12: 69676933-69676935 on Build GRCh38
rs199594674 Chr.12: 69677031-69677031 on Build GRCh38
rs753205163 Chr.12: 69677242-69677242 on Build GRCh38
rs746443148 Chr.12: 69676882-69676882 on Build GRCh38
rs763747167 Chr.12: 69676951-69676951 on Build GRCh38
rs760549863 Chr.12: 69676936-69676936 on Build GRCh38
rs545548955 Chr.12: 69676888-69676888 on Build GRCh38
rs768866074 Chr.12: 69676910-69676910 on Build GRCh38
rs11609972 Chr.12: 69677113-69677113 on Build GRCh38
rs747033835 Chr.12: 69677184-69677184 on Build GRCh38
rs755438978 Chr.12: 69677118-69677118 on Build GRCh38
rs771667441 Chr.12: 69677038-69677038 on Build GRCh38
rs778735903 Chr.12: 69677161-69677161 on Build GRCh38
rs202161560 Chr.12: 69677266-69677266 on Build GRCh38
rs745517606 Chr.12: 69677248-69677248 on Build GRCh38
rs748065485 Chr.12: 69677269-69677269 on Build GRCh38
rs927233694 Chr.12: 69677164-69677164 on Build GRCh38
rs759531399 Chr.12: 69677000-69677000 on Build GRCh38
rs774049665 Chr.12: 69677033-69677033 on Build GRCh38
rs368113693 Chr.12: 69677056-69677056 on Build GRCh38
rs552071746 Chr.12: 69677320-69677320 on Build GRCh38
rs746472418 Chr.12: 69677259-69677259 on Build GRCh38
rs568405722 Chr.12: 69677004-69677004 on Build GRCh38
rs758498539 Chr.12: 69677101-69677101 on Build GRCh38
rs374461067 Chr.12: 69677013-69677013 on Build GRCh38
rs368490759 Chr.12: 69677019-69677019 on Build GRCh38
rs200887965 Chr.12: 69677075-69677075 on Build GRCh38
rs768172565 Chr.12: 69677261-69677261 on Build GRCh38
rs754102676 Chr.12: 69676953-69676953 on Build GRCh38
rs778490693 Chr.12: 69676897-69676897 on Build GRCh38
rs145416134 Chr.12: 69676964-69676964 on Build GRCh38
rs775882500 Chr.12: 69676892-69676892 on Build GRCh38
rs769082184 Chr.12: 69677014-69677014 on Build GRCh38
rs114061225 Chr.12: 69676990-69676990 on Build GRCh38
rs763098809 Chr.12: 69677197-69677197 on Build GRCh38
rs767876879 Chr.12: 69677241-69677241 on Build GRCh38
rs751581328 Chr.12: 69677232-69677232 on Build GRCh38
rs140310153 Chr.12: 69677196-69677196 on Build GRCh38
rs766515260 Chr.12: 69677218-69677218 on Build GRCh38
rs759561779 Chr.12: 69677300-69677300 on Build GRCh38
rs78300740 Chr.12: 69677081-69677081 on Build GRCh38
rs79464531 Chr.12: 69677114-69677114 on Build GRCh38
rs370129964 Chr.12: 69677279-69677279 on Build GRCh38
rs892646497 Chr.12: 69677324-69677324 on Build GRCh38
rs1004584936 Chr.12: 69677008-69677008 on Build GRCh38
rs556546964 Chr.12: 69677243-69677243 on Build GRCh38
rs200668742 Chr.12: 69677020-69677020 on Build GRCh38
rs770481382 Chr.12: 69676914-69676914 on Build GRCh38
rs1027883495 Chr.12: 69676966-69676966 on Build GRCh38
rs752565721 Chr.12: 69677175-69677179 on Build GRCh38
rs752576624 Chr.12: 69676935-69676935 on Build GRCh38
rs17106943 Chr.12: 69677298-69677298 on Build GRCh38
rs780570896 Chr.12: 69676991-69676991 on Build GRCh38
rs201638085 Chr.12: 69676911-69676911 on Build GRCh38
rs372783693 Chr.12: 69677247-69677247 on Build GRCh38
rs946762426 Chr.12: 69676887-69676887 on Build GRCh38
rs767536005 Chr.12: 69676932-69676932 on Build GRCh38
rs748436719 Chr.12: 69677135-69677135 on Build GRCh38
rs760469467 Chr.12: 69677049-69677049 on Build GRCh38
rs145342394 Chr.12: 69677314-69677314 on Build GRCh38
rs763952985 Chr.12: 69677061-69677061 on Build GRCh38
rs776531139 Chr.12: 69677193-69677193 on Build GRCh38
rs779761425 Chr.12: 69677253-69677253 on Build GRCh38
rs374833609 Chr.12: 69677034-69677034 on Build GRCh38
rs768412741 Chr.12: 69677219-69677219 on Build GRCh38
rs759601669 Chr.12: 69676922-69676922 on Build GRCh38
rs745673258 Chr.12: 69676940-69676940 on Build GRCh38
rs757459680 Chr.12: 69676913-69676913 on Build GRCh38
rs755812933 Chr.12: 69677216-69677232 on Build GRCh38
rs1039937279 Chr.12: 69676994-69676994 on Build GRCh38
rs772990158 Chr.12: 69677287-69677287 on Build GRCh38
rs750482090 Chr.12: 69676979-69676979 on Build GRCh38
rs369833223 Chr.12: 69677119-69677119 on Build GRCh38
rs199950482 Chr.12: 69677195-69677195 on Build GRCh38
rs375603707 Chr.12: 69677182-69677182 on Build GRCh38
rs768457172 Chr.12: 69677192-69677192 on Build GRCh38

View Full Product Details