Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 70310754-70311171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT2
Entrez Gene ID
4848
Gene Name
CCR4-NOT transcription complex subunit 2
Gene Aliases
CDC36, HSPC131, NOT2, NOT2H
UniGene
Hs.133350
Gene Chromosome Location
Chr.12: 70242993-70354993 on Build GRCh38
Transcripts
NM_001199302.1 NM_001199303.1 NM_014515.5 NR_037615.1 XM_006719429.3 XM_006719430.3 XM_006719432.3 XM_006719433.3 XM_011538399.1 XM_011538400.2 XM_017019348.1 XM_017019349.1 XM_017019350.1 XM_017019351.1 XM_017019352.1 XM_017019353.1 XM_017019354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5605642 c.169G>A p.D57N Chr.12: 70311015-70311015 on Build GRCh38
1323087 c.97G>T p.V33F Chr.12: 70310943-70310943 on Build GRCh38
3812924 c.77G>T p.R26I Chr.12: 70310923-70310923 on Build GRCh38
6339970 c.113A>G p.H38R Chr.12: 70310959-70310959 on Build GRCh38
4044578 c.139T>C p.S47P Chr.12: 70310985-70310985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752390701 Chr.12: 70310833-70310833 on Build GRCh38
rs138621902 Chr.12: 70310991-70310991 on Build GRCh38
rs761266832 Chr.12: 70310946-70310946 on Build GRCh38
rs192745125 Chr.12: 70310874-70310874 on Build GRCh38
rs374877154 Chr.12: 70311057-70311057 on Build GRCh38
rs751765559 Chr.12: 70310979-70310979 on Build GRCh38
rs903740126 Chr.12: 70311112-70311112 on Build GRCh38
rs768196233 Chr.12: 70310941-70310941 on Build GRCh38
rs952546442 Chr.12: 70310831-70310831 on Build GRCh38
rs982981280 Chr.12: 70310867-70310867 on Build GRCh38
rs780642158 Chr.12: 70311115-70311115 on Build GRCh38
rs376768007 Chr.12: 70310882-70310882 on Build GRCh38
rs755850489 Chr.12: 70310836-70310836 on Build GRCh38
rs762442117 Chr.12: 70311041-70311041 on Build GRCh38
rs371872752 Chr.12: 70311052-70311052 on Build GRCh38
rs868265413 Chr.12: 70311148-70311148 on Build GRCh38
rs554598990 Chr.12: 70311073-70311073 on Build GRCh38
rs770182270 Chr.12: 70311061-70311062 on Build GRCh38
rs781387012 Chr.12: 70310865-70310865 on Build GRCh38
rs965494704 Chr.12: 70310789-70310789 on Build GRCh38
rs537449907 Chr.12: 70311100-70311100 on Build GRCh38
rs1033893512 Chr.12: 70311139-70311139 on Build GRCh38
rs767456270 Chr.12: 70310826-70310826 on Build GRCh38
rs1050869264 Chr.12: 70311003-70311003 on Build GRCh38
rs778209685 Chr.12: 70311049-70311049 on Build GRCh38
rs569088774 Chr.12: 70311040-70311040 on Build GRCh38
rs368572050 Chr.12: 70310963-70310963 on Build GRCh38
rs528219139 Chr.12: 70310832-70310832 on Build GRCh38
rs200035743 Chr.12: 70311058-70311058 on Build GRCh38
rs372247691 Chr.12: 70310863-70310863 on Build GRCh38
rs768985607 Chr.12: 70311140-70311140 on Build GRCh38
rs756708431 Chr.12: 70310873-70310873 on Build GRCh38
rs746653135 Chr.12: 70310927-70310927 on Build GRCh38
rs147120637 Chr.12: 70310973-70310973 on Build GRCh38
rs748195995 Chr.12: 70310870-70310870 on Build GRCh38
rs771321443 Chr.12: 70310888-70310888 on Build GRCh38
rs747555054 Chr.12: 70311121-70311121 on Build GRCh38
rs753403636 Chr.12: 70310970-70310970 on Build GRCh38
rs939678362 Chr.12: 70310860-70310860 on Build GRCh38
rs189356828 Chr.12: 70310850-70310850 on Build GRCh38
rs148064945 Chr.12: 70310829-70310829 on Build GRCh38
rs775063432 Chr.12: 70310904-70310904 on Build GRCh38
rs530445826 Chr.12: 70310799-70310799 on Build GRCh38
rs550624923 Chr.12: 70310984-70310984 on Build GRCh38
rs532301707 Chr.12: 70310945-70310945 on Build GRCh38
rs201352266 Chr.12: 70310861-70310861 on Build GRCh38
rs772954450 Chr.12: 70311123-70311123 on Build GRCh38
rs758158044 Chr.12: 70310847-70310847 on Build GRCh38
rs141549168 Chr.12: 70310959-70310959 on Build GRCh38
rs752836290 Chr.12: 70311030-70311030 on Build GRCh38
rs757611855 Chr.12: 70311053-70311053 on Build GRCh38
rs941637228 Chr.12: 70310886-70310886 on Build GRCh38
rs769740163 Chr.12: 70310954-70310954 on Build GRCh38
rs548531642 Chr.12: 70310828-70310828 on Build GRCh38

View Full Product Details