Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 70335308-70335850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT2
Entrez Gene ID
4848
Gene Name
CCR4-NOT transcription complex subunit 2
Gene Aliases
CDC36, HSPC131, NOT2, NOT2H
UniGene
Hs.133350
Gene Chromosome Location
Chr.12: 70242993-70354993 on Build GRCh38
Transcripts
NM_001199302.1 NM_001199303.1 NM_014515.5 NR_037615.1 XM_006719429.3 XM_006719430.3 XM_006719432.3 XM_006719433.3 XM_011538399.1 XM_011538400.2 XM_017019348.1 XM_017019349.1 XM_017019350.1 XM_017019351.1 XM_017019352.1 XM_017019353.1 XM_017019354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3812925 c.683C>T p.S228L Chr.12: 70335471-70335471 on Build GRCh38
6631438 c.652G>A p.G218R Chr.12: 70335440-70335440 on Build GRCh38
6631446 c.707G>A p.R236Q Chr.12: 70335495-70335495 on Build GRCh38
3731126 c.732C>T p.N244N Chr.12: 70335520-70335520 on Build GRCh38
3725059 c.694G>A p.A232T Chr.12: 70335482-70335482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908760553 Chr.12: 70335767-70335767 on Build GRCh38
rs964814422 Chr.12: 70335652-70335652 on Build GRCh38
rs574586147 Chr.12: 70335610-70335610 on Build GRCh38
rs761647671 Chr.12: 70335648-70335648 on Build GRCh38
rs187301196 Chr.12: 70335806-70335806 on Build GRCh38
rs749026769 Chr.12: 70335345-70335345 on Build GRCh38
rs774116127 Chr.12: 70335613-70335614 on Build GRCh38
rs1003209318 Chr.12: 70335523-70335523 on Build GRCh38
rs568763287 Chr.12: 70335415-70335415 on Build GRCh38
rs966669992 Chr.12: 70335636-70335636 on Build GRCh38
rs989342181 Chr.12: 70335592-70335592 on Build GRCh38
rs377527305 Chr.12: 70335440-70335440 on Build GRCh38
rs776005787 Chr.12: 70335455-70335455 on Build GRCh38
rs921420609 Chr.12: 70335713-70335713 on Build GRCh38
rs755945149 Chr.12: 70335574-70335574 on Build GRCh38
rs987219314 Chr.12: 70335371-70335371 on Build GRCh38
rs1048615056 Chr.12: 70335747-70335747 on Build GRCh38
rs772460902 Chr.12: 70335439-70335439 on Build GRCh38
rs373103481 Chr.12: 70335499-70335499 on Build GRCh38
rs900041337 Chr.12: 70335421-70335421 on Build GRCh38
rs767414377 Chr.12: 70335532-70335532 on Build GRCh38
rs201309251 Chr.12: 70335498-70335498 on Build GRCh38
rs1027546877 Chr.12: 70335693-70335693 on Build GRCh38
rs762668312 Chr.12: 70335495-70335495 on Build GRCh38
rs758131724 Chr.12: 70335422-70335425 on Build GRCh38
rs763856567 Chr.12: 70335587-70335587 on Build GRCh38
rs750239547 Chr.12: 70335390-70335392 on Build GRCh38
rs1046788719 Chr.12: 70335771-70335771 on Build GRCh38
rs561007259 Chr.12: 70335337-70335337 on Build GRCh38
rs148665302 Chr.12: 70335487-70335487 on Build GRCh38
rs147223048 Chr.12: 70335422-70335422 on Build GRCh38
rs778037628 Chr.12: 70335408-70335408 on Build GRCh38
rs747433744 Chr.12: 70335458-70335458 on Build GRCh38
rs961922717 Chr.12: 70335626-70335626 on Build GRCh38
rs769136149 Chr.12: 70335483-70335483 on Build GRCh38
rs565460226 Chr.12: 70335357-70335357 on Build GRCh38
rs906578599 Chr.12: 70335504-70335504 on Build GRCh38
rs911297851 Chr.12: 70335369-70335369 on Build GRCh38
rs1036722227 Chr.12: 70335683-70335683 on Build GRCh38
rs942682806 Chr.12: 70335382-70335382 on Build GRCh38
rs931494904 Chr.12: 70335739-70335739 on Build GRCh38
rs528556957 Chr.12: 70335346-70335346 on Build GRCh38
rs755892278 Chr.12: 70335407-70335407 on Build GRCh38
rs554788354 Chr.12: 70335344-70335344 on Build GRCh38
rs779123769 Chr.12: 70335599-70335599 on Build GRCh38
rs144986592 Chr.12: 70335410-70335410 on Build GRCh38
rs752450857 Chr.12: 70335406-70335406 on Build GRCh38
rs752629512 Chr.12: 70335538-70335538 on Build GRCh38
rs370680853 Chr.12: 70335675-70335675 on Build GRCh38
rs781120437 Chr.12: 70335388-70335388 on Build GRCh38
rs377431152 Chr.12: 70335423-70335423 on Build GRCh38
rs546837598 Chr.12: 70335356-70335356 on Build GRCh38
rs758489572 Chr.12: 70335609-70335609 on Build GRCh38
rs757502017 Chr.12: 70335590-70335590 on Build GRCh38
rs745947810 Chr.12: 70335608-70335608 on Build GRCh38
rs1039855671 Chr.12: 70335419-70335419 on Build GRCh38
rs759033217 Chr.12: 70335508-70335508 on Build GRCh38
rs911607034 Chr.12: 70335381-70335381 on Build GRCh38
rs1052590986 Chr.12: 70335449-70335449 on Build GRCh38
rs73324133 Chr.12: 70335816-70335816 on Build GRCh38
rs961974741 Chr.12: 70335332-70335332 on Build GRCh38
rs951873746 Chr.12: 70335768-70335768 on Build GRCh38
rs764123798 Chr.12: 70335364-70335364 on Build GRCh38
rs532707995 Chr.12: 70335370-70335370 on Build GRCh38
rs753581040 Chr.12: 70335588-70335588 on Build GRCh38
rs987513745 Chr.12: 70335638-70335638 on Build GRCh38

View Full Product Details