Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 70337224-70337771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT2
Entrez Gene ID
4848
Gene Name
CCR4-NOT transcription complex subunit 2
Gene Aliases
CDC36, HSPC131, NOT2, NOT2H
UniGene
Hs.133350
Gene Chromosome Location
Chr.12: 70242993-70354993 on Build GRCh38
Transcripts
NM_001199302.1 NM_001199303.1 NM_014515.5 NR_037615.1 XM_006719429.3 XM_006719430.3 XM_006719432.3 XM_006719433.3 XM_011538399.1 XM_011538400.2 XM_017019348.1 XM_017019349.1 XM_017019350.1 XM_017019351.1 XM_017019352.1 XM_017019353.1 XM_017019354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4673188 c.806A>G p.Q269R Chr.12: 70337419-70337419 on Build GRCh38
6207802 c.864T>A p.Y288* Chr.12: 70337477-70337477 on Build GRCh38
4805884 c.809C>T p.S270F Chr.12: 70337422-70337422 on Build GRCh38
5900568 c.868G>A p.D290N Chr.12: 70337481-70337481 on Build GRCh38
695261 c.869A>C p.D290A Chr.12: 70337482-70337482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748461514 Chr.12: 70337347-70337347 on Build GRCh38
rs879136353 Chr.12: 70337712-70337712 on Build GRCh38
rs754486698 Chr.12: 70337310-70337310 on Build GRCh38
rs10506590 Chr.12: 70337343-70337343 on Build GRCh38
rs751206936 Chr.12: 70337275-70337275 on Build GRCh38
rs753634148 Chr.12: 70337435-70337435 on Build GRCh38
rs932990982 Chr.12: 70337704-70337704 on Build GRCh38
rs966472310 Chr.12: 70337651-70337651 on Build GRCh38
rs754307062 Chr.12: 70337385-70337385 on Build GRCh38
rs556813944 Chr.12: 70337546-70337546 on Build GRCh38
rs763979513 Chr.12: 70337414-70337414 on Build GRCh38
rs751636975 Chr.12: 70337500-70337500 on Build GRCh38
rs760629038 Chr.12: 70337413-70337413 on Build GRCh38
rs745512873 Chr.12: 70337564-70337564 on Build GRCh38
rs1055492314 Chr.12: 70337393-70337393 on Build GRCh38
rs74101497 Chr.12: 70337298-70337298 on Build GRCh38
rs775457376 Chr.12: 70337383-70337383 on Build GRCh38
rs757018617 Chr.12: 70337440-70337440 on Build GRCh38
rs774042073 Chr.12: 70337360-70337360 on Build GRCh38
rs769187626 Chr.12: 70337338-70337338 on Build GRCh38
rs1022425396 Chr.12: 70337565-70337565 on Build GRCh38
rs563137400 Chr.12: 70337291-70337291 on Build GRCh38
rs984983228 Chr.12: 70337476-70337476 on Build GRCh38
rs560729533 Chr.12: 70337685-70337685 on Build GRCh38
rs921751993 Chr.12: 70337290-70337290 on Build GRCh38
rs779203056 Chr.12: 70337652-70337652 on Build GRCh38
rs896729577 Chr.12: 70337358-70337358 on Build GRCh38
rs537935006 Chr.12: 70337531-70337531 on Build GRCh38
rs765433684 Chr.12: 70337473-70337473 on Build GRCh38
rs1012339382 Chr.12: 70337557-70337557 on Build GRCh38
rs1000223762 Chr.12: 70337602-70337602 on Build GRCh38
rs770507441 Chr.12: 70337359-70337359 on Build GRCh38
rs572641166 Chr.12: 70337724-70337724 on Build GRCh38
rs577293161 Chr.12: 70337530-70337530 on Build GRCh38
rs752324339 Chr.12: 70337528-70337528 on Build GRCh38
rs903970464 Chr.12: 70337315-70337315 on Build GRCh38
rs894246645 Chr.12: 70337465-70337465 on Build GRCh38
rs758444384 Chr.12: 70337482-70337482 on Build GRCh38
rs770192961 Chr.12: 70337542-70337542 on Build GRCh38
rs553603414 Chr.12: 70337252-70337252 on Build GRCh38
rs780256665 Chr.12: 70337492-70337492 on Build GRCh38
rs767025113 Chr.12: 70337368-70337368 on Build GRCh38
rs976149544 Chr.12: 70337679-70337679 on Build GRCh38
rs146802441 Chr.12: 70337601-70337601 on Build GRCh38
rs1030991036 Chr.12: 70337637-70337637 on Build GRCh38
rs750652017 Chr.12: 70337474-70337474 on Build GRCh38
rs1016425771 Chr.12: 70337533-70337533 on Build GRCh38
rs1002613041 Chr.12: 70337322-70337322 on Build GRCh38
rs568136154 Chr.12: 70337698-70337698 on Build GRCh38
rs891611852 Chr.12: 70337271-70337271 on Build GRCh38
rs757721881 Chr.12: 70337579-70337579 on Build GRCh38
rs558674730 Chr.12: 70337367-70337367 on Build GRCh38
rs184638104 Chr.12: 70337596-70337596 on Build GRCh38

View Full Product Details