Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 85280154-85280658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX1
Entrez Gene ID
8092
Gene Name
ALX homeobox 1
Gene Aliases
CART1, FND3, HEL23
UniGene
Hs.41683
Gene Chromosome Location
Chr.12: 85280258-85301784 on Build GRCh38
Transcripts
NM_006982.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598952 c.1A>G p.M1V Chr.12: 85280262-85280262 on Build GRCh38
3955266 c.165C>A p.P55P Chr.12: 85280426-85280426 on Build GRCh38
6598951 c.195G>A p.L65L Chr.12: 85280456-85280456 on Build GRCh38
6587594 c.184C>A p.H62N Chr.12: 85280445-85280445 on Build GRCh38
3813142 c.106G>C p.D36H Chr.12: 85280367-85280367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780196140 Chr.12: 85280220-85280220 on Build GRCh38
rs1048636970 Chr.12: 85280448-85280448 on Build GRCh38
rs776648954 Chr.12: 85280490-85280491 on Build GRCh38
rs746804639 Chr.12: 85280495-85280495 on Build GRCh38
rs370968616 Chr.12: 85280234-85280234 on Build GRCh38
rs746407374 Chr.12: 85280212-85280212 on Build GRCh38
rs559221908 Chr.12: 85280586-85280586 on Build GRCh38
rs750883922 Chr.12: 85280399-85280399 on Build GRCh38
rs760099352 Chr.12: 85280261-85280261 on Build GRCh38
rs757462532 Chr.12: 85280494-85280494 on Build GRCh38
rs915407375 Chr.12: 85280618-85280618 on Build GRCh38
rs963285579 Chr.12: 85280201-85280201 on Build GRCh38
rs372544988 Chr.12: 85280370-85280370 on Build GRCh38
rs981808018 Chr.12: 85280245-85280245 on Build GRCh38
rs761123701 Chr.12: 85280529-85280529 on Build GRCh38
rs774149369 Chr.12: 85280362-85280362 on Build GRCh38
rs375884213 Chr.12: 85280524-85280524 on Build GRCh38
rs575941794 Chr.12: 85280515-85280515 on Build GRCh38
rs369826150 Chr.12: 85280375-85280375 on Build GRCh38
rs756389093 Chr.12: 85280422-85280422 on Build GRCh38
rs775308419 Chr.12: 85280432-85280432 on Build GRCh38
rs748172443 Chr.12: 85280413-85280413 on Build GRCh38
rs762520703 Chr.12: 85280512-85280512 on Build GRCh38
rs777754230 Chr.12: 85280472-85280472 on Build GRCh38
rs986158806 Chr.12: 85280395-85280395 on Build GRCh38
rs764192292 Chr.12: 85280530-85280530 on Build GRCh38
rs759945013 Chr.12: 85280516-85280516 on Build GRCh38
rs376329142 Chr.12: 85280373-85280373 on Build GRCh38
rs752310288 Chr.12: 85280258-85280258 on Build GRCh38
rs549640285 Chr.12: 85280196-85280196 on Build GRCh38
rs548908336 Chr.12: 85280180-85280180 on Build GRCh38
rs776338410 Chr.12: 85280438-85280438 on Build GRCh38
rs768003680 Chr.12: 85280465-85280465 on Build GRCh38
rs375614518 Chr.12: 85280426-85280426 on Build GRCh38
rs764969720 Chr.12: 85280398-85280398 on Build GRCh38
rs770562894 Chr.12: 85280514-85280514 on Build GRCh38
rs368906149 Chr.12: 85280361-85280361 on Build GRCh38
rs776683842 Chr.12: 85280229-85280229 on Build GRCh38
rs768593991 Chr.12: 85280316-85280316 on Build GRCh38
rs753895342 Chr.12: 85280534-85280534 on Build GRCh38
rs779423593 Chr.12: 85280291-85280291 on Build GRCh38
rs755474505 Chr.12: 85280411-85280411 on Build GRCh38
rs376214478 Chr.12: 85280287-85280287 on Build GRCh38
rs751887592 Chr.12: 85280457-85280457 on Build GRCh38
rs763849212 Chr.12: 85280276-85280276 on Build GRCh38
rs138946669 Chr.12: 85280433-85280433 on Build GRCh38
rs150692244 Chr.12: 85280410-85280410 on Build GRCh38
rs916103290 Chr.12: 85280619-85280619 on Build GRCh38
rs772806072 Chr.12: 85280510-85280510 on Build GRCh38
rs781138367 Chr.12: 85280412-85280412 on Build GRCh38
rs370293266 Chr.12: 85280285-85280285 on Build GRCh38
rs567417111 Chr.12: 85280181-85280181 on Build GRCh38
rs760284048 Chr.12: 85280372-85280372 on Build GRCh38
rs970386725 Chr.12: 85280535-85280535 on Build GRCh38
rs116409037 Chr.12: 85280309-85280309 on Build GRCh38
rs766955975 Chr.12: 85280409-85280409 on Build GRCh38
rs989648503 Chr.12: 85280613-85280613 on Build GRCh38
rs772972571 Chr.12: 85280447-85280447 on Build GRCh38
rs527881274 Chr.12: 85280628-85280628 on Build GRCh38
rs781280416 Chr.12: 85280347-85280347 on Build GRCh38
rs564559514 Chr.12: 85280552-85280552 on Build GRCh38
rs866146541 Chr.12: 85280605-85280605 on Build GRCh38
rs776678208 Chr.12: 85280502-85280502 on Build GRCh38
rs1015342746 Chr.12: 85280305-85280305 on Build GRCh38
rs532007095 Chr.12: 85280566-85280566 on Build GRCh38
rs1004003622 Chr.12: 85280281-85280281 on Build GRCh38
rs115596276 Chr.12: 85280452-85280452 on Build GRCh38
rs746323343 Chr.12: 85280544-85280544 on Build GRCh38
rs769939946 Chr.12: 85280353-85280353 on Build GRCh38
rs141968055 Chr.12: 85280408-85280408 on Build GRCh38
rs748403633 Chr.12: 85280352-85280352 on Build GRCh38
rs759478419 Chr.12: 85280247-85280247 on Build GRCh38
rs903402171 Chr.12: 85280189-85280189 on Build GRCh38
rs780416730 Chr.12: 85280299-85280299 on Build GRCh38
rs561233322 Chr.12: 85280461-85280461 on Build GRCh38
rs368794193 Chr.12: 85280246-85280246 on Build GRCh38
rs776213513 Chr.12: 85280385-85280385 on Build GRCh38
rs368289293 Chr.12: 85280427-85280427 on Build GRCh38
rs947442823 Chr.12: 85280184-85280184 on Build GRCh38
rs201314651 Chr.12: 85280341-85280341 on Build GRCh38
rs202073009 Chr.12: 85280214-85280214 on Build GRCh38
rs750759166 Chr.12: 85280292-85280292 on Build GRCh38
rs754396020 Chr.12: 85280187-85280188 on Build GRCh38
rs547073411 Chr.12: 85280582-85280582 on Build GRCh38
rs145944049 Chr.12: 85280451-85280451 on Build GRCh38
rs914521573 Chr.12: 85280407-85280407 on Build GRCh38
rs981716042 Chr.12: 85280541-85280541 on Build GRCh38
rs778019022 Chr.12: 85280486-85280486 on Build GRCh38
rs142324538 Chr.12: 85280243-85280243 on Build GRCh38
rs768579174 Chr.12: 85280434-85280434 on Build GRCh38
rs767303774 Chr.12: 85280255-85280255 on Build GRCh38
rs148136812 Chr.12: 85280386-85280386 on Build GRCh38
rs373333417 Chr.12: 85280518-85280518 on Build GRCh38
rs115440539 Chr.12: 85280443-85280443 on Build GRCh38
rs747042802 Chr.12: 85280221-85280221 on Build GRCh38
rs761403611 Chr.12: 85280441-85280441 on Build GRCh38
rs868556823 Chr.12: 85280205-85280205 on Build GRCh38
rs1057301365 Chr.12: 85280591-85280591 on Build GRCh38
rs1035967247 Chr.12: 85280207-85280207 on Build GRCh38
rs749536472 Chr.12: 85280487-85280487 on Build GRCh38
rs775260906 Chr.12: 85280371-85280371 on Build GRCh38
rs759032123 Chr.12: 85280298-85280298 on Build GRCh38
rs568150157 Chr.12: 85280581-85280582 on Build GRCh38
rs752844254 Chr.12: 85280471-85280471 on Build GRCh38
rs779440782 Chr.12: 85280509-85280509 on Build GRCh38
rs193089444 Chr.12: 85280226-85280226 on Build GRCh38

View Full Product Details