Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 88043511-88043988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf29
Entrez Gene ID
91298
Gene Name
chromosome 12 open reading frame 29
Gene Aliases
-
UniGene
Hs.591009
Gene Chromosome Location
Chr.12: 88035491-88050160 on Build GRCh38
Transcripts
NM_001009894.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6445829 c.325G>A p.E109K Chr.12: 88043619-88043619 on Build GRCh38
5416647 c.369G>C p.E123D Chr.12: 88043663-88043663 on Build GRCh38
6265683 c.400G>T p.D134Y Chr.12: 88043694-88043694 on Build GRCh38
6614158 c.363A>G p.A121A Chr.12: 88043657-88043657 on Build GRCh38
6488466 c.410G>A p.G137E Chr.12: 88043704-88043704 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776308926 Chr.12: 88043586-88043586 on Build GRCh38
rs1047996506 Chr.12: 88043906-88043906 on Build GRCh38
rs777981140 Chr.12: 88043702-88043702 on Build GRCh38
rs990927686 Chr.12: 88043847-88043847 on Build GRCh38
rs886619738 Chr.12: 88043936-88043936 on Build GRCh38
rs1020770802 Chr.12: 88043669-88043669 on Build GRCh38
rs375590917 Chr.12: 88043574-88043574 on Build GRCh38
rs566366134 Chr.12: 88043874-88043874 on Build GRCh38
rs74511812 Chr.12: 88043607-88043607 on Build GRCh38
rs375953239 Chr.12: 88043746-88043746 on Build GRCh38
rs368205004 Chr.12: 88043727-88043727 on Build GRCh38
rs139590799 Chr.12: 88043613-88043613 on Build GRCh38
rs966485513 Chr.12: 88043693-88043693 on Build GRCh38
rs746557782 Chr.12: 88043700-88043701 on Build GRCh38
rs771905453 Chr.12: 88043549-88043549 on Build GRCh38
rs773463842 Chr.12: 88043612-88043612 on Build GRCh38
rs780549339 Chr.12: 88043762-88043763 on Build GRCh38
rs375123507 Chr.12: 88043748-88043748 on Build GRCh38
rs779999230 Chr.12: 88043568-88043568 on Build GRCh38
rs1049044376 Chr.12: 88043952-88043952 on Build GRCh38
rs766827837 Chr.12: 88043624-88043624 on Build GRCh38
rs779907567 Chr.12: 88043751-88043751 on Build GRCh38
rs111667577 Chr.12: 88043938-88043938 on Build GRCh38
rs546570897 Chr.12: 88043801-88043801 on Build GRCh38
rs370459592 Chr.12: 88043754-88043754 on Build GRCh38
rs779611287 Chr.12: 88043600-88043601 on Build GRCh38
rs767699571 Chr.12: 88043677-88043677 on Build GRCh38
rs756413980 Chr.12: 88043687-88043687 on Build GRCh38
rs1029348506 Chr.12: 88043630-88043630 on Build GRCh38
rs950190855 Chr.12: 88043888-88043888 on Build GRCh38
rs1010447536 Chr.12: 88043656-88043656 on Build GRCh38
rs913037056 Chr.12: 88043876-88043876 on Build GRCh38
rs199508607 Chr.12: 88043623-88043623 on Build GRCh38
rs144278479 Chr.12: 88043638-88043638 on Build GRCh38
rs867810227 Chr.12: 88043550-88043550 on Build GRCh38
rs761454607 Chr.12: 88043591-88043591 on Build GRCh38
rs758056302 Chr.12: 88043582-88043584 on Build GRCh38
rs753026398 Chr.12: 88043685-88043685 on Build GRCh38
rs112835312 Chr.12: 88043569-88043569 on Build GRCh38
rs564208657 Chr.12: 88043592-88043592 on Build GRCh38
rs878916734 Chr.12: 88043562-88043562 on Build GRCh38
rs957506057 Chr.12: 88043763-88043763 on Build GRCh38
rs927315924 Chr.12: 88043941-88043941 on Build GRCh38
rs961179330 Chr.12: 88043540-88043540 on Build GRCh38
rs937308991 Chr.12: 88043948-88043948 on Build GRCh38
rs267603710 Chr.12: 88043639-88043639 on Build GRCh38
rs768166650 Chr.12: 88043718-88043720 on Build GRCh38
rs756241301 Chr.12: 88043920-88043920 on Build GRCh38
rs937702396 Chr.12: 88043824-88043824 on Build GRCh38

View Full Product Details