Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 88048628-88049129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf29
Entrez Gene ID
91298
Gene Name
chromosome 12 open reading frame 29
Gene Aliases
-
UniGene
Hs.591009
Gene Chromosome Location
Chr.12: 88035491-88050160 on Build GRCh38
Transcripts
NM_001009894.2 NM_025114.3 XM_011538756.2 XM_011538757.2 XM_011538758.2 XM_011538759.2 XM_011538760.1 XM_011538761.1 XM_011538762.2 XM_011538763.2 XM_011538766.2 XM_017019980.1 XM_017019981.1 XM_017019983.1

Gene Symbol
CEP290
Entrez Gene ID
80184
Gene Name
centrosomal protein 290
Gene Aliases
3H11Ag, BBS14, CT87, JBTS5, LCA10, MKS4, NPHP6, POC3, SLSN6, rd16
UniGene
Hs.150444
Gene Chromosome Location
Chr.12: 88049013-88142216 on Build GRCh38
Transcripts
NM_001009894.2 NM_025114.3 XM_011538756.2 XM_011538757.2 XM_011538758.2 XM_011538759.2 XM_011538760.1 XM_011538761.1 XM_011538762.2 XM_011538763.2 XM_011538766.2 XM_017019980.1 XM_017019981.1 XM_017019983.1

SNP Detail

SNP ID Chromosome Location
rs919854612 Chr.12: 88048760-88048760 on Build GRCh38
rs917054146 Chr.12: 88048783-88048783 on Build GRCh38
rs371409424 Chr.12: 88048777-88048777 on Build GRCh38
rs929758721 Chr.12: 88049056-88049056 on Build GRCh38
rs1047227678 Chr.12: 88048805-88048805 on Build GRCh38
rs546314965 Chr.12: 88048962-88048962 on Build GRCh38
rs1029859737 Chr.12: 88048881-88048881 on Build GRCh38
rs917105368 Chr.12: 88049042-88049042 on Build GRCh38
rs559836257 Chr.12: 88048985-88048985 on Build GRCh38
rs998375451 Chr.12: 88048870-88048870 on Build GRCh38
rs182200948 Chr.12: 88048752-88048752 on Build GRCh38
rs888122889 Chr.12: 88048836-88048836 on Build GRCh38
rs909671522 Chr.12: 88049098-88049098 on Build GRCh38
rs528583149 Chr.12: 88049015-88049015 on Build GRCh38
rs753974685 Chr.12: 88048697-88048699 on Build GRCh38
rs1033661984 Chr.12: 88049017-88049019 on Build GRCh38
rs879240697 Chr.12: 88049069-88049069 on Build GRCh38
rs982562258 Chr.12: 88049074-88049074 on Build GRCh38
rs201340582 Chr.12: 88048731-88048732 on Build GRCh38
rs142295832 Chr.12: 88048900-88048900 on Build GRCh38
rs368073560 Chr.12: 88049036-88049040 on Build GRCh38
rs929753521 Chr.12: 88048791-88048791 on Build GRCh38
rs77752413 Chr.12: 88048717-88048717 on Build GRCh38
rs138991901 Chr.12: 88048763-88048763 on Build GRCh38
rs1039339291 Chr.12: 88048854-88048854 on Build GRCh38
rs376016171 Chr.12: 88049016-88049016 on Build GRCh38
rs746495319 Chr.12: 88048832-88048832 on Build GRCh38
rs561964994 Chr.12: 88049061-88049061 on Build GRCh38
rs937239024 Chr.12: 88048683-88048683 on Build GRCh38
rs886049876 Chr.12: 88049038-88049041 on Build GRCh38
rs771476327 Chr.12: 88049014-88049014 on Build GRCh38
rs574755919 Chr.12: 88048838-88048838 on Build GRCh38
rs968115899 Chr.12: 88048943-88048943 on Build GRCh38
rs1009674440 Chr.12: 88048912-88048912 on Build GRCh38
rs187923463 Chr.12: 88048867-88048867 on Build GRCh38
rs146362418 Chr.12: 88048927-88048927 on Build GRCh38
rs993123845 Chr.12: 88049040-88049040 on Build GRCh38

View Full Product Details