Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 88049396-88049901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf29
Entrez Gene ID
91298
Gene Name
chromosome 12 open reading frame 29
Gene Aliases
-
UniGene
Hs.591009
Gene Chromosome Location
Chr.12: 88035491-88050160 on Build GRCh38
Transcripts
NM_001009894.2 NM_025114.3 XM_011538756.2 XM_011538757.2 XM_011538758.2 XM_011538759.2 XM_011538760.1 XM_011538761.1 XM_011538762.2 XM_011538763.2 XM_011538766.2 XM_017019980.1 XM_017019981.1 XM_017019983.1

Gene Symbol
CEP290
Entrez Gene ID
80184
Gene Name
centrosomal protein 290
Gene Aliases
3H11Ag, BBS14, CT87, JBTS5, LCA10, MKS4, NPHP6, POC3, SLSN6, rd16
UniGene
Hs.150444
Gene Chromosome Location
Chr.12: 88049013-88142216 on Build GRCh38
Transcripts
NM_001009894.2 NM_025114.3 XM_011538756.2 XM_011538757.2 XM_011538758.2 XM_011538759.2 XM_011538760.1 XM_011538761.1 XM_011538762.2 XM_011538763.2 XM_011538766.2 XM_017019980.1 XM_017019981.1 XM_017019983.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6197750 c.7216-7C>T p.? Chr.12: 88049421-88049421 on Build GRCh38
6197751 c.7210-7C>T p.? Chr.12: 88049421-88049421 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs112178328 Chr.12: 88049570-88049570 on Build GRCh38
rs753383814 Chr.12: 88049427-88049432 on Build GRCh38
rs1026313896 Chr.12: 88049587-88049587 on Build GRCh38
rs751890225 Chr.12: 88049462-88049462 on Build GRCh38
rs761577764 Chr.12: 88049575-88049575 on Build GRCh38
rs557337043 Chr.12: 88049534-88049534 on Build GRCh38
rs773421938 Chr.12: 88049808-88049808 on Build GRCh38
rs962495876 Chr.12: 88049696-88049696 on Build GRCh38
rs914178008 Chr.12: 88049495-88049495 on Build GRCh38
rs769991891 Chr.12: 88049673-88049673 on Build GRCh38
rs373069036 Chr.12: 88049467-88049467 on Build GRCh38
rs866667812 Chr.12: 88049540-88049540 on Build GRCh38
rs573262714 Chr.12: 88049666-88049666 on Build GRCh38
rs192835210 Chr.12: 88049564-88049564 on Build GRCh38
rs986895282 Chr.12: 88049736-88049736 on Build GRCh38
rs188748213 Chr.12: 88049551-88049551 on Build GRCh38
rs925443420 Chr.12: 88049834-88049834 on Build GRCh38
rs1055173296 Chr.12: 88049569-88049569 on Build GRCh38
rs868648519 Chr.12: 88049631-88049631 on Build GRCh38
rs1043975780 Chr.12: 88049827-88049827 on Build GRCh38
rs552892778 Chr.12: 88049652-88049652 on Build GRCh38
rs45613639 Chr.12: 88049436-88049436 on Build GRCh38
rs762987255 Chr.12: 88049430-88049430 on Build GRCh38
rs1014039465 Chr.12: 88049584-88049587 on Build GRCh38
rs975135594 Chr.12: 88049711-88049711 on Build GRCh38
rs748744450 Chr.12: 88049523-88049523 on Build GRCh38
rs1043946724 Chr.12: 88049539-88049539 on Build GRCh38
rs562100795 Chr.12: 88049722-88049722 on Build GRCh38
rs78391352 Chr.12: 88049717-88049717 on Build GRCh38
rs183076076 Chr.12: 88049496-88049496 on Build GRCh38
rs951401140 Chr.12: 88049627-88049631 on Build GRCh38
rs1004123187 Chr.12: 88049632-88049632 on Build GRCh38
rs920949297 Chr.12: 88049462-88049462 on Build GRCh38
rs945630753 Chr.12: 88049503-88049503 on Build GRCh38
rs548615399 Chr.12: 88049761-88049761 on Build GRCh38
rs975103980 Chr.12: 88049451-88049452 on Build GRCh38
rs933614087 Chr.12: 88049464-88049464 on Build GRCh38
rs145452070 Chr.12: 88049811-88049812 on Build GRCh38
rs945630027 Chr.12: 88049821-88049821 on Build GRCh38

View Full Product Details