Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89351548-89352043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP6
Entrez Gene ID
1848
Gene Name
dual specificity phosphatase 6
Gene Aliases
HH19, MKP3, PYST1
UniGene
Hs.298654
Gene Chromosome Location
Chr.12: 89347825-89352859 on Build GRCh38
Transcripts
NM_001946.3 NM_022652.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7187025 c.343C>T p.L115F Chr.12: 89351697-89351697 on Build GRCh38
5695539 c.351G>A p.L117L Chr.12: 89351689-89351689 on Build GRCh38
5611256 c.346_347GG>AA p.G116K Chr.12: 89351693-89351694 on Build GRCh38
3753523 c.340G>T p.V114L Chr.12: 89351700-89351700 on Build GRCh38
7365676 c.383G>C p.R128P Chr.12: 89351657-89351657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587776978 Chr.12: 89351811-89351811 on Build GRCh38
rs779578737 Chr.12: 89351578-89351578 on Build GRCh38
rs558694866 Chr.12: 89351749-89351749 on Build GRCh38
rs144083822 Chr.12: 89351893-89351893 on Build GRCh38
rs370311640 Chr.12: 89351761-89351761 on Build GRCh38
rs759892905 Chr.12: 89351883-89351883 on Build GRCh38
rs764950348 Chr.12: 89351755-89351755 on Build GRCh38
rs774213055 Chr.12: 89351941-89351941 on Build GRCh38
rs148576220 Chr.12: 89351989-89351989 on Build GRCh38
rs754016535 Chr.12: 89351896-89351896 on Build GRCh38
rs200020030 Chr.12: 89351695-89351695 on Build GRCh38
rs774021182 Chr.12: 89351875-89351875 on Build GRCh38
rs999990772 Chr.12: 89351644-89351644 on Build GRCh38
rs755252865 Chr.12: 89351612-89351613 on Build GRCh38
rs778040558 Chr.12: 89351793-89351793 on Build GRCh38
rs769538456 Chr.12: 89351835-89351835 on Build GRCh38
rs572067782 Chr.12: 89351752-89351752 on Build GRCh38
rs146480840 Chr.12: 89351940-89351940 on Build GRCh38
rs755373899 Chr.12: 89351668-89351668 on Build GRCh38
rs866581285 Chr.12: 89351627-89351627 on Build GRCh38
rs748901060 Chr.12: 89351929-89351929 on Build GRCh38
rs1055835044 Chr.12: 89351874-89351874 on Build GRCh38
rs768291739 Chr.12: 89351744-89351744 on Build GRCh38
rs752643011 Chr.12: 89352005-89352005 on Build GRCh38
rs1048553876 Chr.12: 89351584-89351584 on Build GRCh38
rs527433314 Chr.12: 89351812-89351812 on Build GRCh38
rs201138185 Chr.12: 89352019-89352019 on Build GRCh38
rs1024180074 Chr.12: 89351602-89351602 on Build GRCh38
rs746895147 Chr.12: 89351823-89351823 on Build GRCh38
rs767725122 Chr.12: 89351787-89351787 on Build GRCh38
rs756696501 Chr.12: 89351592-89351592 on Build GRCh38
rs753109290 Chr.12: 89351788-89351788 on Build GRCh38
rs780038667 Chr.12: 89351997-89351997 on Build GRCh38
rs757293671 Chr.12: 89351806-89351806 on Build GRCh38
rs542439690 Chr.12: 89351913-89351913 on Build GRCh38
rs777646559 Chr.12: 89351713-89351713 on Build GRCh38
rs952749852 Chr.12: 89351740-89351740 on Build GRCh38
rs1041734103 Chr.12: 89351768-89351768 on Build GRCh38
rs376038752 Chr.12: 89351658-89351658 on Build GRCh38
rs776032408 Chr.12: 89351953-89351953 on Build GRCh38
rs748087431 Chr.12: 89351920-89351920 on Build GRCh38
rs753582413 Chr.12: 89351591-89351591 on Build GRCh38
rs192081420 Chr.12: 89351952-89351952 on Build GRCh38
rs772978587 Chr.12: 89351851-89351851 on Build GRCh38
rs752082427 Chr.12: 89351778-89351778 on Build GRCh38
rs936222145 Chr.12: 89351871-89351871 on Build GRCh38
rs778684685 Chr.12: 89352020-89352020 on Build GRCh38
rs780512205 Chr.12: 89351601-89351601 on Build GRCh38
rs868768607 Chr.12: 89351647-89351647 on Build GRCh38
rs749090170 Chr.12: 89352013-89352013 on Build GRCh38
rs771054626 Chr.12: 89351730-89351730 on Build GRCh38
rs984302574 Chr.12: 89351742-89351742 on Build GRCh38
rs778521021 Chr.12: 89351595-89351595 on Build GRCh38
rs781650709 Chr.12: 89351670-89351670 on Build GRCh38
rs766038388 Chr.12: 89351770-89351770 on Build GRCh38
rs764390682 Chr.12: 89351961-89351961 on Build GRCh38
rs762737360 Chr.12: 89351859-89351859 on Build GRCh38
rs367702814 Chr.12: 89351633-89351633 on Build GRCh38
rs541128548 Chr.12: 89351758-89351758 on Build GRCh38
rs764598740 Chr.12: 89351640-89351640 on Build GRCh38
rs777743665 Chr.12: 89351925-89351925 on Build GRCh38
rs753398886 Chr.12: 89351636-89351636 on Build GRCh38
rs748279152 Chr.12: 89351686-89351686 on Build GRCh38
rs1014088065 Chr.12: 89351978-89351978 on Build GRCh38
rs775471646 Chr.12: 89351632-89351632 on Build GRCh38
rs955003812 Chr.12: 89352017-89352017 on Build GRCh38
rs955857289 Chr.12: 89351680-89351680 on Build GRCh38
rs61734372 Chr.12: 89351780-89351780 on Build GRCh38
rs2279574 Chr.12: 89351700-89351700 on Build GRCh38
rs76842227 Chr.12: 89351615-89351615 on Build GRCh38
rs752172017 Chr.12: 89351660-89351660 on Build GRCh38
rs773372664 Chr.12: 89351621-89351621 on Build GRCh38
rs139215127 Chr.12: 89351825-89351825 on Build GRCh38
rs750510747 Chr.12: 89351911-89351911 on Build GRCh38
rs757489434 Chr.12: 89351908-89351908 on Build GRCh38
rs141049010 Chr.12: 89351946-89351946 on Build GRCh38
rs533891390 Chr.12: 89351618-89351618 on Build GRCh38
rs766301392 Chr.12: 89352016-89352016 on Build GRCh38
rs1041286983 Chr.12: 89351610-89351610 on Build GRCh38
rs756143915 Chr.12: 89352008-89352008 on Build GRCh38
rs749364594 Chr.12: 89351722-89351722 on Build GRCh38
rs758485593 Chr.12: 89351996-89351996 on Build GRCh38
rs768064247 Chr.12: 89351885-89351885 on Build GRCh38
rs1014339213 Chr.12: 89351600-89351600 on Build GRCh38
rs187310303 Chr.12: 89351646-89351646 on Build GRCh38
rs926696619 Chr.12: 89351739-89351739 on Build GRCh38
rs778903834 Chr.12: 89351910-89351910 on Build GRCh38
rs182761061 Chr.12: 89351575-89351575 on Build GRCh38
rs753200673 Chr.12: 89351886-89351886 on Build GRCh38
rs746986341 Chr.12: 89351743-89351743 on Build GRCh38
rs926176218 Chr.12: 89351857-89351857 on Build GRCh38
rs750382042 Chr.12: 89351570-89351570 on Build GRCh38
rs756502676 Chr.12: 89351792-89351792 on Build GRCh38
rs562832283 Chr.12: 89351965-89351965 on Build GRCh38
rs776225487 Chr.12: 89351826-89351826 on Build GRCh38
rs758911284 Chr.12: 89351654-89351654 on Build GRCh38
rs1027986241 Chr.12: 89352015-89352015 on Build GRCh38
rs761953520 Chr.12: 89351968-89351968 on Build GRCh38
rs747973701 Chr.12: 89351830-89351830 on Build GRCh38
rs756477775 Chr.12: 89351888-89351888 on Build GRCh38
rs747386398 Chr.12: 89351603-89351603 on Build GRCh38
rs200947366 Chr.12: 89351599-89351599 on Build GRCh38
rs377057060 Chr.12: 89351628-89351628 on Build GRCh38
rs765947766 Chr.12: 89351866-89351866 on Build GRCh38
rs765444393 Chr.12: 89351971-89351971 on Build GRCh38
rs750850768 Chr.12: 89351598-89351598 on Build GRCh38
rs759410967 Chr.12: 89351626-89351626 on Build GRCh38
rs749559072 Chr.12: 89351797-89351797 on Build GRCh38
rs999928543 Chr.12: 89351904-89351904 on Build GRCh38
rs375201922 Chr.12: 89352010-89352010 on Build GRCh38
rs971168899 Chr.12: 89351719-89351719 on Build GRCh38
rs372428608 Chr.12: 89351634-89351634 on Build GRCh38
rs112255849 Chr.12: 89351609-89351609 on Build GRCh38
rs777653842 Chr.12: 89352012-89352012 on Build GRCh38
rs769601359 Chr.12: 89351922-89351922 on Build GRCh38
rs1052782471 Chr.12: 89351917-89351917 on Build GRCh38
rs748453854 Chr.12: 89351616-89351616 on Build GRCh38
rs761555413 Chr.12: 89351637-89351637 on Build GRCh38
rs908813384 Chr.12: 89351587-89351587 on Build GRCh38

View Full Product Details