Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 90972484-90972980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPYC
Entrez Gene ID
1833
Gene Name
epiphycan
Gene Aliases
DSPG3, PGLB, Pg-Lb, SLRR3B
UniGene
Hs.435680
Gene Chromosome Location
Chr.12: 90963679-91005026 on Build GRCh38
Transcripts
NM_004950.4 XM_011538008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
695469 c.410C>A p.A137D Chr.12: 90972911-90972911 on Build GRCh38
4839043 c.400G>T p.E134* Chr.12: 90972921-90972921 on Build GRCh38
2044897 c.355C>A p.L119I Chr.12: 90972966-90972966 on Build GRCh38
2044895 c.402A>G p.E134E Chr.12: 90972919-90972919 on Build GRCh38
6655637 c.482delA p.N161fs*6 Chr.12: 90972839-90972839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373463291 Chr.12: 90972879-90972879 on Build GRCh38
rs773105055 Chr.12: 90972600-90972600 on Build GRCh38
rs183846621 Chr.12: 90972914-90972914 on Build GRCh38
rs766917163 Chr.12: 90972818-90972818 on Build GRCh38
rs777788937 Chr.12: 90972847-90972847 on Build GRCh38
rs185118507 Chr.12: 90972552-90972552 on Build GRCh38
rs765893904 Chr.12: 90972777-90972777 on Build GRCh38
rs761511520 Chr.12: 90972885-90972885 on Build GRCh38
rs912216974 Chr.12: 90972700-90972700 on Build GRCh38
rs778902385 Chr.12: 90972942-90972942 on Build GRCh38
rs952902062 Chr.12: 90972837-90972837 on Build GRCh38
rs115968659 Chr.12: 90972892-90972892 on Build GRCh38
rs769830387 Chr.12: 90972781-90972781 on Build GRCh38
rs111936337 Chr.12: 90972784-90972784 on Build GRCh38
rs140043768 Chr.12: 90972906-90972906 on Build GRCh38
rs190547958 Chr.12: 90972902-90972902 on Build GRCh38
rs751906525 Chr.12: 90972562-90972562 on Build GRCh38
rs1028102388 Chr.12: 90972604-90972604 on Build GRCh38
rs761599491 Chr.12: 90972802-90972802 on Build GRCh38
rs781447797 Chr.12: 90972827-90972827 on Build GRCh38
rs1008571346 Chr.12: 90972937-90972937 on Build GRCh38
rs931746486 Chr.12: 90972663-90972663 on Build GRCh38
rs766987779 Chr.12: 90972950-90972950 on Build GRCh38
rs895306063 Chr.12: 90972755-90972755 on Build GRCh38
rs377287806 Chr.12: 90972782-90972782 on Build GRCh38
rs114231682 Chr.12: 90972870-90972870 on Build GRCh38
rs189927191 Chr.12: 90972800-90972800 on Build GRCh38
rs975744525 Chr.12: 90972642-90972642 on Build GRCh38
rs756262038 Chr.12: 90972934-90972934 on Build GRCh38
rs756349998 Chr.12: 90972844-90972844 on Build GRCh38
rs777983818 Chr.12: 90972939-90972939 on Build GRCh38
rs936386070 Chr.12: 90972742-90972742 on Build GRCh38
rs1050159224 Chr.12: 90972672-90972672 on Build GRCh38
rs148048305 Chr.12: 90972595-90972595 on Build GRCh38
rs752891135 Chr.12: 90972831-90972831 on Build GRCh38
rs745754970 Chr.12: 90972792-90972792 on Build GRCh38
rs150301480 Chr.12: 90972910-90972910 on Build GRCh38
rs1016202421 Chr.12: 90972817-90972817 on Build GRCh38
rs1055328063 Chr.12: 90972752-90972752 on Build GRCh38
rs767872227 Chr.12: 90972911-90972911 on Build GRCh38
rs17784152 Chr.12: 90972872-90972872 on Build GRCh38
rs568659983 Chr.12: 90972798-90972798 on Build GRCh38
rs949706231 Chr.12: 90972756-90972756 on Build GRCh38
rs566219786 Chr.12: 90972635-90972635 on Build GRCh38
rs577030828 Chr.12: 90972869-90972869 on Build GRCh38
rs369410885 Chr.12: 90972791-90972791 on Build GRCh38
rs370001487 Chr.12: 90972811-90972811 on Build GRCh38
rs936551677 Chr.12: 90972738-90972738 on Build GRCh38
rs187299979 Chr.12: 90972896-90972896 on Build GRCh38
rs755362136 Chr.12: 90972820-90972820 on Build GRCh38
rs892411306 Chr.12: 90972759-90972759 on Build GRCh38
rs199845057 Chr.12: 90972871-90972871 on Build GRCh38
rs376854703 Chr.12: 90972785-90972785 on Build GRCh38
rs1005377807 Chr.12: 90972535-90972535 on Build GRCh38
rs775079767 Chr.12: 90972797-90972797 on Build GRCh38
rs151128680 Chr.12: 90972886-90972886 on Build GRCh38
rs556981118 Chr.12: 90972801-90972801 on Build GRCh38
rs962538734 Chr.12: 90972587-90972587 on Build GRCh38
rs757354237 Chr.12: 90972941-90972941 on Build GRCh38
rs144618983 Chr.12: 90972588-90972588 on Build GRCh38
rs754118538 Chr.12: 90972940-90972940 on Build GRCh38
rs749374663 Chr.12: 90972852-90972852 on Build GRCh38
rs978258743 Chr.12: 90972703-90972703 on Build GRCh38
rs535017715 Chr.12: 90972649-90972649 on Build GRCh38
rs548907901 Chr.12: 90972664-90972664 on Build GRCh38
rs925115918 Chr.12: 90972711-90972711 on Build GRCh38
rs1013494959 Chr.12: 90972766-90972766 on Build GRCh38
rs752060272 Chr.12: 90972819-90972819 on Build GRCh38
rs375024769 Chr.12: 90972809-90972809 on Build GRCh38
rs952873255 Chr.12: 90972530-90972530 on Build GRCh38
rs772077640 Chr.12: 90972793-90972793 on Build GRCh38
rs986733669 Chr.12: 90972667-90972671 on Build GRCh38
rs1019498217 Chr.12: 90972951-90972951 on Build GRCh38
rs755265811 Chr.12: 90972751-90972751 on Build GRCh38

View Full Product Details