Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 91178219-91178723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCN
Entrez Gene ID
1634
Gene Name
decorin
Gene Aliases
CSCD, DSPG2, PG40, PGII, PGS2, SLRR1B
UniGene
Hs.156316
Gene Chromosome Location
Chr.12: 91143277-91183124 on Build GRCh38
Transcripts
NM_001920.4 NM_133503.3 NM_133504.3 NM_133505.3 NM_133506.3 NM_133507.3 XM_005268693.1 XM_006719270.1 XM_017018917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3359960 c.76G>T p.D26Y Chr.12: 91178477-91178477 on Build GRCh38
7133576 c.138C>G p.F46L Chr.12: 91178415-91178415 on Build GRCh38
5724139 c.105G>A p.G35G Chr.12: 91178448-91178448 on Build GRCh38
7266321 c.44G>A p.W15* Chr.12: 91178509-91178509 on Build GRCh38
4045362 c.176A>G p.Q59R Chr.12: 91178377-91178377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770622864 Chr.12: 91178598-91178598 on Build GRCh38
rs912177379 Chr.12: 91178508-91178508 on Build GRCh38
rs903995914 Chr.12: 91178321-91178321 on Build GRCh38
rs899227075 Chr.12: 91178406-91178406 on Build GRCh38
rs755803507 Chr.12: 91178582-91178582 on Build GRCh38
rs767135015 Chr.12: 91178432-91178432 on Build GRCh38
rs558487692 Chr.12: 91178294-91178294 on Build GRCh38
rs921127899 Chr.12: 91178454-91178454 on Build GRCh38
rs772310105 Chr.12: 91178612-91178616 on Build GRCh38
rs752978349 Chr.12: 91178414-91178414 on Build GRCh38
rs3138168 Chr.12: 91178627-91178627 on Build GRCh38
rs779957336 Chr.12: 91178542-91178542 on Build GRCh38
rs775866553 Chr.12: 91178613-91178618 on Build GRCh38
rs376291263 Chr.12: 91178331-91178331 on Build GRCh38
rs747838243 Chr.12: 91178474-91178474 on Build GRCh38
rs771981030 Chr.12: 91178317-91178317 on Build GRCh38
rs1042640557 Chr.12: 91178523-91178523 on Build GRCh38
rs377465489 Chr.12: 91178586-91178586 on Build GRCh38
rs973997608 Chr.12: 91178672-91178672 on Build GRCh38
rs983075026 Chr.12: 91178404-91178404 on Build GRCh38
rs543023495 Chr.12: 91178557-91178557 on Build GRCh38
rs771532366 Chr.12: 91178495-91178495 on Build GRCh38
rs751523402 Chr.12: 91178553-91178553 on Build GRCh38
rs554860301 Chr.12: 91178506-91178506 on Build GRCh38
rs768558722 Chr.12: 91178450-91178450 on Build GRCh38
rs753886599 Chr.12: 91178526-91178526 on Build GRCh38
rs774031411 Chr.12: 91178527-91178529 on Build GRCh38
rs1022924687 Chr.12: 91178546-91178546 on Build GRCh38
rs994026250 Chr.12: 91178410-91178410 on Build GRCh38
rs768166776 Chr.12: 91178327-91178327 on Build GRCh38
rs200516355 Chr.12: 91178343-91178343 on Build GRCh38
rs1004821307 Chr.12: 91178647-91178647 on Build GRCh38
rs17018909 Chr.12: 91178363-91178363 on Build GRCh38
rs145597441 Chr.12: 91178499-91178499 on Build GRCh38
rs763583161 Chr.12: 91178362-91178362 on Build GRCh38
rs968488826 Chr.12: 91178618-91178618 on Build GRCh38
rs769608190 Chr.12: 91178634-91178634 on Build GRCh38
rs141738168 Chr.12: 91178520-91178520 on Build GRCh38
rs752822867 Chr.12: 91178516-91178516 on Build GRCh38
rs948667177 Chr.12: 91178643-91178643 on Build GRCh38
rs780010356 Chr.12: 91178437-91178437 on Build GRCh38
rs751949231 Chr.12: 91178391-91178391 on Build GRCh38
rs748801281 Chr.12: 91178588-91178588 on Build GRCh38
rs767929330 Chr.12: 91178510-91178510 on Build GRCh38
rs777864432 Chr.12: 91178420-91178420 on Build GRCh38
rs759983929 Chr.12: 91178403-91178403 on Build GRCh38
rs745812009 Chr.12: 91178308-91178308 on Build GRCh38
rs781069074 Chr.12: 91178559-91178559 on Build GRCh38
rs372859376 Chr.12: 91178384-91178384 on Build GRCh38
rs764351704 Chr.12: 91178525-91178525 on Build GRCh38
rs780647595 Chr.12: 91178457-91178457 on Build GRCh38
rs538859858 Chr.12: 91178278-91178278 on Build GRCh38
rs767628814 Chr.12: 91178409-91178409 on Build GRCh38
rs968896573 Chr.12: 91178255-91178255 on Build GRCh38
rs756082215 Chr.12: 91178415-91178415 on Build GRCh38
rs757397794 Chr.12: 91178539-91178539 on Build GRCh38
rs369115115 Chr.12: 91178339-91178339 on Build GRCh38
rs761210879 Chr.12: 91178332-91178332 on Build GRCh38
rs771816792 Chr.12: 91178312-91178312 on Build GRCh38
rs145908340 Chr.12: 91178639-91178639 on Build GRCh38
rs1031301377 Chr.12: 91178621-91178621 on Build GRCh38
rs746806929 Chr.12: 91178446-91178446 on Build GRCh38
rs373826429 Chr.12: 91178563-91178563 on Build GRCh38
rs3138167 Chr.12: 91178638-91178638 on Build GRCh38
rs973256749 Chr.12: 91178656-91178656 on Build GRCh38
rs936938061 Chr.12: 91178601-91178601 on Build GRCh38
rs769143874 Chr.12: 91178478-91178478 on Build GRCh38
rs759071149 Chr.12: 91178559-91178560 on Build GRCh38
rs762598758 Chr.12: 91178487-91178487 on Build GRCh38
rs565341409 Chr.12: 91178682-91178682 on Build GRCh38
rs756283703 Chr.12: 91178291-91178291 on Build GRCh38
rs950751495 Chr.12: 91178396-91178396 on Build GRCh38
rs1011057527 Chr.12: 91178543-91178543 on Build GRCh38
rs920433897 Chr.12: 91178690-91178690 on Build GRCh38
rs973863511 Chr.12: 91178443-91178443 on Build GRCh38
rs886414085 Chr.12: 91178645-91178645 on Build GRCh38
rs746858264 Chr.12: 91178322-91178322 on Build GRCh38
rs1047691756 Chr.12: 91178412-91178412 on Build GRCh38
rs775198942 Chr.12: 91178453-91178453 on Build GRCh38
rs376051357 Chr.12: 91178421-91178421 on Build GRCh38
rs759052669 Chr.12: 91178281-91178281 on Build GRCh38
rs1016247871 Chr.12: 91178652-91178652 on Build GRCh38
rs962625629 Chr.12: 91178423-91178423 on Build GRCh38
rs977582085 Chr.12: 91178619-91178619 on Build GRCh38
rs924307214 Chr.12: 91178290-91178290 on Build GRCh38

View Full Product Details