Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 91182821-91183325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCN
Entrez Gene ID
1634
Gene Name
decorin
Gene Aliases
CSCD, DSPG2, PG40, PGII, PGS2, SLRR1B
UniGene
Hs.156316
Gene Chromosome Location
Chr.12: 91143277-91183124 on Build GRCh38
Transcripts
NM_001920.4

SNP Detail

SNP ID Chromosome Location
rs955524396 Chr.12: 91182868-91182868 on Build GRCh38
rs141784528 Chr.12: 91183270-91183270 on Build GRCh38
rs922672268 Chr.12: 91182850-91182850 on Build GRCh38
rs964190857 Chr.12: 91183070-91183070 on Build GRCh38
rs540013218 Chr.12: 91182917-91182917 on Build GRCh38
rs762842245 Chr.12: 91183278-91183278 on Build GRCh38
rs567042173 Chr.12: 91183276-91183276 on Build GRCh38
rs957405055 Chr.12: 91183217-91183217 on Build GRCh38
rs765067149 Chr.12: 91182880-91182880 on Build GRCh38
rs897103914 Chr.12: 91183047-91183047 on Build GRCh38
rs573611790 Chr.12: 91183005-91183005 on Build GRCh38
rs1001592345 Chr.12: 91182943-91182943 on Build GRCh38
rs530804842 Chr.12: 91183095-91183095 on Build GRCh38
rs13312820 Chr.12: 91183269-91183269 on Build GRCh38
rs933187244 Chr.12: 91183133-91183133 on Build GRCh38
rs554139939 Chr.12: 91183125-91183125 on Build GRCh38
rs560656144 Chr.12: 91183069-91183070 on Build GRCh38
rs981574465 Chr.12: 91183262-91183262 on Build GRCh38
rs946320371 Chr.12: 91183259-91183259 on Build GRCh38
rs972572136 Chr.12: 91182849-91182849 on Build GRCh38
rs13312818 Chr.12: 91183285-91183285 on Build GRCh38
rs569934519 Chr.12: 91183263-91183263 on Build GRCh38
rs527310415 Chr.12: 91182920-91182921 on Build GRCh38
rs886049891 Chr.12: 91182928-91182928 on Build GRCh38
rs990178020 Chr.12: 91183221-91183221 on Build GRCh38
rs13312821 Chr.12: 91183071-91183071 on Build GRCh38
rs189853358 Chr.12: 91182870-91182870 on Build GRCh38
rs951560834 Chr.12: 91183046-91183046 on Build GRCh38
rs779869744 Chr.12: 91183002-91183002 on Build GRCh38
rs147956869 Chr.12: 91182946-91182946 on Build GRCh38
rs1014462227 Chr.12: 91182949-91182949 on Build GRCh38
rs370700926 Chr.12: 91182934-91182934 on Build GRCh38
rs936208926 Chr.12: 91182916-91182916 on Build GRCh38
rs946183547 Chr.12: 91182931-91182931 on Build GRCh38
rs906270916 Chr.12: 91182937-91182937 on Build GRCh38
rs1051690544 Chr.12: 91183247-91183247 on Build GRCh38
rs13312819 Chr.12: 91183271-91183271 on Build GRCh38
rs577975617 Chr.12: 91182996-91182996 on Build GRCh38
rs913374604 Chr.12: 91182928-91182928 on Build GRCh38
rs911231583 Chr.12: 91182911-91182911 on Build GRCh38
rs551101682 Chr.12: 91183186-91183186 on Build GRCh38
rs1018666177 Chr.12: 91183197-91183197 on Build GRCh38
rs993961102 Chr.12: 91183052-91183052 on Build GRCh38
rs1034051253 Chr.12: 91182944-91182944 on Build GRCh38
rs181496130 Chr.12: 91183092-91183092 on Build GRCh38
rs879664812 Chr.12: 91183178-91183178 on Build GRCh38
rs990416856 Chr.12: 91182919-91182919 on Build GRCh38
rs370171529 Chr.12: 91182893-91182893 on Build GRCh38
rs1055159830 Chr.12: 91182953-91182953 on Build GRCh38
rs894829505 Chr.12: 91182981-91182981 on Build GRCh38
rs1005396151 Chr.12: 91182994-91182994 on Build GRCh38
rs369813559 Chr.12: 91183075-91183075 on Build GRCh38
rs913467575 Chr.12: 91183244-91183244 on Build GRCh38
rs538921668 Chr.12: 91183123-91183123 on Build GRCh38

View Full Product Details