Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 92141028-92141538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTG1
Entrez Gene ID
694
Gene Name
BTG anti-proliferation factor 1
Gene Aliases
APRO2
UniGene
Hs.255935
Gene Chromosome Location
Chr.12: 92140278-92145897 on Build GRCh38
Transcripts
NM_001731.2 NR_046159.2 NR_046160.2 NR_132339.1 NR_132340.1 NR_132341.1

Gene Symbol
LINC01619
Entrez Gene ID
256021
Gene Name
long intergenic non-protein coding RNA 1619
Gene Aliases
C12orf79
UniGene
Hs.651357
Gene Chromosome Location
Chr.12: 91984976-92142831 on Build GRCh38
Transcripts
NM_001731.2 NR_046159.2 NR_046160.2 NR_132339.1 NR_132340.1 NR_132341.1

SNP Detail

SNP ID Chromosome Location
rs559358462 Chr.12: 92141063-92141063 on Build GRCh38
rs1053225290 Chr.12: 92141066-92141066 on Build GRCh38
rs77462034 Chr.12: 92141070-92141070 on Build GRCh38
rs997821327 Chr.12: 92141218-92141218 on Build GRCh38
rs969432680 Chr.12: 92141316-92141316 on Build GRCh38
rs561860844 Chr.12: 92141284-92141284 on Build GRCh38
rs1048327882 Chr.12: 92141501-92141501 on Build GRCh38
rs530136086 Chr.12: 92141298-92141298 on Build GRCh38
rs1033167312 Chr.12: 92141329-92141329 on Build GRCh38
rs979440053 Chr.12: 92141295-92141295 on Build GRCh38
rs1015329956 Chr.12: 92141176-92141176 on Build GRCh38
rs893003767 Chr.12: 92141147-92141147 on Build GRCh38
rs577138031 Chr.12: 92141382-92141382 on Build GRCh38
rs950187233 Chr.12: 92141400-92141400 on Build GRCh38
rs78917531 Chr.12: 92141084-92141084 on Build GRCh38
rs765407584 Chr.12: 92141142-92141142 on Build GRCh38
rs527643820 Chr.12: 92141348-92141348 on Build GRCh38
rs988893286 Chr.12: 92141340-92141340 on Build GRCh38
rs755707334 Chr.12: 92141476-92141476 on Build GRCh38
rs750665843 Chr.12: 92141245-92141245 on Build GRCh38
rs943097582 Chr.12: 92141479-92141479 on Build GRCh38
rs529287845 Chr.12: 92141189-92141189 on Build GRCh38
rs1044793969 Chr.12: 92141113-92141113 on Build GRCh38
rs919502181 Chr.12: 92141351-92141351 on Build GRCh38
rs888844885 Chr.12: 92141079-92141079 on Build GRCh38
rs2384848 Chr.12: 92141148-92141148 on Build GRCh38
rs533881128 Chr.12: 92141060-92141060 on Build GRCh38
rs28399551 Chr.12: 92141333-92141333 on Build GRCh38
rs755555573 Chr.12: 92141386-92141386 on Build GRCh38
rs868463290 Chr.12: 92141124-92141124 on Build GRCh38
rs900844182 Chr.12: 92141207-92141207 on Build GRCh38
rs984908959 Chr.12: 92141447-92141447 on Build GRCh38
rs1028414007 Chr.12: 92141224-92141224 on Build GRCh38
rs149250109 Chr.12: 92141342-92141343 on Build GRCh38
rs954289245 Chr.12: 92141233-92141233 on Build GRCh38
rs961257801 Chr.12: 92141347-92141347 on Build GRCh38
rs747619859 Chr.12: 92141470-92141470 on Build GRCh38
rs141996513 Chr.12: 92141379-92141379 on Build GRCh38
rs752007180 Chr.12: 92141376-92141376 on Build GRCh38
rs937938653 Chr.12: 92141116-92141116 on Build GRCh38
rs917248261 Chr.12: 92141077-92141077 on Build GRCh38
rs1011412064 Chr.12: 92141185-92141185 on Build GRCh38
rs1048202930 Chr.12: 92141123-92141123 on Build GRCh38
rs1031929354 Chr.12: 92141134-92141134 on Build GRCh38
rs980098983 Chr.12: 92141449-92141449 on Build GRCh38
rs780535263 Chr.12: 92141412-92141412 on Build GRCh38
rs758705621 Chr.12: 92141254-92141254 on Build GRCh38
rs777506429 Chr.12: 92141509-92141509 on Build GRCh38
rs1003904651 Chr.12: 92141174-92141174 on Build GRCh38
rs548067987 Chr.12: 92141115-92141115 on Build GRCh38
rs561462815 Chr.12: 92141118-92141118 on Build GRCh38
rs930839394 Chr.12: 92141380-92141381 on Build GRCh38
rs978916578 Chr.12: 92141056-92141056 on Build GRCh38

View Full Product Details