Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 8015154-8015651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERRFI1
Entrez Gene ID
54206
Gene Name
ERBB receptor feedback inhibitor 1
Gene Aliases
GENE-33, MIG-6, MIG6, RALT
UniGene
Hs.605445
Gene Chromosome Location
Chr.1: 8011719-8026333 on Build GRCh38
Transcripts
NM_018948.3 XM_005263477.2 XM_006710697.2 XM_011541596.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5847357 c.102G>A p.W34* Chr.1: 8015518-8015518 on Build GRCh38
6460714 c.112A>T p.S38C Chr.1: 8015508-8015508 on Build GRCh38
6968214 c.80G>A p.G27E Chr.1: 8015540-8015540 on Build GRCh38
6953132 c.5C>T p.S2L Chr.1: 8015615-8015615 on Build GRCh38
6930815 c.71G>A p.R24Q Chr.1: 8015549-8015549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144399366 Chr.1: 8015251-8015251 on Build GRCh38
rs369117984 Chr.1: 8015414-8015414 on Build GRCh38
rs750577494 Chr.1: 8015368-8015368 on Build GRCh38
rs529402636 Chr.1: 8015432-8015432 on Build GRCh38
rs201907699 Chr.1: 8015580-8015580 on Build GRCh38
rs765586292 Chr.1: 8015525-8015525 on Build GRCh38
rs886760787 Chr.1: 8015318-8015318 on Build GRCh38
rs746929809 Chr.1: 8015316-8015316 on Build GRCh38
rs773420542 Chr.1: 8015506-8015506 on Build GRCh38
rs776574075 Chr.1: 8015445-8015445 on Build GRCh38
rs145113484 Chr.1: 8015540-8015540 on Build GRCh38
rs971253551 Chr.1: 8015426-8015426 on Build GRCh38
rs777235827 Chr.1: 8015422-8015422 on Build GRCh38
rs776593719 Chr.1: 8015613-8015613 on Build GRCh38
rs762102577 Chr.1: 8015482-8015482 on Build GRCh38
rs376892066 Chr.1: 8015479-8015479 on Build GRCh38
rs769669957 Chr.1: 8015563-8015563 on Build GRCh38
rs1004361134 Chr.1: 8015416-8015416 on Build GRCh38
rs199654778 Chr.1: 8015404-8015404 on Build GRCh38
rs758066492 Chr.1: 8015418-8015418 on Build GRCh38
rs982848694 Chr.1: 8015181-8015181 on Build GRCh38
rs767560134 Chr.1: 8015436-8015437 on Build GRCh38
rs991416337 Chr.1: 8015504-8015504 on Build GRCh38
rs370227993 Chr.1: 8015511-8015511 on Build GRCh38
rs1003594715 Chr.1: 8015427-8015427 on Build GRCh38
rs776290618 Chr.1: 8015328-8015328 on Build GRCh38
rs756656981 Chr.1: 8015378-8015378 on Build GRCh38
rs368971771 Chr.1: 8015294-8015294 on Build GRCh38
rs778682070 Chr.1: 8015296-8015296 on Build GRCh38
rs765734320 Chr.1: 8015616-8015616 on Build GRCh38
rs544401150 Chr.1: 8015466-8015466 on Build GRCh38
rs750920915 Chr.1: 8015228-8015228 on Build GRCh38
rs970818780 Chr.1: 8015250-8015250 on Build GRCh38
rs760063562 Chr.1: 8015614-8015614 on Build GRCh38
rs772410037 Chr.1: 8015595-8015595 on Build GRCh38
rs545922916 Chr.1: 8015198-8015198 on Build GRCh38
rs775450610 Chr.1: 8015467-8015467 on Build GRCh38
rs770897374 Chr.1: 8015323-8015323 on Build GRCh38
rs760807582 Chr.1: 8015606-8015606 on Build GRCh38
rs200259274 Chr.1: 8015558-8015558 on Build GRCh38
rs773510363 Chr.1: 8015604-8015604 on Build GRCh38
rs914522975 Chr.1: 8015537-8015537 on Build GRCh38
rs749701648 Chr.1: 8015285-8015285 on Build GRCh38
rs745798544 Chr.1: 8015455-8015455 on Build GRCh38
rs770998621 Chr.1: 8015612-8015612 on Build GRCh38
rs751230334 Chr.1: 8015550-8015550 on Build GRCh38
rs1027171161 Chr.1: 8015381-8015381 on Build GRCh38
rs558318143 Chr.1: 8015359-8015359 on Build GRCh38
rs902956661 Chr.1: 8015321-8015321 on Build GRCh38
rs372927205 Chr.1: 8015417-8015417 on Build GRCh38
rs771500410 Chr.1: 8015311-8015311 on Build GRCh38
rs759438365 Chr.1: 8015343-8015343 on Build GRCh38
rs937247399 Chr.1: 8015263-8015263 on Build GRCh38
rs562057462 Chr.1: 8015347-8015347 on Build GRCh38
rs766976133 Chr.1: 8015392-8015392 on Build GRCh38
rs747808154 Chr.1: 8015309-8015309 on Build GRCh38
rs749161578 Chr.1: 8015581-8015581 on Build GRCh38
rs753050395 Chr.1: 8015623-8015623 on Build GRCh38
rs759989048 Chr.1: 8015519-8015519 on Build GRCh38
rs756719677 Chr.1: 8015551-8015551 on Build GRCh38
rs772461595 Chr.1: 8015486-8015486 on Build GRCh38
rs374119208 Chr.1: 8015364-8015364 on Build GRCh38
rs1034096610 Chr.1: 8015438-8015438 on Build GRCh38
rs762159468 Chr.1: 8015363-8015363 on Build GRCh38
rs760895809 Chr.1: 8015509-8015509 on Build GRCh38
rs147802351 Chr.1: 8015312-8015312 on Build GRCh38
rs957635030 Chr.1: 8015430-8015430 on Build GRCh38
rs754301065 Chr.1: 8015518-8015518 on Build GRCh38
rs753103592 Chr.1: 8015531-8015531 on Build GRCh38
rs761848847 Chr.1: 8015417-8015418 on Build GRCh38
rs12087168 Chr.1: 8015560-8015560 on Build GRCh38
rs76755344 Chr.1: 8015252-8015252 on Build GRCh38
rs746592564 Chr.1: 8015424-8015424 on Build GRCh38
rs755307255 Chr.1: 8015396-8015396 on Build GRCh38
rs754139728 Chr.1: 8015394-8015394 on Build GRCh38

View Full Product Details