Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 8970872-8971307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA6
Entrez Gene ID
765
Gene Name
carbonic anhydrase 6
Gene Aliases
CA-VI, GUSTIN
UniGene
Hs.100322
Gene Chromosome Location
Chr.1: 8945786-8975092 on Build GRCh38
Transcripts
NM_001215.3 NM_001270500.1 NM_001270501.1 NM_001270502.1 XM_011542083.2 XM_011542084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7304143 c.811C>A p.H271N Chr.1: 8970948-8970948 on Build GRCh38
4749660 c.790C>T p.R264C Chr.1: 8970927-8970927 on Build GRCh38
5687219 c.795G>T p.R265S Chr.1: 8970932-8970932 on Build GRCh38
3751447 c.768C>T p.N256N Chr.1: 8970905-8970905 on Build GRCh38
4795367 c.794G>A p.R265K Chr.1: 8970931-8970931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758271140 Chr.1: 8970901-8970901 on Build GRCh38
rs1006370658 Chr.1: 8971181-8971181 on Build GRCh38
rs78602434 Chr.1: 8971199-8971199 on Build GRCh38
rs761039346 Chr.1: 8971146-8971146 on Build GRCh38
rs759136963 Chr.1: 8970986-8970986 on Build GRCh38
rs960942489 Chr.1: 8971224-8971224 on Build GRCh38
rs542173751 Chr.1: 8971174-8971174 on Build GRCh38
rs549441507 Chr.1: 8970920-8970920 on Build GRCh38
rs3737665 Chr.1: 8970905-8970905 on Build GRCh38
rs1028607971 Chr.1: 8971153-8971153 on Build GRCh38
rs959636595 Chr.1: 8971204-8971205 on Build GRCh38
rs902329065 Chr.1: 8971048-8971048 on Build GRCh38
rs578136302 Chr.1: 8970993-8970993 on Build GRCh38
rs200520970 Chr.1: 8970974-8970974 on Build GRCh38
rs1040213215 Chr.1: 8971280-8971280 on Build GRCh38
rs181233618 Chr.1: 8970992-8970992 on Build GRCh38
rs972929399 Chr.1: 8971204-8971204 on Build GRCh38
rs111279535 Chr.1: 8971035-8971035 on Build GRCh38
rs974050004 Chr.1: 8971203-8971203 on Build GRCh38
rs756984549 Chr.1: 8970909-8970909 on Build GRCh38
rs200133195 Chr.1: 8971030-8971030 on Build GRCh38
rs749482487 Chr.1: 8970921-8970921 on Build GRCh38
rs1054408977 Chr.1: 8971124-8971124 on Build GRCh38
rs4908799 Chr.1: 8970994-8970994 on Build GRCh38
rs928256805 Chr.1: 8970939-8970939 on Build GRCh38
rs999378277 Chr.1: 8971051-8971051 on Build GRCh38
rs893067142 Chr.1: 8971155-8971155 on Build GRCh38
rs776292459 Chr.1: 8970978-8970978 on Build GRCh38
rs901769781 Chr.1: 8971136-8971136 on Build GRCh38
rs1044169956 Chr.1: 8971268-8971268 on Build GRCh38
rs751421530 Chr.1: 8970996-8970996 on Build GRCh38
rs946726485 Chr.1: 8971251-8971251 on Build GRCh38
rs267598758 Chr.1: 8970914-8970914 on Build GRCh38
rs868741591 Chr.1: 8970971-8970971 on Build GRCh38
rs139363760 Chr.1: 8970900-8970900 on Build GRCh38
rs760352504 Chr.1: 8970990-8970993 on Build GRCh38
rs116134104 Chr.1: 8971212-8971212 on Build GRCh38
rs772700668 Chr.1: 8970957-8970957 on Build GRCh38
rs367775539 Chr.1: 8971022-8971022 on Build GRCh38
rs753777901 Chr.1: 8971171-8971171 on Build GRCh38
rs761972360 Chr.1: 8971244-8971244 on Build GRCh38
rs916569036 Chr.1: 8971250-8971250 on Build GRCh38
rs766622056 Chr.1: 8970975-8970975 on Build GRCh38
rs754519428 Chr.1: 8970898-8970898 on Build GRCh38
rs183507557 Chr.1: 8971164-8971164 on Build GRCh38
rs532310180 Chr.1: 8971276-8971276 on Build GRCh38
rs998770654 Chr.1: 8971050-8971050 on Build GRCh38
rs576024936 Chr.1: 8971189-8971189 on Build GRCh38
rs748672108 Chr.1: 8970949-8970949 on Build GRCh38
rs757110971 Chr.1: 8971023-8971023 on Build GRCh38
rs775943143 Chr.1: 8971055-8971055 on Build GRCh38
rs996643539 Chr.1: 8971158-8971158 on Build GRCh38
rs768880697 Chr.1: 8970927-8970927 on Build GRCh38
rs774516209 Chr.1: 8970947-8970947 on Build GRCh38
rs111580237 Chr.1: 8971188-8971188 on Build GRCh38
rs973366993 Chr.1: 8970930-8970930 on Build GRCh38
rs140431319 Chr.1: 8970973-8970973 on Build GRCh38
rs116366173 Chr.1: 8971277-8971277 on Build GRCh38

View Full Product Details