Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9728561-9729056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSTN1
Entrez Gene ID
22883
Gene Name
calsyntenin 1
Gene Aliases
ALC-ALPHA, CDHR12, CST-1, CSTN1, PIK3CD, XB31alpha, alcalpha1, alcalpha2
UniGene
Hs.29665
Gene Chromosome Location
Chr.1: 9729021-9824526 on Build GRCh38
Transcripts
NM_001009566.2 NM_001302883.1 NM_014944.4 NM_005026.3 XM_006710687.2 XM_006710689.2 XM_017001476.1 XM_017001477.1 XM_017001478.1 XM_017001479.1 XM_017001480.1 XM_017001481.1 XM_017001482.1 XM_017001483.1 XM_017001484.1 XM_017001485.1

Gene Symbol
PIK3CD
Entrez Gene ID
5293
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta
Gene Aliases
APDS, IMD14, P110DELTA, PI3K, p110D
UniGene
Hs.518451
Gene Chromosome Location
Chr.1: 9629889-9729114 on Build GRCh38
Transcripts
NM_001009566.2 NM_001302883.1 NM_014944.4 NM_005026.3 XM_006710687.2 XM_006710689.2 XM_017001476.1 XM_017001477.1 XM_017001478.1 XM_017001479.1 XM_017001480.1 XM_017001481.1 XM_017001482.1 XM_017001483.1 XM_017001484.1 XM_017001485.1

SNP Detail

SNP ID Chromosome Location
rs555451470 Chr.1: 9728625-9728625 on Build GRCh38
rs993175235 Chr.1: 9728679-9728679 on Build GRCh38
rs142343550 Chr.1: 9728781-9728782 on Build GRCh38
rs893112935 Chr.1: 9728591-9728591 on Build GRCh38
rs145984383 Chr.1: 9728837-9728840 on Build GRCh38
rs775634541 Chr.1: 9729018-9729018 on Build GRCh38
rs889058615 Chr.1: 9728620-9728620 on Build GRCh38
rs934829552 Chr.1: 9728713-9728713 on Build GRCh38
rs1008722681 Chr.1: 9728633-9728633 on Build GRCh38
rs1044164107 Chr.1: 9729023-9729024 on Build GRCh38
rs1040674574 Chr.1: 9728969-9728970 on Build GRCh38
rs777380551 Chr.1: 9728712-9728712 on Build GRCh38
rs914650673 Chr.1: 9728916-9728916 on Build GRCh38
rs749444829 Chr.1: 9728683-9728684 on Build GRCh38
rs746127837 Chr.1: 9728784-9728785 on Build GRCh38
rs990207437 Chr.1: 9728714-9728714 on Build GRCh38
rs982046414 Chr.1: 9728587-9728587 on Build GRCh38
rs912811973 Chr.1: 9729027-9729027 on Build GRCh38
rs956172024 Chr.1: 9728913-9728916 on Build GRCh38
rs577785029 Chr.1: 9728770-9728770 on Build GRCh38
rs139933669 Chr.1: 9728676-9728676 on Build GRCh38
rs184417664 Chr.1: 9729013-9729013 on Build GRCh38
rs573315808 Chr.1: 9728781-9728781 on Build GRCh38
rs540223155 Chr.1: 9728834-9728834 on Build GRCh38
rs1048780021 Chr.1: 9728791-9728791 on Build GRCh38
rs912521239 Chr.1: 9728937-9728941 on Build GRCh38
rs529632901 Chr.1: 9729024-9729027 on Build GRCh38
rs990343279 Chr.1: 9728915-9728915 on Build GRCh38
rs80109509 Chr.1: 9728705-9728705 on Build GRCh38
rs914681834 Chr.1: 9728723-9728724 on Build GRCh38
rs774506013 Chr.1: 9728836-9728837 on Build GRCh38
rs564409139 Chr.1: 9728780-9728780 on Build GRCh38
rs748783576 Chr.1: 9728907-9728908 on Build GRCh38
rs756686923 Chr.1: 9728590-9728590 on Build GRCh38
rs932624080 Chr.1: 9728727-9728727 on Build GRCh38
rs919199747 Chr.1: 9728998-9729000 on Build GRCh38
rs944068442 Chr.1: 9728937-9728937 on Build GRCh38
rs76391189 Chr.1: 9729020-9729020 on Build GRCh38
rs932655103 Chr.1: 9728609-9728609 on Build GRCh38
rs778141996 Chr.1: 9728677-9728677 on Build GRCh38
rs1048917590 Chr.1: 9729017-9729017 on Build GRCh38
rs567461402 Chr.1: 9728673-9728673 on Build GRCh38
rs971094372 Chr.1: 9728849-9728849 on Build GRCh38
rs770517598 Chr.1: 9728815-9728815 on Build GRCh38
rs546244841 Chr.1: 9728772-9728772 on Build GRCh38
rs561755558 Chr.1: 9728952-9728952 on Build GRCh38
rs934692204 Chr.1: 9728589-9728589 on Build GRCh38
rs556349176 Chr.1: 9728711-9728711 on Build GRCh38
rs553526734 Chr.1: 9728588-9728588 on Build GRCh38
rs985565907 Chr.1: 9728936-9728937 on Build GRCh38
rs1023204723 Chr.1: 9728700-9728700 on Build GRCh38
rs1050343358 Chr.1: 9728610-9728610 on Build GRCh38
rs757225268 Chr.1: 9728782-9728784 on Build GRCh38
rs562228504 Chr.1: 9728840-9728841 on Build GRCh38
rs1050222350 Chr.1: 9728764-9728765 on Build GRCh38
rs951692208 Chr.1: 9728683-9728683 on Build GRCh38
rs142831718 Chr.1: 9729023-9729023 on Build GRCh38
rs954147676 Chr.1: 9728923-9728923 on Build GRCh38
rs1011675348 Chr.1: 9728980-9728980 on Build GRCh38
rs1012577407 Chr.1: 9728826-9728826 on Build GRCh38
rs775070981 Chr.1: 9728684-9728687 on Build GRCh38
rs956101145 Chr.1: 9728974-9728974 on Build GRCh38
rs772119200 Chr.1: 9728975-9728975 on Build GRCh38
rs745663402 Chr.1: 9728935-9728935 on Build GRCh38
rs981718815 Chr.1: 9728707-9728707 on Build GRCh38
rs1027375361 Chr.1: 9728681-9728681 on Build GRCh38
rs201307826 Chr.1: 9728857-9728857 on Build GRCh38
rs1003507326 Chr.1: 9728871-9728871 on Build GRCh38
rs1031946974 Chr.1: 9728909-9728909 on Build GRCh38
rs762165239 Chr.1: 9728910-9728911 on Build GRCh38
rs778702520 Chr.1: 9728823-9728823 on Build GRCh38
rs750811249 Chr.1: 9728632-9728633 on Build GRCh38
rs1022156678 Chr.1: 9729012-9729012 on Build GRCh38

View Full Product Details