Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10636771-10637312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASZ1
Entrez Gene ID
54897
Gene Name
castor zinc finger 1
Gene Aliases
CAS11, CST, SRG, ZNF693, dJ734G22.1
UniGene
Hs.439894
Gene Chromosome Location
Chr.1: 10636602-10796676 on Build GRCh38
Transcripts
NM_001079843.2 XM_017001539.1 XM_017001540.1 XM_017001541.1

SNP Detail

SNP ID Chromosome Location
rs536617450 Chr.1: 10637100-10637100 on Build GRCh38
rs949689315 Chr.1: 10636954-10636954 on Build GRCh38
rs1056155361 Chr.1: 10636927-10636927 on Build GRCh38
rs538862111 Chr.1: 10636992-10636992 on Build GRCh38
rs961440113 Chr.1: 10637193-10637193 on Build GRCh38
rs576925129 Chr.1: 10637225-10637225 on Build GRCh38
rs530608326 Chr.1: 10636901-10636901 on Build GRCh38
rs982492959 Chr.1: 10636895-10636895 on Build GRCh38
rs181999733 Chr.1: 10636951-10636951 on Build GRCh38
rs920436865 Chr.1: 10637206-10637206 on Build GRCh38
rs897746241 Chr.1: 10637105-10637107 on Build GRCh38
rs910307939 Chr.1: 10637244-10637244 on Build GRCh38
rs1045386523 Chr.1: 10636964-10636964 on Build GRCh38
rs1027519439 Chr.1: 10637119-10637120 on Build GRCh38
rs955619336 Chr.1: 10637116-10637116 on Build GRCh38
rs566389318 Chr.1: 10637050-10637050 on Build GRCh38
rs199954187 Chr.1: 10637176-10637177 on Build GRCh38
rs377187327 Chr.1: 10637177-10637177 on Build GRCh38
rs547798657 Chr.1: 10636860-10636860 on Build GRCh38
rs372373282 Chr.1: 10637123-10637123 on Build GRCh38
rs986673340 Chr.1: 10637236-10637236 on Build GRCh38
rs961469587 Chr.1: 10636871-10636871 on Build GRCh38
rs190890391 Chr.1: 10637151-10637151 on Build GRCh38
rs537545466 Chr.1: 10637182-10637182 on Build GRCh38
rs1009252275 Chr.1: 10636842-10636842 on Build GRCh38
rs529605282 Chr.1: 10636800-10636800 on Build GRCh38
rs993337355 Chr.1: 10636862-10636862 on Build GRCh38
rs748430278 Chr.1: 10636834-10636834 on Build GRCh38
rs1034982058 Chr.1: 10637209-10637209 on Build GRCh38
rs142525076 Chr.1: 10636990-10636990 on Build GRCh38
rs1023736282 Chr.1: 10637164-10637165 on Build GRCh38
rs146005610 Chr.1: 10637259-10637259 on Build GRCh38
rs1024082502 Chr.1: 10636886-10636887 on Build GRCh38
rs765073839 Chr.1: 10637268-10637268 on Build GRCh38
rs908596579 Chr.1: 10636850-10636850 on Build GRCh38
rs893330154 Chr.1: 10637059-10637059 on Build GRCh38
rs776056531 Chr.1: 10637115-10637117 on Build GRCh38
rs1054619135 Chr.1: 10637018-10637018 on Build GRCh38
rs574118680 Chr.1: 10636817-10636818 on Build GRCh38
rs951932910 Chr.1: 10637213-10637213 on Build GRCh38
rs930568648 Chr.1: 10636923-10636923 on Build GRCh38
rs988309174 Chr.1: 10636951-10636953 on Build GRCh38
rs1010652155 Chr.1: 10636809-10636809 on Build GRCh38
rs753650111 Chr.1: 10636905-10636905 on Build GRCh38
rs969436337 Chr.1: 10636894-10636895 on Build GRCh38
rs935547682 Chr.1: 10636921-10636922 on Build GRCh38
rs1011398211 Chr.1: 10637229-10637229 on Build GRCh38
rs949989638 Chr.1: 10637254-10637254 on Build GRCh38
rs764301559 Chr.1: 10636837-10636846 on Build GRCh38
rs755939640 Chr.1: 10637226-10637226 on Build GRCh38
rs186499528 Chr.1: 10637017-10637017 on Build GRCh38
rs367556741 Chr.1: 10637001-10637001 on Build GRCh38
rs946486203 Chr.1: 10636961-10636961 on Build GRCh38
rs369702021 Chr.1: 10637214-10637214 on Build GRCh38
rs543561768 Chr.1: 10637120-10637121 on Build GRCh38
rs1013143015 Chr.1: 10637132-10637133 on Build GRCh38
rs1018879667 Chr.1: 10636853-10636853 on Build GRCh38
rs757231361 Chr.1: 10637103-10637103 on Build GRCh38
rs920452106 Chr.1: 10636888-10636888 on Build GRCh38
rs974187452 Chr.1: 10637205-10637205 on Build GRCh38
rs190708048 Chr.1: 10636885-10636885 on Build GRCh38
rs952808880 Chr.1: 10636883-10636883 on Build GRCh38

View Full Product Details