Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10949545-10950031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf127
Entrez Gene ID
148345
Gene Name
chromosome 1 open reading frame 127
Gene Aliases
-
UniGene
Hs.127026
Gene Chromosome Location
Chr.1: 10946473-10982037 on Build GRCh38
Transcripts
NM_001170754.1 XM_017000373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1583187 c.1115C>T p.S372L Chr.1: 10949799-10949799 on Build GRCh38
3470932 c.1239G>A p.Q413Q Chr.1: 10949675-10949675 on Build GRCh38
3975853 c.1254G>T p.R418R Chr.1: 10949660-10949660 on Build GRCh38
4667297 c.1159T>C p.S387P Chr.1: 10949755-10949755 on Build GRCh38
893536 c.614C>T p.S205L Chr.1: 10949799-10949799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376799038 Chr.1: 10949709-10949709 on Build GRCh38
rs578152471 Chr.1: 10949797-10949797 on Build GRCh38
rs760131101 Chr.1: 10949849-10949849 on Build GRCh38
rs183542170 Chr.1: 10949605-10949605 on Build GRCh38
rs560318412 Chr.1: 10949830-10949830 on Build GRCh38
rs759522674 Chr.1: 10949685-10949685 on Build GRCh38
rs1281013 Chr.1: 10949622-10949622 on Build GRCh38
rs752832963 Chr.1: 10949692-10949692 on Build GRCh38
rs34236308 Chr.1: 10949945-10949946 on Build GRCh38
rs973763373 Chr.1: 10949669-10949669 on Build GRCh38
rs951449503 Chr.1: 10949900-10949900 on Build GRCh38
rs571052057 Chr.1: 10949635-10949635 on Build GRCh38
rs757212121 Chr.1: 10949638-10949638 on Build GRCh38
rs895574095 Chr.1: 10949567-10949567 on Build GRCh38
rs750415326 Chr.1: 10949730-10949730 on Build GRCh38
rs368723168 Chr.1: 10949617-10949617 on Build GRCh38
rs374114840 Chr.1: 10949578-10949578 on Build GRCh38
rs769725311 Chr.1: 10949850-10949850 on Build GRCh38
rs1005688371 Chr.1: 10949765-10949765 on Build GRCh38
rs773140391 Chr.1: 10949751-10949751 on Build GRCh38
rs755554107 Chr.1: 10949741-10949741 on Build GRCh38
rs527461195 Chr.1: 10949863-10949863 on Build GRCh38
rs765408636 Chr.1: 10949688-10949688 on Build GRCh38
rs780626374 Chr.1: 10949639-10949639 on Build GRCh38
rs151254184 Chr.1: 10949647-10949647 on Build GRCh38
rs979226978 Chr.1: 10950003-10950003 on Build GRCh38
rs927182078 Chr.1: 10949967-10949967 on Build GRCh38
rs531491611 Chr.1: 10949982-10949982 on Build GRCh38
rs980801590 Chr.1: 10949951-10949951 on Build GRCh38
rs762077217 Chr.1: 10949604-10949604 on Build GRCh38
rs141484983 Chr.1: 10949919-10949919 on Build GRCh38
rs138476809 Chr.1: 10949787-10949787 on Build GRCh38
rs757115935 Chr.1: 10949725-10949725 on Build GRCh38
rs534520725 Chr.1: 10949661-10949661 on Build GRCh38
rs748851491 Chr.1: 10949779-10949779 on Build GRCh38
rs963001901 Chr.1: 10949950-10949950 on Build GRCh38
rs1020665301 Chr.1: 10949966-10949966 on Build GRCh38
rs751894293 Chr.1: 10949739-10949739 on Build GRCh38
rs982870029 Chr.1: 10949912-10949912 on Build GRCh38
rs144101789 Chr.1: 10949640-10949640 on Build GRCh38
rs998280555 Chr.1: 10949885-10949885 on Build GRCh38
rs773831399 Chr.1: 10949607-10949607 on Build GRCh38
rs763270360 Chr.1: 10949706-10949706 on Build GRCh38
rs145096321 Chr.1: 10949798-10949798 on Build GRCh38
rs865811981 Chr.1: 10949782-10949782 on Build GRCh38
rs766680398 Chr.1: 10949662-10949662 on Build GRCh38
rs574255703 Chr.1: 10949758-10949758 on Build GRCh38
rs906057691 Chr.1: 10949817-10949817 on Build GRCh38
rs1029814651 Chr.1: 10949888-10949888 on Build GRCh38
rs140420878 Chr.1: 10949659-10949659 on Build GRCh38
rs767673160 Chr.1: 10949704-10949704 on Build GRCh38
rs769340376 Chr.1: 10949593-10949593 on Build GRCh38
rs367695286 Chr.1: 10949579-10949579 on Build GRCh38
rs372353780 Chr.1: 10949636-10949636 on Build GRCh38
rs762120545 Chr.1: 10949687-10949688 on Build GRCh38
rs774707145 Chr.1: 10949596-10949596 on Build GRCh38
rs183368244 Chr.1: 10949771-10949771 on Build GRCh38
rs929685940 Chr.1: 10949702-10949702 on Build GRCh38
rs199606004 Chr.1: 10949615-10949615 on Build GRCh38
rs367886125 Chr.1: 10949729-10949729 on Build GRCh38
rs188534393 Chr.1: 10949920-10949920 on Build GRCh38
rs1281012 Chr.1: 10949646-10949646 on Build GRCh38
rs765373523 Chr.1: 10949620-10949620 on Build GRCh38
rs760666482 Chr.1: 10949585-10949585 on Build GRCh38
rs111866188 Chr.1: 10949678-10949678 on Build GRCh38
rs777344923 Chr.1: 10949684-10949684 on Build GRCh38
rs1749888 Chr.1: 10949869-10949869 on Build GRCh38
rs751373265 Chr.1: 10949716-10949716 on Build GRCh38
rs545472559 Chr.1: 10949799-10949799 on Build GRCh38
rs988835030 Chr.1: 10949934-10949934 on Build GRCh38
rs12724985 Chr.1: 10949633-10949633 on Build GRCh38
rs144408399 Chr.1: 10949644-10949644 on Build GRCh38
rs771577554 Chr.1: 10949674-10949674 on Build GRCh38
rs753993390 Chr.1: 10949619-10949619 on Build GRCh38

View Full Product Details