Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11073542-11074040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC10
Entrez Gene ID
5394
Gene Name
exosome component 10
Gene Aliases
PM-Scl, PM/Scl-100, PMSCL, PMSCL2, RRP6, Rrp6p, p2, p3, p4
UniGene
Hs.632368
Gene Chromosome Location
Chr.1: 11066613-11099910 on Build GRCh38
Transcripts
NM_001001998.2 NM_002685.3 XM_005263475.2 XM_017001494.1 XM_017001495.1 XM_017001496.1 XM_017001497.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
893720 c.2113G>A p.V705I Chr.1: 11073978-11073978 on Build GRCh38
4534855 c.2127G>T p.A709A Chr.1: 11073964-11073964 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs990813609 Chr.1: 11074011-11074013 on Build GRCh38
rs755186517 Chr.1: 11073887-11073887 on Build GRCh38
rs377479487 Chr.1: 11073899-11073900 on Build GRCh38
rs936963097 Chr.1: 11073846-11073846 on Build GRCh38
rs761215129 Chr.1: 11073957-11073957 on Build GRCh38
rs868160189 Chr.1: 11073814-11073814 on Build GRCh38
rs553504837 Chr.1: 11073661-11073661 on Build GRCh38
rs937641903 Chr.1: 11073637-11073637 on Build GRCh38
rs771367983 Chr.1: 11073892-11073893 on Build GRCh38
rs369649478 Chr.1: 11073904-11073904 on Build GRCh38
rs1057503906 Chr.1: 11073647-11073647 on Build GRCh38
rs779004519 Chr.1: 11073889-11073889 on Build GRCh38
rs917879126 Chr.1: 11073709-11073709 on Build GRCh38
rs780636359 Chr.1: 11073923-11073923 on Build GRCh38
rs35294839 Chr.1: 11073727-11073727 on Build GRCh38
rs10082000 Chr.1: 11073899-11073899 on Build GRCh38
rs765413962 Chr.1: 11073966-11073966 on Build GRCh38
rs140844627 Chr.1: 11074000-11074000 on Build GRCh38
rs143492861 Chr.1: 11073630-11073630 on Build GRCh38
rs117771172 Chr.1: 11073986-11073986 on Build GRCh38
rs747914166 Chr.1: 11073891-11073891 on Build GRCh38
rs145888478 Chr.1: 11073927-11073927 on Build GRCh38
rs190900126 Chr.1: 11073964-11073964 on Build GRCh38
rs755868644 Chr.1: 11073883-11073884 on Build GRCh38
rs772373112 Chr.1: 11073938-11073938 on Build GRCh38
rs937896146 Chr.1: 11073641-11073641 on Build GRCh38
rs563576180 Chr.1: 11073865-11073865 on Build GRCh38
rs900173909 Chr.1: 11073833-11073833 on Build GRCh38
rs376441584 Chr.1: 11073949-11073949 on Build GRCh38
rs541243766 Chr.1: 11073648-11073648 on Build GRCh38
rs373184731 Chr.1: 11073958-11073958 on Build GRCh38
rs770907306 Chr.1: 11073922-11073922 on Build GRCh38
rs983226019 Chr.1: 11073598-11073598 on Build GRCh38
rs895988846 Chr.1: 11073669-11073669 on Build GRCh38
rs200670819 Chr.1: 11073992-11073992 on Build GRCh38
rs917460818 Chr.1: 11073851-11073851 on Build GRCh38
rs115028803 Chr.1: 11073616-11073616 on Build GRCh38
rs775018603 Chr.1: 11073941-11073941 on Build GRCh38
rs919940758 Chr.1: 11073640-11073640 on Build GRCh38
rs773671843 Chr.1: 11074011-11074011 on Build GRCh38
rs996150053 Chr.1: 11073706-11073706 on Build GRCh38
rs778366565 Chr.1: 11073889-11073890 on Build GRCh38
rs1010787377 Chr.1: 11073680-11073680 on Build GRCh38
rs1041838255 Chr.1: 11073683-11073683 on Build GRCh38
rs1037351815 Chr.1: 11073628-11073628 on Build GRCh38
rs753993336 Chr.1: 11073886-11073886 on Build GRCh38
rs112419013 Chr.1: 11073746-11073746 on Build GRCh38
rs777609061 Chr.1: 11073903-11073903 on Build GRCh38
rs747603625 Chr.1: 11073890-11073891 on Build GRCh38
rs748922914 Chr.1: 11073887-11073888 on Build GRCh38
rs781108031 Chr.1: 11073805-11073805 on Build GRCh38
rs1030626097 Chr.1: 11073740-11073740 on Build GRCh38
rs374800195 Chr.1: 11073900-11073901 on Build GRCh38
rs983222228 Chr.1: 11073752-11073752 on Build GRCh38
rs770965502 Chr.1: 11073877-11073878 on Build GRCh38
rs1054968323 Chr.1: 11073656-11073656 on Build GRCh38
rs908605201 Chr.1: 11073615-11073615 on Build GRCh38
rs564553987 Chr.1: 11073911-11073911 on Build GRCh38
rs754754467 Chr.1: 11073759-11073759 on Build GRCh38
rs964385281 Chr.1: 11073701-11073701 on Build GRCh38
rs926999943 Chr.1: 11073791-11073791 on Build GRCh38
rs115976717 Chr.1: 11073948-11073948 on Build GRCh38
rs754546374 Chr.1: 11073888-11073889 on Build GRCh38
rs1042248219 Chr.1: 11073737-11073737 on Build GRCh38
rs566253732 Chr.1: 11074012-11074012 on Build GRCh38
rs768443779 Chr.1: 11074005-11074005 on Build GRCh38
rs71568312 Chr.1: 11073850-11073850 on Build GRCh38
rs771422316 Chr.1: 11073688-11073688 on Build GRCh38
rs200016047 Chr.1: 11073979-11073979 on Build GRCh38
rs568198105 Chr.1: 11073955-11073955 on Build GRCh38
rs745325342 Chr.1: 11073989-11073989 on Build GRCh38
rs752855218 Chr.1: 11073978-11073978 on Build GRCh38
rs779691370 Chr.1: 11073886-11073887 on Build GRCh38
rs369390713 Chr.1: 11073950-11073950 on Build GRCh38
rs904585335 Chr.1: 11073687-11073687 on Build GRCh38
rs530927758 Chr.1: 11073873-11073873 on Build GRCh38
rs907600343 Chr.1: 11073599-11073599 on Build GRCh38
rs373290835 Chr.1: 11073758-11073758 on Build GRCh38
rs973117843 Chr.1: 11073782-11073782 on Build GRCh38
rs1018318328 Chr.1: 11073690-11073690 on Build GRCh38
rs147771843 Chr.1: 11073981-11073981 on Build GRCh38
rs187838179 Chr.1: 11073850-11073850 on Build GRCh38

View Full Product Details