Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11076926-11077468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC10
Entrez Gene ID
5394
Gene Name
exosome component 10
Gene Aliases
PM-Scl, PM/Scl-100, PMSCL, PMSCL2, RRP6, Rrp6p, p2, p3, p4
UniGene
Hs.632368
Gene Chromosome Location
Chr.1: 11066613-11099910 on Build GRCh38
Transcripts
NM_001001998.2 NM_002685.3 XM_005263475.2 XM_017001494.1 XM_017001495.1 XM_017001496.1 XM_017001497.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5641401 c.1859A>G p.Y620C Chr.1: 11077385-11077385 on Build GRCh38
893723 c.1830C>T p.H610H Chr.1: 11077414-11077414 on Build GRCh38
423426 c.1807G>C p.E603Q Chr.1: 11077437-11077437 on Build GRCh38
5377165 c.2011T>C p.W671R Chr.1: 11077390-11077390 on Build GRCh38
2118907 c.1844C>T p.A615V Chr.1: 11077400-11077400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938515292 Chr.1: 11077013-11077014 on Build GRCh38
rs779405374 Chr.1: 11076959-11076959 on Build GRCh38
rs944770595 Chr.1: 11077173-11077173 on Build GRCh38
rs761479912 Chr.1: 11077342-11077342 on Build GRCh38
rs1047850762 Chr.1: 11077419-11077419 on Build GRCh38
rs942398076 Chr.1: 11077090-11077090 on Build GRCh38
rs554713898 Chr.1: 11077143-11077143 on Build GRCh38
rs753531583 Chr.1: 11076993-11076997 on Build GRCh38
rs768502027 Chr.1: 11077433-11077433 on Build GRCh38
rs923523099 Chr.1: 11077363-11077363 on Build GRCh38
rs551658765 Chr.1: 11076994-11076996 on Build GRCh38
rs746910891 Chr.1: 11077279-11077279 on Build GRCh38
rs747087664 Chr.1: 11077382-11077382 on Build GRCh38
rs3737619 Chr.1: 11077237-11077237 on Build GRCh38
rs910621797 Chr.1: 11077145-11077145 on Build GRCh38
rs574442275 Chr.1: 11077290-11077290 on Build GRCh38
rs371930082 Chr.1: 11077369-11077369 on Build GRCh38
rs753213386 Chr.1: 11077357-11077357 on Build GRCh38
rs749227398 Chr.1: 11077427-11077427 on Build GRCh38
rs776529668 Chr.1: 11077284-11077284 on Build GRCh38
rs902121015 Chr.1: 11077105-11077105 on Build GRCh38
rs766154688 Chr.1: 11076989-11076992 on Build GRCh38
rs538787034 Chr.1: 11077076-11077076 on Build GRCh38
rs565893804 Chr.1: 11077085-11077085 on Build GRCh38
rs1003263558 Chr.1: 11077286-11077286 on Build GRCh38
rs532389846 Chr.1: 11077046-11077046 on Build GRCh38
rs1040367727 Chr.1: 11077182-11077182 on Build GRCh38
rs929897324 Chr.1: 11077222-11077222 on Build GRCh38
rs912963923 Chr.1: 11077034-11077034 on Build GRCh38
rs968239347 Chr.1: 11077095-11077095 on Build GRCh38
rs1005370393 Chr.1: 11077273-11077273 on Build GRCh38
rs1037778487 Chr.1: 11077445-11077445 on Build GRCh38
rs988356320 Chr.1: 11077078-11077078 on Build GRCh38
rs757859493 Chr.1: 11077441-11077442 on Build GRCh38
rs1026457698 Chr.1: 11077148-11077148 on Build GRCh38
rs541149821 Chr.1: 11077270-11077270 on Build GRCh38
rs767417779 Chr.1: 11076975-11076975 on Build GRCh38
rs748712010 Chr.1: 11076960-11076960 on Build GRCh38
rs376796152 Chr.1: 11077393-11077393 on Build GRCh38
rs886496120 Chr.1: 11077169-11077169 on Build GRCh38
rs369031958 Chr.1: 11076995-11076995 on Build GRCh38
rs375027201 Chr.1: 11076954-11076954 on Build GRCh38
rs746153780 Chr.1: 11077406-11077406 on Build GRCh38
rs3737618 Chr.1: 11077136-11077136 on Build GRCh38
rs778493046 Chr.1: 11076996-11076997 on Build GRCh38
rs968646606 Chr.1: 11077337-11077337 on Build GRCh38
rs777857502 Chr.1: 11077380-11077380 on Build GRCh38
rs368328243 Chr.1: 11077356-11077356 on Build GRCh38
rs777750877 Chr.1: 11076979-11076979 on Build GRCh38
rs3737617 Chr.1: 11077080-11077080 on Build GRCh38
rs758978498 Chr.1: 11076965-11076965 on Build GRCh38
rs761671077 Chr.1: 11077287-11077287 on Build GRCh38
rs976099323 Chr.1: 11077410-11077410 on Build GRCh38
rs200673142 Chr.1: 11076985-11076985 on Build GRCh38
rs769800074 Chr.1: 11077328-11077328 on Build GRCh38
rs966108022 Chr.1: 11077383-11077383 on Build GRCh38
rs771322224 Chr.1: 11076972-11076972 on Build GRCh38
rs956319687 Chr.1: 11077303-11077303 on Build GRCh38
rs199634937 Chr.1: 11077394-11077394 on Build GRCh38
rs377424028 Chr.1: 11077336-11077336 on Build GRCh38
rs141772850 Chr.1: 11077266-11077266 on Build GRCh38
rs1009860624 Chr.1: 11077318-11077318 on Build GRCh38
rs759413264 Chr.1: 11077315-11077315 on Build GRCh38
rs201435830 Chr.1: 11076966-11076966 on Build GRCh38
rs773873451 Chr.1: 11077437-11077437 on Build GRCh38
rs1006269627 Chr.1: 11077172-11077172 on Build GRCh38
rs778602736 Chr.1: 11077068-11077068 on Build GRCh38
rs776529475 Chr.1: 11077000-11077000 on Build GRCh38
rs372812090 Chr.1: 11077335-11077335 on Build GRCh38
rs936133679 Chr.1: 11077234-11077234 on Build GRCh38
rs768739581 Chr.1: 11077282-11077282 on Build GRCh38
rs752182962 Chr.1: 11077343-11077344 on Build GRCh38
rs767159302 Chr.1: 11077344-11077344 on Build GRCh38
rs755409722 Chr.1: 11076955-11076955 on Build GRCh38
rs956954439 Chr.1: 11077077-11077077 on Build GRCh38
rs369539353 Chr.1: 11077413-11077413 on Build GRCh38
rs576426252 Chr.1: 11077163-11077163 on Build GRCh38
rs368616524 Chr.1: 11077330-11077330 on Build GRCh38
rs376966222 Chr.1: 11077347-11077347 on Build GRCh38
rs769712983 Chr.1: 11077414-11077414 on Build GRCh38
rs1046964332 Chr.1: 11077229-11077229 on Build GRCh38
rs969488752 Chr.1: 11077021-11077021 on Build GRCh38
rs534515362 Chr.1: 11077378-11077378 on Build GRCh38
rs35589071 Chr.1: 11077238-11077238 on Build GRCh38
rs559170110 Chr.1: 11077283-11077283 on Build GRCh38
rs919225935 Chr.1: 11077202-11077202 on Build GRCh38
rs773784614 Chr.1: 11077338-11077338 on Build GRCh38
rs372601184 Chr.1: 11077418-11077418 on Build GRCh38
rs1039277694 Chr.1: 11077101-11077101 on Build GRCh38
rs755321885 Chr.1: 11077349-11077349 on Build GRCh38

View Full Product Details