Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98645212-98645712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APAF1
Entrez Gene ID
317
Gene Name
apoptotic peptidase activating factor 1
Gene Aliases
APAF-1, CED4
UniGene
Hs.552567
Gene Chromosome Location
Chr.12: 98645141-98735433 on Build GRCh38
Transcripts
NM_001160.2 NM_013229.2 NM_181861.1 NM_181868.1 NM_181869.1 XM_017019250.1

SNP Detail

SNP ID Chromosome Location
rs947886126 Chr.12: 98645269-98645269 on Build GRCh38
rs956564671 Chr.12: 98645686-98645686 on Build GRCh38
rs1044797570 Chr.12: 98645283-98645283 on Build GRCh38
rs1047205903 Chr.12: 98645336-98645336 on Build GRCh38
rs927730729 Chr.12: 98645501-98645501 on Build GRCh38
rs568973668 Chr.12: 98645249-98645249 on Build GRCh38
rs930886000 Chr.12: 98645503-98645503 on Build GRCh38
rs957560455 Chr.12: 98645640-98645640 on Build GRCh38
rs921921760 Chr.12: 98645488-98645488 on Build GRCh38
rs939020004 Chr.12: 98645312-98645312 on Build GRCh38
rs1030864592 Chr.12: 98645656-98645656 on Build GRCh38
rs1032118639 Chr.12: 98645437-98645437 on Build GRCh38
rs935269431 Chr.12: 98645260-98645260 on Build GRCh38
rs906289557 Chr.12: 98645308-98645308 on Build GRCh38
rs923745480 Chr.12: 98645270-98645270 on Build GRCh38
rs923878015 Chr.12: 98645255-98645255 on Build GRCh38
rs192062352 Chr.12: 98645433-98645433 on Build GRCh38
rs957408395 Chr.12: 98645439-98645439 on Build GRCh38
rs11109556 Chr.12: 98645389-98645389 on Build GRCh38
rs1035174432 Chr.12: 98645395-98645395 on Build GRCh38
rs772164094 Chr.12: 98645264-98645264 on Build GRCh38
rs955370388 Chr.12: 98645247-98645247 on Build GRCh38
rs911948787 Chr.12: 98645475-98645475 on Build GRCh38
rs980580034 Chr.12: 98645673-98645673 on Build GRCh38
rs922693544 Chr.12: 98645237-98645237 on Build GRCh38
rs989372171 Chr.12: 98645442-98645442 on Build GRCh38
rs1046712859 Chr.12: 98645529-98645529 on Build GRCh38
rs113577911 Chr.12: 98645368-98645368 on Build GRCh38
rs139550753 Chr.12: 98645592-98645592 on Build GRCh38
rs528518338 Chr.12: 98645477-98645477 on Build GRCh38
rs911933081 Chr.12: 98645321-98645321 on Build GRCh38
rs554357301 Chr.12: 98645276-98645276 on Build GRCh38
rs988887412 Chr.12: 98645450-98645450 on Build GRCh38
rs996967929 Chr.12: 98645582-98645582 on Build GRCh38
rs373647874 Chr.12: 98645261-98645261 on Build GRCh38
rs990784730 Chr.12: 98645236-98645236 on Build GRCh38
rs956532118 Chr.12: 98645438-98645438 on Build GRCh38
rs1049267666 Chr.12: 98645380-98645380 on Build GRCh38
rs1023727866 Chr.12: 98645595-98645595 on Build GRCh38
rs980632296 Chr.12: 98645490-98645490 on Build GRCh38
rs367782321 Chr.12: 98645319-98645319 on Build GRCh38
rs2118851 Chr.12: 98645548-98645548 on Build GRCh38
rs1023273658 Chr.12: 98645428-98645428 on Build GRCh38
rs565597531 Chr.12: 98645567-98645567 on Build GRCh38
rs561224847 Chr.12: 98645476-98645476 on Build GRCh38
rs1029696736 Chr.12: 98645422-98645422 on Build GRCh38
rs996980845 Chr.12: 98645416-98645416 on Build GRCh38
rs979283283 Chr.12: 98645434-98645434 on Build GRCh38
rs1049340194 Chr.12: 98645525-98645525 on Build GRCh38
rs930853786 Chr.12: 98645379-98645379 on Build GRCh38
rs372261485 Chr.12: 98645241-98645241 on Build GRCh38
rs534751877 Chr.12: 98645636-98645636 on Build GRCh38
rs999317642 Chr.12: 98645635-98645635 on Build GRCh38
rs576691742 Chr.12: 98645378-98645378 on Build GRCh38
rs955682952 Chr.12: 98645424-98645424 on Build GRCh38
rs941035202 Chr.12: 98645547-98645547 on Build GRCh38
rs969331138 Chr.12: 98645663-98645663 on Build GRCh38
rs116501350 Chr.12: 98645337-98645337 on Build GRCh38
rs886752674 Chr.12: 98645544-98645544 on Build GRCh38
rs371572763 Chr.12: 98645367-98645367 on Build GRCh38
rs1029809676 Chr.12: 98645583-98645583 on Build GRCh38
rs113907867 Chr.12: 98645393-98645393 on Build GRCh38
rs886639437 Chr.12: 98645530-98645530 on Build GRCh38
rs2118852 Chr.12: 98645689-98645689 on Build GRCh38
rs569512005 Chr.12: 98645647-98645647 on Build GRCh38
rs939119783 Chr.12: 98645513-98645513 on Build GRCh38
rs927814739 Chr.12: 98645684-98645684 on Build GRCh38
rs967949512 Chr.12: 98645430-98645430 on Build GRCh38
rs990156761 Chr.12: 98645641-98645641 on Build GRCh38
rs896240801 Chr.12: 98645579-98645579 on Build GRCh38
rs74619561 Chr.12: 98645405-98645405 on Build GRCh38
rs572842999 Chr.12: 98645310-98645310 on Build GRCh38

View Full Product Details