Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98645592-98646087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APAF1
Entrez Gene ID
317
Gene Name
apoptotic peptidase activating factor 1
Gene Aliases
APAF-1, CED4
UniGene
Hs.552567
Gene Chromosome Location
Chr.12: 98645141-98735433 on Build GRCh38
Transcripts
NM_001160.2 NM_013229.2 NM_181861.1 NM_181868.1 NM_181869.1 XM_017019250.1

SNP Detail

SNP ID Chromosome Location
rs956564671 Chr.12: 98645686-98645686 on Build GRCh38
rs770317808 Chr.12: 98645795-98645795 on Build GRCh38
rs957560455 Chr.12: 98645640-98645640 on Build GRCh38
rs893254141 Chr.12: 98645826-98645826 on Build GRCh38
rs548294189 Chr.12: 98645695-98645695 on Build GRCh38
rs1019871295 Chr.12: 98646033-98646033 on Build GRCh38
rs1030864592 Chr.12: 98645656-98645656 on Build GRCh38
rs908289188 Chr.12: 98645954-98645954 on Build GRCh38
rs1057045088 Chr.12: 98645791-98645791 on Build GRCh38
rs566429449 Chr.12: 98645769-98645769 on Build GRCh38
rs1011581401 Chr.12: 98645846-98645846 on Build GRCh38
rs769975027 Chr.12: 98645790-98645790 on Build GRCh38
rs891266588 Chr.12: 98645765-98645765 on Build GRCh38
rs921498087 Chr.12: 98645747-98645747 on Build GRCh38
rs775866105 Chr.12: 98645805-98645805 on Build GRCh38
rs969278972 Chr.12: 98645904-98645904 on Build GRCh38
rs947797225 Chr.12: 98645811-98645811 on Build GRCh38
rs577757125 Chr.12: 98645925-98645925 on Build GRCh38
rs920901854 Chr.12: 98646016-98646016 on Build GRCh38
rs966534951 Chr.12: 98645705-98645705 on Build GRCh38
rs1022993063 Chr.12: 98645877-98645877 on Build GRCh38
rs960790860 Chr.12: 98645939-98645939 on Build GRCh38
rs1000471035 Chr.12: 98645945-98645945 on Build GRCh38
rs999317642 Chr.12: 98645635-98645635 on Build GRCh38
rs534751877 Chr.12: 98645636-98645636 on Build GRCh38
rs908106280 Chr.12: 98645786-98645786 on Build GRCh38
rs1008114272 Chr.12: 98646024-98646024 on Build GRCh38
rs969331138 Chr.12: 98645663-98645663 on Build GRCh38
rs932934983 Chr.12: 98645762-98645762 on Build GRCh38
rs1051364562 Chr.12: 98645764-98645764 on Build GRCh38
rs576079253 Chr.12: 98645830-98645830 on Build GRCh38
rs773325915 Chr.12: 98645957-98645957 on Build GRCh38
rs2118852 Chr.12: 98645689-98645689 on Build GRCh38
rs776457513 Chr.12: 98645858-98645858 on Build GRCh38
rs1038043181 Chr.12: 98645732-98645732 on Build GRCh38
rs569512005 Chr.12: 98645647-98645647 on Build GRCh38
rs980580034 Chr.12: 98645673-98645673 on Build GRCh38
rs113987233 Chr.12: 98646006-98646006 on Build GRCh38
rs763257040 Chr.12: 98645946-98645946 on Build GRCh38
rs941025187 Chr.12: 98645721-98645721 on Build GRCh38
rs1053577258 Chr.12: 98645806-98645806 on Build GRCh38
rs962459517 Chr.12: 98645997-98645997 on Build GRCh38
rs1002090302 Chr.12: 98645920-98645920 on Build GRCh38
rs927814739 Chr.12: 98645684-98645684 on Build GRCh38
rs974816679 Chr.12: 98645710-98645710 on Build GRCh38
rs908296519 Chr.12: 98645701-98645701 on Build GRCh38
rs990156761 Chr.12: 98645641-98645641 on Build GRCh38

View Full Product Details