Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98662363-98662911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APAF1
Entrez Gene ID
317
Gene Name
apoptotic peptidase activating factor 1
Gene Aliases
APAF-1, CED4
UniGene
Hs.552567
Gene Chromosome Location
Chr.12: 98645141-98735433 on Build GRCh38
Transcripts
NM_001160.2 NM_013229.2 NM_181861.1 NM_181868.1 NM_181869.1 XM_017019250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
696225 c.945A>T p.K315N Chr.12: 98662796-98662796 on Build GRCh38
6074177 c.771G>A p.Q257Q Chr.12: 98662516-98662516 on Build GRCh38
5376260 c.854C>T p.S285F Chr.12: 98662705-98662705 on Build GRCh38
318717 c.793A>G p.R265G Chr.12: 98662538-98662538 on Build GRCh38
5423516 c.824-10T>C p.? Chr.12: 98662665-98662665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371399207 Chr.12: 98662425-98662425 on Build GRCh38
rs758870589 Chr.12: 98662428-98662428 on Build GRCh38
rs753107484 Chr.12: 98662813-98662816 on Build GRCh38
rs942947636 Chr.12: 98662667-98662667 on Build GRCh38
rs775507125 Chr.12: 98662491-98662491 on Build GRCh38
rs750499402 Chr.12: 98662777-98662777 on Build GRCh38
rs756319379 Chr.12: 98662456-98662456 on Build GRCh38
rs756411622 Chr.12: 98662820-98662823 on Build GRCh38
rs144728788 Chr.12: 98662710-98662710 on Build GRCh38
rs770864896 Chr.12: 98662488-98662488 on Build GRCh38
rs753042213 Chr.12: 98662602-98662602 on Build GRCh38
rs771508556 Chr.12: 98662834-98662834 on Build GRCh38
rs778075105 Chr.12: 98662610-98662610 on Build GRCh38
rs139788089 Chr.12: 98662811-98662811 on Build GRCh38
rs752069194 Chr.12: 98662591-98662591 on Build GRCh38
rs199783677 Chr.12: 98662550-98662550 on Build GRCh38
rs779183383 Chr.12: 98662763-98662763 on Build GRCh38
rs769637846 Chr.12: 98662805-98662805 on Build GRCh38
rs745909861 Chr.12: 98662524-98662526 on Build GRCh38
rs145270473 Chr.12: 98662526-98662526 on Build GRCh38
rs552228477 Chr.12: 98662448-98662448 on Build GRCh38
rs773894374 Chr.12: 98662826-98662826 on Build GRCh38
rs769974597 Chr.12: 98662753-98662753 on Build GRCh38
rs1046826037 Chr.12: 98662728-98662728 on Build GRCh38
rs773840959 Chr.12: 98662671-98662671 on Build GRCh38
rs1018119143 Chr.12: 98662668-98662668 on Build GRCh38
rs768637465 Chr.12: 98662518-98662518 on Build GRCh38
rs537644742 Chr.12: 98662460-98662460 on Build GRCh38
rs753148886 Chr.12: 98662723-98662723 on Build GRCh38
rs368863812 Chr.12: 98662652-98662652 on Build GRCh38
rs772701293 Chr.12: 98662579-98662579 on Build GRCh38
rs773514371 Chr.12: 98662432-98662435 on Build GRCh38
rs770472412 Chr.12: 98662818-98662818 on Build GRCh38
rs767905121 Chr.12: 98662732-98662732 on Build GRCh38
rs766941395 Chr.12: 98662438-98662438 on Build GRCh38
rs544169539 Chr.12: 98662809-98662809 on Build GRCh38
rs768692708 Chr.12: 98662641-98662641 on Build GRCh38
rs757360181 Chr.12: 98662617-98662617 on Build GRCh38
rs192898520 Chr.12: 98662788-98662788 on Build GRCh38
rs781301099 Chr.12: 98662796-98662796 on Build GRCh38
rs778853424 Chr.12: 98662628-98662628 on Build GRCh38
rs201309144 Chr.12: 98662582-98662582 on Build GRCh38
rs751568034 Chr.12: 98662438-98662439 on Build GRCh38
rs764913306 Chr.12: 98662574-98662574 on Build GRCh38
rs766516357 Chr.12: 98662439-98662439 on Build GRCh38
rs376462245 Chr.12: 98662632-98662632 on Build GRCh38
rs1048377806 Chr.12: 98662613-98662613 on Build GRCh38
rs749410709 Chr.12: 98662611-98662611 on Build GRCh38
rs761550622 Chr.12: 98662416-98662416 on Build GRCh38
rs760089515 Chr.12: 98662714-98662714 on Build GRCh38
rs756390473 Chr.12: 98662604-98662604 on Build GRCh38
rs762135366 Chr.12: 98662851-98662851 on Build GRCh38
rs748107954 Chr.12: 98662438-98662438 on Build GRCh38
rs764710758 Chr.12: 98662420-98662420 on Build GRCh38
rs746875464 Chr.12: 98662517-98662517 on Build GRCh38
rs950176672 Chr.12: 98662533-98662533 on Build GRCh38
rs765587353 Chr.12: 98662855-98662855 on Build GRCh38
rs781471884 Chr.12: 98662449-98662449 on Build GRCh38
rs759065424 Chr.12: 98662831-98662831 on Build GRCh38
rs773664804 Chr.12: 98662535-98662535 on Build GRCh38
rs776607206 Chr.12: 98662647-98662647 on Build GRCh38
rs769427886 Chr.12: 98662653-98662653 on Build GRCh38
rs754127964 Chr.12: 98662843-98662843 on Build GRCh38
rs755473881 Chr.12: 98662597-98662597 on Build GRCh38
rs573318327 Chr.12: 98662656-98662656 on Build GRCh38
rs749412360 Chr.12: 98662471-98662471 on Build GRCh38
rs747979101 Chr.12: 98662802-98662802 on Build GRCh38
rs200771041 Chr.12: 98662640-98662640 on Build GRCh38
rs138616141 Chr.12: 98662715-98662715 on Build GRCh38
rs74552945 Chr.12: 98662440-98662440 on Build GRCh38
rs570685568 Chr.12: 98662451-98662451 on Build GRCh38
rs544496420 Chr.12: 98662838-98662838 on Build GRCh38
rs201562861 Chr.12: 98662749-98662749 on Build GRCh38
rs978990703 Chr.12: 98662389-98662389 on Build GRCh38
rs775597203 Chr.12: 98662800-98662801 on Build GRCh38
rs111885484 Chr.12: 98662552-98662552 on Build GRCh38
rs189237320 Chr.12: 98662762-98662762 on Build GRCh38
rs765984017 Chr.12: 98662665-98662665 on Build GRCh38
rs894424376 Chr.12: 98662863-98662863 on Build GRCh38
rs149321598 Chr.12: 98662761-98662761 on Build GRCh38
rs79290726 Chr.12: 98662581-98662581 on Build GRCh38
rs762447515 Chr.12: 98662657-98662657 on Build GRCh38
rs765565025 Chr.12: 98662413-98662415 on Build GRCh38
rs779199907 Chr.12: 98662512-98662512 on Build GRCh38
rs369909924 Chr.12: 98662798-98662798 on Build GRCh38
rs764575621 Chr.12: 98662841-98662841 on Build GRCh38
rs898651129 Chr.12: 98662684-98662684 on Build GRCh38

View Full Product Details