Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100617507-100618040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS2L3
Entrez Gene ID
283431
Gene Name
growth arrest specific 2 like 3
Gene Aliases
G2L3
UniGene
Hs.20575
Gene Chromosome Location
Chr.12: 100573661-100628288 on Build GRCh38
Transcripts
NM_001303130.1 NM_001303131.1 NM_174942.2 XM_005268804.3 XM_011538219.2 XM_011538220.2 XM_011538221.2 XM_017019206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5051258 c.509+2T>C p.? Chr.12: 100617809-100617809 on Build GRCh38
4497448 c.496C>T p.R166* Chr.12: 100617794-100617794 on Build GRCh38
5692460 c.446-2A>T p.? Chr.12: 100617742-100617742 on Build GRCh38
1207929 c.472T>C p.Y158H Chr.12: 100617770-100617770 on Build GRCh38
4882670 c.488A>G p.E163G Chr.12: 100617786-100617786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772717576 Chr.12: 100617756-100617756 on Build GRCh38
rs754474377 Chr.12: 100617785-100617785 on Build GRCh38
rs1007696555 Chr.12: 100617600-100617601 on Build GRCh38
rs902011847 Chr.12: 100617981-100617981 on Build GRCh38
rs750819579 Chr.12: 100617740-100617740 on Build GRCh38
rs1003862960 Chr.12: 100617882-100617882 on Build GRCh38
rs980934440 Chr.12: 100617915-100617915 on Build GRCh38
rs568968694 Chr.12: 100617805-100617805 on Build GRCh38
rs1054174132 Chr.12: 100617566-100617566 on Build GRCh38
rs1006719983 Chr.12: 100617691-100617691 on Build GRCh38
rs60123844 Chr.12: 100617643-100617643 on Build GRCh38
rs1026306675 Chr.12: 100617678-100617678 on Build GRCh38
rs886406587 Chr.12: 100617690-100617690 on Build GRCh38
rs949929962 Chr.12: 100617556-100617557 on Build GRCh38
rs183921162 Chr.12: 100617564-100617564 on Build GRCh38
rs759595301 Chr.12: 100617834-100617834 on Build GRCh38
rs766375184 Chr.12: 100617807-100617807 on Build GRCh38
rs1025088681 Chr.12: 100617892-100617892 on Build GRCh38
rs751691347 Chr.12: 100617996-100617996 on Build GRCh38
rs757538599 Chr.12: 100617715-100617715 on Build GRCh38
rs936534951 Chr.12: 100617614-100617614 on Build GRCh38
rs1039153826 Chr.12: 100617767-100617767 on Build GRCh38
rs765506754 Chr.12: 100617850-100617850 on Build GRCh38
rs886456589 Chr.12: 100617995-100617995 on Build GRCh38
rs201874715 Chr.12: 100617699-100617699 on Build GRCh38
rs899342337 Chr.12: 100617798-100617798 on Build GRCh38
rs759497804 Chr.12: 100617859-100617859 on Build GRCh38
rs186658303 Chr.12: 100617661-100617661 on Build GRCh38
rs374154516 Chr.12: 100617800-100617800 on Build GRCh38
rs150008707 Chr.12: 100617895-100617896 on Build GRCh38
rs572875678 Chr.12: 100617986-100617986 on Build GRCh38
rs774416187 Chr.12: 100617827-100617827 on Build GRCh38
rs765184088 Chr.12: 100617909-100617909 on Build GRCh38
rs780732441 Chr.12: 100617789-100617789 on Build GRCh38
rs191513607 Chr.12: 100617836-100617836 on Build GRCh38
rs747726961 Chr.12: 100617792-100617792 on Build GRCh38
rs970905773 Chr.12: 100617547-100617547 on Build GRCh38
rs780540035 Chr.12: 100617762-100617762 on Build GRCh38
rs990317476 Chr.12: 100617890-100617890 on Build GRCh38
rs771641414 Chr.12: 100617700-100617700 on Build GRCh38
rs926463109 Chr.12: 100617567-100617567 on Build GRCh38
rs1006250938 Chr.12: 100618007-100618007 on Build GRCh38
rs750683002 Chr.12: 100617856-100617856 on Build GRCh38
rs913856915 Chr.12: 100617647-100617647 on Build GRCh38
rs934415334 Chr.12: 100617565-100617565 on Build GRCh38
rs776801349 Chr.12: 100617838-100617838 on Build GRCh38
rs761889392 Chr.12: 100617844-100617844 on Build GRCh38
rs752130851 Chr.12: 100617766-100617766 on Build GRCh38
rs760058489 Chr.12: 100617771-100617771 on Build GRCh38
rs369578521 Chr.12: 100617716-100617716 on Build GRCh38
rs770925403 Chr.12: 100617826-100617826 on Build GRCh38
rs943091948 Chr.12: 100617903-100617903 on Build GRCh38
rs1033480561 Chr.12: 100617943-100617943 on Build GRCh38
rs188644318 Chr.12: 100617987-100617987 on Build GRCh38
rs1045463870 Chr.12: 100617557-100617557 on Build GRCh38
rs769519509 Chr.12: 100617795-100617795 on Build GRCh38
rs957851658 Chr.12: 100617961-100617961 on Build GRCh38
rs183920990 Chr.12: 100617973-100617973 on Build GRCh38
rs146806070 Chr.12: 100617757-100617757 on Build GRCh38

View Full Product Details