Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100974656-100975151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO4
Entrez Gene ID
121601
Gene Name
anoctamin 4
Gene Aliases
TMEM16D
UniGene
Hs.58785
Gene Chromosome Location
Chr.12: 100717457-101128641 on Build GRCh38
Transcripts
NM_001286615.1 NM_001286616.1 NM_178826.3 XM_011537911.2 XM_011537912.2 XM_011537913.2 XM_011537914.2 XM_011537915.2 XM_011537916.2 XM_011537918.2 XM_017018808.1 XM_017018809.1 XM_017018810.1 XM_017018811.1 XM_017018812.1 XM_017018813.1 XM_017018814.1 XM_017018815.1 XM_017018816.1 XM_017018817.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4590381 c.467A>C p.Y156S Chr.12: 100974859-100974859 on Build GRCh38
933870 c.489C>A p.F163L Chr.12: 100974881-100974881 on Build GRCh38
933869 c.462C>A p.I154I Chr.12: 100974854-100974854 on Build GRCh38
691695 c.456A>T p.R152S Chr.12: 100974848-100974848 on Build GRCh38
1470519 c.478C>G p.R160G Chr.12: 100974870-100974870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774199585 Chr.12: 100974937-100974937 on Build GRCh38
rs749904401 Chr.12: 100974814-100974814 on Build GRCh38
rs778988665 Chr.12: 100974871-100974871 on Build GRCh38
rs936796935 Chr.12: 100974855-100974855 on Build GRCh38
rs745933521 Chr.12: 100974894-100974894 on Build GRCh38
rs768174700 Chr.12: 100974754-100974754 on Build GRCh38
rs79379625 Chr.12: 100974862-100974862 on Build GRCh38
rs377503312 Chr.12: 100975080-100975080 on Build GRCh38
rs543313023 Chr.12: 100975040-100975040 on Build GRCh38
rs1033625466 Chr.12: 100975097-100975097 on Build GRCh38
rs779296268 Chr.12: 100974939-100974940 on Build GRCh38
rs781164710 Chr.12: 100974866-100974866 on Build GRCh38
rs747258486 Chr.12: 100974904-100974904 on Build GRCh38
rs771912966 Chr.12: 100975079-100975079 on Build GRCh38
rs761720673 Chr.12: 100974993-100974993 on Build GRCh38
rs1019152047 Chr.12: 100975051-100975051 on Build GRCh38
rs963913200 Chr.12: 100974963-100974963 on Build GRCh38
rs777078363 Chr.12: 100974787-100974787 on Build GRCh38
rs768956820 Chr.12: 100974910-100974910 on Build GRCh38
rs775852343 Chr.12: 100974763-100974763 on Build GRCh38
rs771096911 Chr.12: 100974750-100974750 on Build GRCh38
rs890911520 Chr.12: 100974908-100974908 on Build GRCh38
rs370605819 Chr.12: 100974863-100974864 on Build GRCh38
rs765934326 Chr.12: 100974858-100974858 on Build GRCh38
rs958646327 Chr.12: 100975127-100975127 on Build GRCh38
rs762589282 Chr.12: 100974856-100974856 on Build GRCh38
rs879724423 Chr.12: 100974683-100974683 on Build GRCh38
rs577783801 Chr.12: 100975015-100975015 on Build GRCh38
rs764744094 Chr.12: 100974804-100974804 on Build GRCh38
rs572208504 Chr.12: 100974901-100974901 on Build GRCh38
rs769044992 Chr.12: 100974781-100974781 on Build GRCh38
rs897966391 Chr.12: 100975063-100975063 on Build GRCh38
rs746047695 Chr.12: 100974756-100974756 on Build GRCh38
rs980875397 Chr.12: 100975107-100975107 on Build GRCh38
rs1018566135 Chr.12: 100974952-100974952 on Build GRCh38
rs752635853 Chr.12: 100974868-100974868 on Build GRCh38
rs374420550 Chr.12: 100974918-100974918 on Build GRCh38
rs144445830 Chr.12: 100974870-100974870 on Build GRCh38
rs762238316 Chr.12: 100974924-100974924 on Build GRCh38
rs949452635 Chr.12: 100974704-100974704 on Build GRCh38
rs199907380 Chr.12: 100974863-100974863 on Build GRCh38
rs376727483 Chr.12: 100974799-100974799 on Build GRCh38
rs770194804 Chr.12: 100974935-100974935 on Build GRCh38
rs776990326 Chr.12: 100974911-100974911 on Build GRCh38
rs878855567 Chr.12: 100974847-100974847 on Build GRCh38
rs904190659 Chr.12: 100974860-100974860 on Build GRCh38
rs772191337 Chr.12: 100974759-100974759 on Build GRCh38
rs373066453 Chr.12: 100975068-100975068 on Build GRCh38
rs182129173 Chr.12: 100974967-100974967 on Build GRCh38
rs761017640 Chr.12: 100974775-100974775 on Build GRCh38
rs773826116 Chr.12: 100974941-100974941 on Build GRCh38
rs548999455 Chr.12: 100975088-100975088 on Build GRCh38
rs905351425 Chr.12: 100974842-100974842 on Build GRCh38
rs571692013 Chr.12: 100975078-100975078 on Build GRCh38
rs201084190 Chr.12: 100974859-100974859 on Build GRCh38
rs78462085 Chr.12: 100974990-100974990 on Build GRCh38
rs564098202 Chr.12: 100974978-100974978 on Build GRCh38
rs892694744 Chr.12: 100974867-100974867 on Build GRCh38
rs917903190 Chr.12: 100975082-100975082 on Build GRCh38
rs187117646 Chr.12: 100975044-100975044 on Build GRCh38

View Full Product Details