Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101914005-101914439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRAM1
Entrez Gene ID
55332
Gene Name
DNA damage regulated autophagy modulator 1
Gene Aliases
DRAM
UniGene
Hs.525634
Gene Chromosome Location
Chr.12: 101877327-101923623 on Build GRCh38
Transcripts
NM_018370.2 XM_005269004.2 XM_005269005.2 XM_017019578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2063709 c.213T>C p.C71C Chr.12: 101914184-101914184 on Build GRCh38
4641601 c.239T>C p.L80P Chr.12: 101914210-101914210 on Build GRCh38
2063706 c.529T>C p.C177R Chr.12: 101914182-101914182 on Build GRCh38
3710653 c.204G>T p.M68I Chr.12: 101914175-101914175 on Build GRCh38
3710652 c.522G>T p.M174I Chr.12: 101914175-101914175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764552698 Chr.12: 101914179-101914179 on Build GRCh38
rs761922080 Chr.12: 101914097-101914097 on Build GRCh38
rs1018095027 Chr.12: 101914279-101914279 on Build GRCh38
rs181747532 Chr.12: 101914305-101914305 on Build GRCh38
rs769597842 Chr.12: 101914269-101914269 on Build GRCh38
rs761733169 Chr.12: 101914129-101914129 on Build GRCh38
rs559922011 Chr.12: 101914336-101914336 on Build GRCh38
rs759893063 Chr.12: 101914233-101914234 on Build GRCh38
rs752804419 Chr.12: 101914171-101914171 on Build GRCh38
rs6539020 Chr.12: 101914412-101914412 on Build GRCh38
rs186329557 Chr.12: 101914411-101914411 on Build GRCh38
rs1018552513 Chr.12: 101914399-101914399 on Build GRCh38
rs982623302 Chr.12: 101914340-101914340 on Build GRCh38
rs764464588 Chr.12: 101914283-101914283 on Build GRCh38
rs373316104 Chr.12: 101914167-101914167 on Build GRCh38
rs980657788 Chr.12: 101914398-101914398 on Build GRCh38
rs929872473 Chr.12: 101914073-101914073 on Build GRCh38
rs752674922 Chr.12: 101914379-101914379 on Build GRCh38
rs766240540 Chr.12: 101914200-101914200 on Build GRCh38
rs949233569 Chr.12: 101914389-101914389 on Build GRCh38
rs759483969 Chr.12: 101914154-101914154 on Build GRCh38
rs375744199 Chr.12: 101914190-101914190 on Build GRCh38
rs973477368 Chr.12: 101914309-101914309 on Build GRCh38
rs779515969 Chr.12: 101914205-101914205 on Build GRCh38
rs188940742 Chr.12: 101914265-101914265 on Build GRCh38
rs115247447 Chr.12: 101914313-101914313 on Build GRCh38
rs746625555 Chr.12: 101914236-101914236 on Build GRCh38
rs761016155 Chr.12: 101914176-101914176 on Build GRCh38
rs534346562 Chr.12: 101914183-101914183 on Build GRCh38
rs779626384 Chr.12: 101914225-101914225 on Build GRCh38
rs200817899 Chr.12: 101914209-101914209 on Build GRCh38
rs1053469717 Chr.12: 101914213-101914213 on Build GRCh38
rs774390362 Chr.12: 101914147-101914147 on Build GRCh38
rs774935938 Chr.12: 101914146-101914146 on Build GRCh38
rs60930790 Chr.12: 101914081-101914081 on Build GRCh38
rs7955730 Chr.12: 101914317-101914317 on Build GRCh38
rs753648410 Chr.12: 101914254-101914254 on Build GRCh38
rs376472149 Chr.12: 101914272-101914272 on Build GRCh38
rs1020474114 Chr.12: 101914284-101914284 on Build GRCh38
rs765199025 Chr.12: 101914145-101914145 on Build GRCh38
rs1048951580 Chr.12: 101914262-101914262 on Build GRCh38
rs759572688 Chr.12: 101914264-101914264 on Build GRCh38
rs543711831 Chr.12: 101914293-101914293 on Build GRCh38
rs987115470 Chr.12: 101914392-101914392 on Build GRCh38
rs950728687 Chr.12: 101914334-101914334 on Build GRCh38
rs982677523 Chr.12: 101914080-101914080 on Build GRCh38
rs750995412 Chr.12: 101914207-101914207 on Build GRCh38

View Full Product Details