Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101923201-101923712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRAM1
Entrez Gene ID
55332
Gene Name
DNA damage regulated autophagy modulator 1
Gene Aliases
DRAM
UniGene
Hs.525634
Gene Chromosome Location
Chr.12: 101877327-101923623 on Build GRCh38
Transcripts
NM_018370.2 XM_005269004.2 XM_005269005.2 XM_017019578.1

SNP Detail

SNP ID Chromosome Location
rs116370125 Chr.12: 101923254-101923254 on Build GRCh38
rs948180961 Chr.12: 101923312-101923312 on Build GRCh38
rs547425653 Chr.12: 101923386-101923386 on Build GRCh38
rs577279046 Chr.12: 101923411-101923411 on Build GRCh38
rs777708332 Chr.12: 101923373-101923373 on Build GRCh38
rs554640524 Chr.12: 101923500-101923500 on Build GRCh38
rs568042197 Chr.12: 101923576-101923576 on Build GRCh38
rs56363228 Chr.12: 101923277-101923277 on Build GRCh38
rs781706279 Chr.12: 101923262-101923262 on Build GRCh38
rs1030435803 Chr.12: 101923649-101923649 on Build GRCh38
rs760615951 Chr.12: 101923686-101923686 on Build GRCh38
rs148342456 Chr.12: 101923615-101923615 on Build GRCh38
rs536102633 Chr.12: 101923491-101923491 on Build GRCh38
rs1038307915 Chr.12: 101923306-101923306 on Build GRCh38
rs773065624 Chr.12: 101923619-101923619 on Build GRCh38
rs35105108 Chr.12: 101923632-101923633 on Build GRCh38
rs907356306 Chr.12: 101923630-101923630 on Build GRCh38
rs79685195 Chr.12: 101923596-101923596 on Build GRCh38
rs550529228 Chr.12: 101923482-101923482 on Build GRCh38
rs570401686 Chr.12: 101923351-101923351 on Build GRCh38
rs954764500 Chr.12: 101923654-101923654 on Build GRCh38
rs186541210 Chr.12: 101923259-101923259 on Build GRCh38
rs570239328 Chr.12: 101923486-101923486 on Build GRCh38
rs749547361 Chr.12: 101923485-101923485 on Build GRCh38
rs562089908 Chr.12: 101923340-101923340 on Build GRCh38
rs7885 Chr.12: 101923454-101923454 on Build GRCh38
rs548257308 Chr.12: 101923309-101923309 on Build GRCh38
rs746351272 Chr.12: 101923293-101923293 on Build GRCh38
rs1018574576 Chr.12: 101923527-101923527 on Build GRCh38
rs893755287 Chr.12: 101923474-101923474 on Build GRCh38
rs543695062 Chr.12: 101923620-101923620 on Build GRCh38
rs557906440 Chr.12: 101923511-101923511 on Build GRCh38
rs778544974 Chr.12: 101923646-101923646 on Build GRCh38
rs540102246 Chr.12: 101923478-101923478 on Build GRCh38
rs10823 Chr.12: 101923363-101923363 on Build GRCh38

View Full Product Details