Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102043725-102044225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC53
Entrez Gene ID
51019
Gene Name
coiled-coil domain containing 53
Gene Aliases
CGI-116
UniGene
Hs.405692
Gene Chromosome Location
Chr.12: 102012840-102062124 on Build GRCh38
Transcripts
NM_001301107.1 NM_016053.3 XM_011538423.1 XM_017019383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
467668 c.237A>G p.L79L Chr.12: 102044192-102044192 on Build GRCh38
430183 c.324+1G>T p.? Chr.12: 102044104-102044104 on Build GRCh38
6324790 c.306C>A p.A102A Chr.12: 102044123-102044123 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778173624 Chr.12: 102044099-102044102 on Build GRCh38
rs770598097 Chr.12: 102044128-102044128 on Build GRCh38
rs370467761 Chr.12: 102044084-102044084 on Build GRCh38
rs147510623 Chr.12: 102044100-102044100 on Build GRCh38
rs552905935 Chr.12: 102043986-102043986 on Build GRCh38
rs940352272 Chr.12: 102043765-102043765 on Build GRCh38
rs748588154 Chr.12: 102044110-102044110 on Build GRCh38
rs750539269 Chr.12: 102044076-102044076 on Build GRCh38
rs201459333 Chr.12: 102044181-102044181 on Build GRCh38
rs554926968 Chr.12: 102044006-102044007 on Build GRCh38
rs115247989 Chr.12: 102043981-102043981 on Build GRCh38
rs911522332 Chr.12: 102043760-102043760 on Build GRCh38
rs371181494 Chr.12: 102044196-102044196 on Build GRCh38
rs1010209658 Chr.12: 102043913-102043913 on Build GRCh38
rs777400483 Chr.12: 102044120-102044120 on Build GRCh38
rs1048549363 Chr.12: 102043793-102043797 on Build GRCh38
rs768625901 Chr.12: 102044189-102044189 on Build GRCh38
rs73382854 Chr.12: 102044085-102044085 on Build GRCh38
rs973915388 Chr.12: 102043788-102043788 on Build GRCh38
rs949303652 Chr.12: 102043922-102043922 on Build GRCh38
rs898898541 Chr.12: 102043781-102043781 on Build GRCh38
rs771754268 Chr.12: 102044171-102044171 on Build GRCh38
rs374642397 Chr.12: 102044185-102044185 on Build GRCh38
rs182922983 Chr.12: 102044195-102044195 on Build GRCh38
rs756751291 Chr.12: 102044115-102044115 on Build GRCh38
rs950783783 Chr.12: 102043819-102043819 on Build GRCh38
rs1026358310 Chr.12: 102043818-102043818 on Build GRCh38
rs113452619 Chr.12: 102043791-102043791 on Build GRCh38
rs751252611 Chr.12: 102043890-102043890 on Build GRCh38
rs963548884 Chr.12: 102043787-102043787 on Build GRCh38
rs1020263749 Chr.12: 102043967-102043967 on Build GRCh38
rs542882642 Chr.12: 102044103-102044103 on Build GRCh38
rs781594560 Chr.12: 102044194-102044194 on Build GRCh38
rs778364917 Chr.12: 102044138-102044138 on Build GRCh38
rs532637842 Chr.12: 102044121-102044121 on Build GRCh38
rs773115383 Chr.12: 102043982-102043982 on Build GRCh38
rs765300992 Chr.12: 102044075-102044075 on Build GRCh38
rs760667950 Chr.12: 102044073-102044073 on Build GRCh38
rs566850310 Chr.12: 102044105-102044105 on Build GRCh38
rs775235274 Chr.12: 102044063-102044063 on Build GRCh38
rs536859682 Chr.12: 102043786-102043786 on Build GRCh38
rs192772507 Chr.12: 102043845-102043845 on Build GRCh38
rs866472985 Chr.12: 102044142-102044142 on Build GRCh38
rs745549584 Chr.12: 102044151-102044151 on Build GRCh38
rs895789980 Chr.12: 102043848-102043848 on Build GRCh38
rs561428463 Chr.12: 102044018-102044018 on Build GRCh38
rs749462374 Chr.12: 102044162-102044163 on Build GRCh38
rs1036090320 Chr.12: 102043767-102043767 on Build GRCh38
rs913686586 Chr.12: 102044089-102044089 on Build GRCh38

View Full Product Details