Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102959685-102960196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCL1
Entrez Gene ID
429
Gene Name
achaete-scute family bHLH transcription factor 1
Gene Aliases
ASH1, HASH1, MASH1, bHLHa46
UniGene
Hs.703025
Gene Chromosome Location
Chr.12: 102957674-102960516 on Build GRCh38
Transcripts
NM_004316.3

SNP Detail

SNP ID Chromosome Location
rs1038988936 Chr.12: 102959986-102959986 on Build GRCh38
rs968726843 Chr.12: 102959860-102959860 on Build GRCh38
rs893031985 Chr.12: 102959746-102959746 on Build GRCh38
rs1391682 Chr.12: 102959747-102959747 on Build GRCh38
rs931720373 Chr.12: 102959972-102959972 on Build GRCh38
rs548027213 Chr.12: 102959823-102959823 on Build GRCh38
rs192574985 Chr.12: 102959730-102959730 on Build GRCh38
rs966241049 Chr.12: 102959894-102959894 on Build GRCh38
rs769986010 Chr.12: 102960003-102960006 on Build GRCh38
rs893002504 Chr.12: 102959974-102959974 on Build GRCh38
rs1036288969 Chr.12: 102960107-102960107 on Build GRCh38
rs547553580 Chr.12: 102959881-102959881 on Build GRCh38
rs921648512 Chr.12: 102959744-102959744 on Build GRCh38
rs1016427896 Chr.12: 102960095-102960095 on Build GRCh38
rs963572537 Chr.12: 102960099-102960099 on Build GRCh38
rs531328423 Chr.12: 102959802-102959802 on Build GRCh38
rs907048203 Chr.12: 102959811-102959811 on Build GRCh38
rs183912670 Chr.12: 102959761-102959761 on Build GRCh38
rs77898161 Chr.12: 102959929-102959929 on Build GRCh38
rs184842404 Chr.12: 102960127-102960127 on Build GRCh38
rs991097417 Chr.12: 102959836-102959836 on Build GRCh38
rs2291854 Chr.12: 102959760-102959760 on Build GRCh38
rs1045817621 Chr.12: 102959887-102959887 on Build GRCh38
rs1011441285 Chr.12: 102959983-102959983 on Build GRCh38
rs534164207 Chr.12: 102959851-102959851 on Build GRCh38
rs201630307 Chr.12: 102960058-102960058 on Build GRCh38
rs189036038 Chr.12: 102960045-102960045 on Build GRCh38
rs539695622 Chr.12: 102960027-102960027 on Build GRCh38
rs940600334 Chr.12: 102959787-102959787 on Build GRCh38
rs769207585 Chr.12: 102959987-102959987 on Build GRCh38
rs144024151 Chr.12: 102960059-102960062 on Build GRCh38
rs1030376150 Chr.12: 102959772-102959772 on Build GRCh38
rs868712126 Chr.12: 102959816-102959816 on Build GRCh38
rs770172230 Chr.12: 102959820-102959820 on Build GRCh38
rs375561937 Chr.12: 102960021-102960021 on Build GRCh38
rs978382950 Chr.12: 102959868-102959868 on Build GRCh38
rs755204233 Chr.12: 102960054-102960055 on Build GRCh38
rs950870912 Chr.12: 102959793-102959793 on Build GRCh38
rs774644837 Chr.12: 102960022-102960022 on Build GRCh38
rs1017546756 Chr.12: 102959757-102959757 on Build GRCh38
rs1031639469 Chr.12: 102959889-102959889 on Build GRCh38
rs540405976 Chr.12: 102959740-102959740 on Build GRCh38
rs900418785 Chr.12: 102959989-102959989 on Build GRCh38
rs975866988 Chr.12: 102959713-102959713 on Build GRCh38
rs887724868 Chr.12: 102959786-102959786 on Build GRCh38
rs576398539 Chr.12: 102960062-102960062 on Build GRCh38
rs146349711 Chr.12: 102960055-102960058 on Build GRCh38
rs1004992987 Chr.12: 102960081-102960081 on Build GRCh38
rs746670132 Chr.12: 102959733-102959733 on Build GRCh38
rs1012286181 Chr.12: 102959840-102959840 on Build GRCh38
rs371263751 Chr.12: 102960058-102960061 on Build GRCh38
rs916805773 Chr.12: 102960115-102960115 on Build GRCh38
rs998614319 Chr.12: 102959732-102959732 on Build GRCh38
rs971400768 Chr.12: 102960139-102960139 on Build GRCh38
rs181547992 Chr.12: 102960109-102960109 on Build GRCh38
rs776116075 Chr.12: 102959915-102959915 on Build GRCh38
rs532413844 Chr.12: 102959748-102959748 on Build GRCh38
rs568127115 Chr.12: 102959837-102959837 on Build GRCh38
rs963603510 Chr.12: 102959827-102959827 on Build GRCh38
rs537464449 Chr.12: 102959811-102959812 on Build GRCh38
rs984445400 Chr.12: 102960016-102960016 on Build GRCh38
rs940321276 Chr.12: 102960036-102960036 on Build GRCh38
rs934432104 Chr.12: 102959896-102959896 on Build GRCh38
rs997489892 Chr.12: 102960018-102960018 on Build GRCh38
rs372307339 Chr.12: 102959866-102959866 on Build GRCh38

View Full Product Details