Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 104304417-104304929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EID3
Entrez Gene ID
493861
Gene Name
EP300 interacting inhibitor of differentiation 3
Gene Aliases
NS4EB, NSE4B, NSMCE4B
UniGene
Hs.659857
Gene Chromosome Location
Chr.12: 104303732-104305205 on Build GRCh38
Transcripts
NM_001008394.2 NM_001093771.2 NM_001261445.1 NM_001261446.1 NM_003330.3 NM_182729.2 NM_182742.2 NM_182743.2

Gene Symbol
TXNRD1
Entrez Gene ID
7296
Gene Name
thioredoxin reductase 1
Gene Aliases
GRIM-12, TR, TR1, TRXR1, TXNR
UniGene
Hs.654922
Gene Chromosome Location
Chr.12: 104215779-104350307 on Build GRCh38
Transcripts
NM_001008394.2 NM_001093771.2 NM_001261445.1 NM_001261446.1 NM_003330.3 NM_182729.2 NM_182742.2 NM_182743.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6232389 c.995C>T p.S332L Chr.12: 104304929-104304929 on Build GRCh38
6231985 c.986C>T p.T329I Chr.12: 104304920-104304920 on Build GRCh38
4038183 c.536G>A p.R179H Chr.12: 104304470-104304470 on Build GRCh38
6272840 c.574G>T p.V192F Chr.12: 104304508-104304508 on Build GRCh38
6234506 c.886G>T p.D296Y Chr.12: 104304820-104304820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74722368 Chr.12: 104304709-104304709 on Build GRCh38
rs748245058 Chr.12: 104304776-104304776 on Build GRCh38
rs745652267 Chr.12: 104304739-104304739 on Build GRCh38
rs767578368 Chr.12: 104304451-104304451 on Build GRCh38
rs775933640 Chr.12: 104304801-104304801 on Build GRCh38
rs750721767 Chr.12: 104304517-104304517 on Build GRCh38
rs761144253 Chr.12: 104304527-104304534 on Build GRCh38
rs763171334 Chr.12: 104304589-104304589 on Build GRCh38
rs776829396 Chr.12: 104304565-104304565 on Build GRCh38
rs766198574 Chr.12: 104304480-104304480 on Build GRCh38
rs780732669 Chr.12: 104304603-104304603 on Build GRCh38
rs765550437 Chr.12: 104304581-104304581 on Build GRCh38
rs777230205 Chr.12: 104304624-104304624 on Build GRCh38
rs756911903 Chr.12: 104304727-104304727 on Build GRCh38
rs753356677 Chr.12: 104304809-104304809 on Build GRCh38
rs369881597 Chr.12: 104304606-104304606 on Build GRCh38
rs745850351 Chr.12: 104304641-104304641 on Build GRCh38
rs769721639 Chr.12: 104304778-104304778 on Build GRCh38
rs760610427 Chr.12: 104304496-104304496 on Build GRCh38
rs758201934 Chr.12: 104304826-104304826 on Build GRCh38
rs754510884 Chr.12: 104304597-104304597 on Build GRCh38
rs753380448 Chr.12: 104304711-104304711 on Build GRCh38
rs773242712 Chr.12: 104304652-104304652 on Build GRCh38
rs868212037 Chr.12: 104304617-104304617 on Build GRCh38
rs772102855 Chr.12: 104304644-104304644 on Build GRCh38
rs778612480 Chr.12: 104304730-104304730 on Build GRCh38
rs1011795155 Chr.12: 104304540-104304540 on Build GRCh38
rs749111044 Chr.12: 104304626-104304626 on Build GRCh38
rs751452445 Chr.12: 104304639-104304639 on Build GRCh38
rs1055476456 Chr.12: 104304796-104304796 on Build GRCh38
rs775694938 Chr.12: 104304487-104304487 on Build GRCh38
rs764411810 Chr.12: 104304503-104304503 on Build GRCh38
rs77135236 Chr.12: 104304659-104304659 on Build GRCh38
rs779653610 Chr.12: 104304444-104304444 on Build GRCh38
rs35757377 Chr.12: 104304897-104304898 on Build GRCh38
rs553038165 Chr.12: 104304690-104304690 on Build GRCh38
rs746544757 Chr.12: 104304538-104304538 on Build GRCh38
rs753238826 Chr.12: 104304519-104304520 on Build GRCh38
rs776208689 Chr.12: 104304452-104304452 on Build GRCh38
rs997225109 Chr.12: 104304886-104304886 on Build GRCh38
rs757509139 Chr.12: 104304767-104304767 on Build GRCh38
rs371354192 Chr.12: 104304543-104304543 on Build GRCh38
rs565042362 Chr.12: 104304755-104304755 on Build GRCh38
rs371562364 Chr.12: 104304461-104304461 on Build GRCh38
rs909100860 Chr.12: 104304860-104304860 on Build GRCh38
rs770156613 Chr.12: 104304650-104304650 on Build GRCh38
rs1042067426 Chr.12: 104304814-104304814 on Build GRCh38
rs766607882 Chr.12: 104304666-104304666 on Build GRCh38
rs763136842 Chr.12: 104304783-104304783 on Build GRCh38
rs201124067 Chr.12: 104304705-104304705 on Build GRCh38
rs936981999 Chr.12: 104304757-104304757 on Build GRCh38
rs1135923 Chr.12: 104304661-104304661 on Build GRCh38
rs774766153 Chr.12: 104304788-104304788 on Build GRCh38
rs544458127 Chr.12: 104304789-104304789 on Build GRCh38
rs763953750 Chr.12: 104304462-104304462 on Build GRCh38
rs768010297 Chr.12: 104304694-104304694 on Build GRCh38
rs772192645 Chr.12: 104304746-104304746 on Build GRCh38
rs779817123 Chr.12: 104304849-104304850 on Build GRCh38
rs1019746769 Chr.12: 104304575-104304575 on Build GRCh38
rs561075240 Chr.12: 104304546-104304546 on Build GRCh38
rs367597572 Chr.12: 104304515-104304515 on Build GRCh38
rs771157795 Chr.12: 104304787-104304787 on Build GRCh38
rs749385426 Chr.12: 104304874-104304874 on Build GRCh38
rs756927289 Chr.12: 104304812-104304812 on Build GRCh38
rs755834351 Chr.12: 104304614-104304614 on Build GRCh38
rs759008834 Chr.12: 104304530-104304530 on Build GRCh38
rs751970447 Chr.12: 104304595-104304595 on Build GRCh38
rs537709102 Chr.12: 104304446-104304446 on Build GRCh38
rs762165593 Chr.12: 104304574-104304574 on Build GRCh38
rs774486177 Chr.12: 104304899-104304899 on Build GRCh38
rs35925321 Chr.12: 104304608-104304609 on Build GRCh38
rs764221279 Chr.12: 104304563-104304563 on Build GRCh38
rs371069815 Chr.12: 104304900-104304900 on Build GRCh38
rs374997109 Chr.12: 104304512-104304512 on Build GRCh38
rs530625247 Chr.12: 104304756-104304756 on Build GRCh38
rs368657994 Chr.12: 104304448-104304448 on Build GRCh38
rs764927719 Chr.12: 104304817-104304817 on Build GRCh38
rs750256191 Chr.12: 104304822-104304822 on Build GRCh38
rs183928257 Chr.12: 104304560-104304560 on Build GRCh38
rs199802893 Chr.12: 104304747-104304747 on Build GRCh38
rs773102131 Chr.12: 104304549-104304549 on Build GRCh38
rs770912295 Chr.12: 104304550-104304550 on Build GRCh38
rs542835354 Chr.12: 104304502-104304502 on Build GRCh38
rs779022548 Chr.12: 104304880-104304880 on Build GRCh38
rs201650501 Chr.12: 104304717-104304717 on Build GRCh38
rs761969497 Chr.12: 104304647-104304647 on Build GRCh38
rs770588903 Chr.12: 104304632-104304632 on Build GRCh38
rs894932620 Chr.12: 104304890-104304890 on Build GRCh38
rs377091329 Chr.12: 104304876-104304876 on Build GRCh38
rs763000507 Chr.12: 104304473-104304473 on Build GRCh38
rs746894135 Chr.12: 104304865-104304865 on Build GRCh38
rs929223145 Chr.12: 104304507-104304507 on Build GRCh38
rs750860139 Chr.12: 104304587-104304587 on Build GRCh38
rs761943719 Chr.12: 104304511-104304511 on Build GRCh38
rs754921010 Chr.12: 104304870-104304870 on Build GRCh38
rs763558119 Chr.12: 104304805-104304805 on Build GRCh38
rs377581157 Chr.12: 104304792-104304792 on Build GRCh38
rs923195110 Chr.12: 104304469-104304469 on Build GRCh38
rs769543323 Chr.12: 104304470-104304470 on Build GRCh38
rs773507190 Chr.12: 104304780-104304780 on Build GRCh38
rs777759118 Chr.12: 104304871-104304871 on Build GRCh38
rs778927712 Chr.12: 104304640-104304640 on Build GRCh38
rs866455204 Chr.12: 104304793-104304793 on Build GRCh38
rs377094192 Chr.12: 104304541-104304541 on Build GRCh38
rs903013554 Chr.12: 104304815-104304815 on Build GRCh38
rs764533697 Chr.12: 104304586-104304587 on Build GRCh38
rs760835291 Chr.12: 104304561-104304561 on Build GRCh38
rs372620381 Chr.12: 104304856-104304856 on Build GRCh38
rs11111987 Chr.12: 104304804-104304804 on Build GRCh38
rs776915935 Chr.12: 104304646-104304646 on Build GRCh38
rs780414452 Chr.12: 104304533-104304533 on Build GRCh38
rs769416916 Chr.12: 104304547-104304547 on Build GRCh38
rs376622366 Chr.12: 104304594-104304594 on Build GRCh38
rs754229041 Chr.12: 104304708-104304719 on Build GRCh38
rs200764339 Chr.12: 104304799-104304799 on Build GRCh38
rs754069073 Chr.12: 104304504-104304504 on Build GRCh38

View Full Product Details