Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 107776259-107776761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCL4
Entrez Gene ID
121549
Gene Name
achaete-scute family bHLH transcription factor 4
Gene Aliases
HASH4, bHLHa44
UniGene
Hs.149910
Gene Chromosome Location
Chr.12: 107774385-107776644 on Build GRCh38
Transcripts
NM_203436.2

SNP Detail

SNP ID Chromosome Location
rs371705892 Chr.12: 107776511-107776511 on Build GRCh38
rs879183599 Chr.12: 107776580-107776580 on Build GRCh38
rs755792996 Chr.12: 107776434-107776434 on Build GRCh38
rs35676126 Chr.12: 107776503-107776504 on Build GRCh38
rs908729663 Chr.12: 107776330-107776330 on Build GRCh38
rs926729568 Chr.12: 107776284-107776284 on Build GRCh38
rs762102084 Chr.12: 107776724-107776725 on Build GRCh38
rs779898402 Chr.12: 107776459-107776459 on Build GRCh38
rs559440274 Chr.12: 107776429-107776429 on Build GRCh38
rs75343959 Chr.12: 107776524-107776524 on Build GRCh38
rs896544327 Chr.12: 107776512-107776512 on Build GRCh38
rs980735597 Chr.12: 107776605-107776605 on Build GRCh38
rs904240185 Chr.12: 107776435-107776435 on Build GRCh38
rs748792936 Chr.12: 107776640-107776640 on Build GRCh38
rs925433858 Chr.12: 107776295-107776295 on Build GRCh38
rs568153001 Chr.12: 107776695-107776695 on Build GRCh38
rs34132780 Chr.12: 107776395-107776396 on Build GRCh38
rs530701731 Chr.12: 107776495-107776495 on Build GRCh38
rs931395253 Chr.12: 107776292-107776292 on Build GRCh38
rs569826808 Chr.12: 107776734-107776734 on Build GRCh38
rs535537156 Chr.12: 107776721-107776721 on Build GRCh38
rs937941903 Chr.12: 107776691-107776691 on Build GRCh38
rs751837542 Chr.12: 107776401-107776401 on Build GRCh38
rs866490591 Chr.12: 107776340-107776340 on Build GRCh38
rs780062502 Chr.12: 107776683-107776683 on Build GRCh38
rs114446870 Chr.12: 107776673-107776673 on Build GRCh38
rs955134497 Chr.12: 107776648-107776648 on Build GRCh38
rs1032555893 Chr.12: 107776574-107776574 on Build GRCh38
rs932352064 Chr.12: 107776388-107776389 on Build GRCh38
rs1033683500 Chr.12: 107776625-107776625 on Build GRCh38
rs756046425 Chr.12: 107776675-107776675 on Build GRCh38
rs546584269 Chr.12: 107776680-107776680 on Build GRCh38
rs879010506 Chr.12: 107776305-107776305 on Build GRCh38
rs973715961 Chr.12: 107776685-107776685 on Build GRCh38
rs185191271 Chr.12: 107776451-107776451 on Build GRCh38
rs908242556 Chr.12: 107776443-107776443 on Build GRCh38
rs986544345 Chr.12: 107776653-107776653 on Build GRCh38
rs984235331 Chr.12: 107776649-107776649 on Build GRCh38
rs923454033 Chr.12: 107776620-107776620 on Build GRCh38
rs376739075 Chr.12: 107776551-107776551 on Build GRCh38
rs919823191 Chr.12: 107776688-107776688 on Build GRCh38
rs761644587 Chr.12: 107776477-107776477 on Build GRCh38
rs964946109 Chr.12: 107776335-107776335 on Build GRCh38
rs866823848 Chr.12: 107776394-107776394 on Build GRCh38
rs1001115278 Chr.12: 107776558-107776558 on Build GRCh38
rs763120684 Chr.12: 107776342-107776342 on Build GRCh38
rs1055379166 Chr.12: 107776510-107776510 on Build GRCh38
rs1049393284 Chr.12: 107776389-107776389 on Build GRCh38
rs1020952911 Chr.12: 107776531-107776531 on Build GRCh38
rs569845338 Chr.12: 107776487-107776487 on Build GRCh38
rs747406935 Chr.12: 107776431-107776431 on Build GRCh38
rs148641153 Chr.12: 107776582-107776582 on Build GRCh38
rs975319189 Chr.12: 107776345-107776345 on Build GRCh38
rs900844303 Chr.12: 107776386-107776386 on Build GRCh38
rs1024269780 Chr.12: 107776552-107776552 on Build GRCh38
rs79082941 Chr.12: 107776471-107776471 on Build GRCh38
rs999965097 Chr.12: 107776448-107776448 on Build GRCh38
rs898419079 Chr.12: 107776483-107776483 on Build GRCh38
rs764374941 Chr.12: 107776363-107776363 on Build GRCh38
rs920990002 Chr.12: 107776359-107776359 on Build GRCh38

View Full Product Details