Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108289422-108289931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMKLR1
Entrez Gene ID
1240
Gene Name
chemerin chemokine-like receptor 1
Gene Aliases
CHEMERINR, ChemR23, DEZ, RVER1
UniGene
Hs.197143
Gene Chromosome Location
Chr.12: 108288044-108339348 on Build GRCh38
Transcripts
NM_001142343.1 NM_001142344.1 NM_001142345.1 NM_004072.2 XM_017018820.1

SNP Detail

SNP ID Chromosome Location
rs551471631 Chr.12: 108289868-108289868 on Build GRCh38
rs1946 Chr.12: 108289601-108289601 on Build GRCh38
rs140751727 Chr.12: 108289513-108289513 on Build GRCh38
rs965650532 Chr.12: 108289822-108289822 on Build GRCh38
rs774525944 Chr.12: 108289593-108289593 on Build GRCh38
rs559560344 Chr.12: 108289609-108289609 on Build GRCh38
rs374910053 Chr.12: 108289689-108289689 on Build GRCh38
rs748039019 Chr.12: 108289741-108289741 on Build GRCh38
rs998925796 Chr.12: 108289643-108289643 on Build GRCh38
rs1036953525 Chr.12: 108289602-108289602 on Build GRCh38
rs148550214 Chr.12: 108289884-108289884 on Build GRCh38
rs553172518 Chr.12: 108289838-108289838 on Build GRCh38
rs903277508 Chr.12: 108289640-108289640 on Build GRCh38
rs534751141 Chr.12: 108289827-108289827 on Build GRCh38
rs1020328325 Chr.12: 108289813-108289813 on Build GRCh38
rs188586237 Chr.12: 108289842-108289842 on Build GRCh38
rs149429396 Chr.12: 108289681-108289681 on Build GRCh38
rs543182203 Chr.12: 108289595-108289595 on Build GRCh38
rs1031253593 Chr.12: 108289664-108289664 on Build GRCh38
rs1138433 Chr.12: 108289626-108289626 on Build GRCh38
rs983453680 Chr.12: 108289510-108289510 on Build GRCh38
rs921047415 Chr.12: 108289545-108289545 on Build GRCh38
rs969455774 Chr.12: 108289448-108289448 on Build GRCh38
rs765103763 Chr.12: 108289875-108289875 on Build GRCh38
rs556094657 Chr.12: 108289537-108289537 on Build GRCh38
rs543412880 Chr.12: 108289885-108289885 on Build GRCh38
rs759619153 Chr.12: 108289817-108289817 on Build GRCh38
rs3179574 Chr.12: 108289711-108289711 on Build GRCh38
rs1028864854 Chr.12: 108289683-108289683 on Build GRCh38
rs757969507 Chr.12: 108289781-108289781 on Build GRCh38
rs894517606 Chr.12: 108289611-108289611 on Build GRCh38
rs568959504 Chr.12: 108289515-108289517 on Build GRCh38
rs781055999 Chr.12: 108289477-108289477 on Build GRCh38
rs777493941 Chr.12: 108289903-108289903 on Build GRCh38
rs953072804 Chr.12: 108289698-108289698 on Build GRCh38
rs916150949 Chr.12: 108289807-108289807 on Build GRCh38
rs903143957 Chr.12: 108289864-108289864 on Build GRCh38
rs991762089 Chr.12: 108289775-108289775 on Build GRCh38
rs191838957 Chr.12: 108289603-108289603 on Build GRCh38
rs144842686 Chr.12: 108289761-108289761 on Build GRCh38
rs1008746392 Chr.12: 108289901-108289901 on Build GRCh38
rs1045876421 Chr.12: 108289632-108289632 on Build GRCh38
rs535780982 Chr.12: 108289876-108289876 on Build GRCh38
rs911530882 Chr.12: 108289716-108289716 on Build GRCh38
rs958211582 Chr.12: 108289485-108289485 on Build GRCh38
rs184297449 Chr.12: 108289729-108289729 on Build GRCh38
rs917186469 Chr.12: 108289887-108289887 on Build GRCh38
rs889045820 Chr.12: 108289891-108289891 on Build GRCh38
rs747013620 Chr.12: 108289512-108289512 on Build GRCh38
rs548486374 Chr.12: 108289647-108289647 on Build GRCh38
rs188550878 Chr.12: 108289476-108289476 on Build GRCh38
rs911624359 Chr.12: 108289589-108289589 on Build GRCh38
rs552432916 Chr.12: 108289725-108289725 on Build GRCh38
rs769074173 Chr.12: 108289538-108289538 on Build GRCh38
rs1030804654 Chr.12: 108289467-108289467 on Build GRCh38
rs976015612 Chr.12: 108289840-108289840 on Build GRCh38
rs752395656 Chr.12: 108289883-108289883 on Build GRCh38
rs1028896165 Chr.12: 108289888-108289888 on Build GRCh38
rs542090225 Chr.12: 108289487-108289487 on Build GRCh38
rs557611490 Chr.12: 108289882-108289882 on Build GRCh38
rs987221611 Chr.12: 108289586-108289586 on Build GRCh38
rs962122863 Chr.12: 108289720-108289720 on Build GRCh38
rs528239371 Chr.12: 108289608-108289608 on Build GRCh38
rs553113051 Chr.12: 108289473-108289473 on Build GRCh38
rs987085864 Chr.12: 108289699-108289699 on Build GRCh38
rs575976866 Chr.12: 108289584-108289584 on Build GRCh38
rs531738570 Chr.12: 108289672-108289672 on Build GRCh38
rs947125084 Chr.12: 108289566-108289566 on Build GRCh38

View Full Product Details