Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108292598-108293095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMKLR1
Entrez Gene ID
1240
Gene Name
chemerin chemokine-like receptor 1
Gene Aliases
CHEMERINR, ChemR23, DEZ, RVER1
UniGene
Hs.197143
Gene Chromosome Location
Chr.12: 108288044-108339348 on Build GRCh38
Transcripts
NM_001142343.1 NM_001142344.1 NM_001142345.1 NM_004072.2 XM_017018820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7155131 c.166C>A p.L56M Chr.12: 108292797-108292797 on Build GRCh38
4818767 c.156C>A p.F52L Chr.12: 108292807-108292807 on Build GRCh38
3870658 c.114G>A p.V38V Chr.12: 108292849-108292849 on Build GRCh38
7088014 c.339G>C p.K113N Chr.12: 108292624-108292624 on Build GRCh38
243742 c.42C>T p.Y14Y Chr.12: 108292921-108292921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200117272 Chr.12: 108292638-108292638 on Build GRCh38
rs376509653 Chr.12: 108292943-108292943 on Build GRCh38
rs1021087305 Chr.12: 108293031-108293031 on Build GRCh38
rs568799764 Chr.12: 108292868-108292868 on Build GRCh38
rs986262868 Chr.12: 108292771-108292771 on Build GRCh38
rs758307028 Chr.12: 108292709-108292709 on Build GRCh38
rs771231971 Chr.12: 108292828-108292828 on Build GRCh38
rs756505533 Chr.12: 108292679-108292679 on Build GRCh38
rs1017711717 Chr.12: 108293016-108293016 on Build GRCh38
rs749519428 Chr.12: 108292745-108292745 on Build GRCh38
rs780083303 Chr.12: 108292961-108292961 on Build GRCh38
rs757850821 Chr.12: 108292804-108292804 on Build GRCh38
rs543389807 Chr.12: 108292726-108292726 on Build GRCh38
rs769110194 Chr.12: 108292967-108292967 on Build GRCh38
rs751158064 Chr.12: 108292908-108292908 on Build GRCh38
rs557311025 Chr.12: 108292977-108292977 on Build GRCh38
rs773693526 Chr.12: 108292986-108292986 on Build GRCh38
rs779887002 Chr.12: 108292718-108292718 on Build GRCh38
rs764435421 Chr.12: 108292787-108292787 on Build GRCh38
rs199548216 Chr.12: 108292854-108292854 on Build GRCh38
rs966403573 Chr.12: 108293065-108293065 on Build GRCh38
rs754239043 Chr.12: 108292793-108292793 on Build GRCh38
rs779984645 Chr.12: 108292806-108292806 on Build GRCh38
rs926636183 Chr.12: 108292641-108292641 on Build GRCh38
rs551039443 Chr.12: 108292988-108292988 on Build GRCh38
rs141421422 Chr.12: 108292778-108292778 on Build GRCh38
rs770141378 Chr.12: 108292770-108292770 on Build GRCh38
rs375387234 Chr.12: 108292747-108292747 on Build GRCh38
rs998469960 Chr.12: 108293064-108293064 on Build GRCh38
rs761225810 Chr.12: 108292785-108292785 on Build GRCh38
rs761137943 Chr.12: 108292846-108292846 on Build GRCh38
rs537099458 Chr.12: 108292889-108292889 on Build GRCh38
rs776627631 Chr.12: 108292741-108292741 on Build GRCh38
rs937791898 Chr.12: 108292622-108292622 on Build GRCh38
rs367921179 Chr.12: 108292689-108292689 on Build GRCh38
rs992175629 Chr.12: 108292898-108292898 on Build GRCh38
rs566373835 Chr.12: 108293021-108293021 on Build GRCh38
rs989256528 Chr.12: 108293024-108293024 on Build GRCh38
rs752505056 Chr.12: 108292927-108292927 on Build GRCh38
rs368753612 Chr.12: 108292642-108292642 on Build GRCh38
rs768584661 Chr.12: 108292740-108292740 on Build GRCh38
rs533727463 Chr.12: 108293061-108293061 on Build GRCh38
rs759151030 Chr.12: 108292994-108292994 on Build GRCh38
rs1055891109 Chr.12: 108292632-108292632 on Build GRCh38
rs775871090 Chr.12: 108292779-108292779 on Build GRCh38
rs958421726 Chr.12: 108293018-108293018 on Build GRCh38
rs764876734 Chr.12: 108292865-108292865 on Build GRCh38
rs202234426 Chr.12: 108292940-108292940 on Build GRCh38
rs373064633 Chr.12: 108292921-108292921 on Build GRCh38
rs773683372 Chr.12: 108292778-108292779 on Build GRCh38
rs775702945 Chr.12: 108292842-108292842 on Build GRCh38
rs369733869 Chr.12: 108292991-108292991 on Build GRCh38
rs755870319 Chr.12: 108292938-108292938 on Build GRCh38
rs756878978 Chr.12: 108292796-108292796 on Build GRCh38
rs748007872 Chr.12: 108292827-108292827 on Build GRCh38
rs777127074 Chr.12: 108292968-108292968 on Build GRCh38
rs555495349 Chr.12: 108293062-108293062 on Build GRCh38
rs770042629 Chr.12: 108292981-108292981 on Build GRCh38
rs761155992 Chr.12: 108292833-108292833 on Build GRCh38
rs369981839 Chr.12: 108292866-108292866 on Build GRCh38
rs201022995 Chr.12: 108292696-108292696 on Build GRCh38
rs369716757 Chr.12: 108292963-108292963 on Build GRCh38
rs778362252 Chr.12: 108292693-108292693 on Build GRCh38
rs764920082 Chr.12: 108292816-108292816 on Build GRCh38
rs200344687 Chr.12: 108292946-108292946 on Build GRCh38
rs772375747 Chr.12: 108292951-108292951 on Build GRCh38
rs746753225 Chr.12: 108292731-108292731 on Build GRCh38
rs754946817 Chr.12: 108292910-108292910 on Build GRCh38
rs1003939483 Chr.12: 108292818-108292818 on Build GRCh38
rs370635582 Chr.12: 108292665-108292665 on Build GRCh38
rs753472328 Chr.12: 108292682-108292682 on Build GRCh38
rs376579038 Chr.12: 108293005-108293005 on Build GRCh38
rs192034694 Chr.12: 108292772-108292772 on Build GRCh38
rs1043775854 Chr.12: 108292972-108292972 on Build GRCh38
rs375250297 Chr.12: 108292666-108292666 on Build GRCh38
rs1006667517 Chr.12: 108292665-108292665 on Build GRCh38
rs531338899 Chr.12: 108292849-108292849 on Build GRCh38
rs945414719 Chr.12: 108292719-108292719 on Build GRCh38
rs997852514 Chr.12: 108292811-108292811 on Build GRCh38
rs899754973 Chr.12: 108293055-108293055 on Build GRCh38
rs779039224 Chr.12: 108292830-108292830 on Build GRCh38
rs202104056 Chr.12: 108292758-108292758 on Build GRCh38
rs1039629989 Chr.12: 108293043-108293043 on Build GRCh38
rs942145947 Chr.12: 108292878-108292878 on Build GRCh38
rs751530283 Chr.12: 108292807-108292807 on Build GRCh38
rs745898843 Chr.12: 108292831-108292831 on Build GRCh38
rs560704906 Chr.12: 108292681-108292681 on Build GRCh38
rs913869104 Chr.12: 108292920-108292920 on Build GRCh38
rs764818755 Chr.12: 108292803-108292803 on Build GRCh38
rs150895979 Chr.12: 108292815-108292815 on Build GRCh38
rs776930995 Chr.12: 108292852-108292852 on Build GRCh38
rs1050798494 Chr.12: 108293066-108293066 on Build GRCh38

View Full Product Details