Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108894161-108894666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAO
Entrez Gene ID
1610
Gene Name
D-amino acid oxidase
Gene Aliases
DAAO, DAMOX, OXDA
UniGene
Hs.113227
Gene Chromosome Location
Chr.12: 108880073-108901043 on Build GRCh38
Transcripts
NM_001917.4 XM_005268692.3 XM_011538004.2 XM_011538005.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3687935 c.600G>T p.G200G Chr.12: 108894355-108894355 on Build GRCh38
430279 c.571C>T p.R191* Chr.12: 108894326-108894326 on Build GRCh38
1628357 c.609G>T p.M203I Chr.12: 108894364-108894364 on Build GRCh38
4776367 c.572G>A p.R191Q Chr.12: 108894327-108894327 on Build GRCh38
6639995 c.612G>T p.K204N Chr.12: 108894367-108894367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902036461 Chr.12: 108894603-108894603 on Build GRCh38
rs374876157 Chr.12: 108894502-108894503 on Build GRCh38
rs369749171 Chr.12: 108894405-108894405 on Build GRCh38
rs192312168 Chr.12: 108894257-108894257 on Build GRCh38
rs980902259 Chr.12: 108894543-108894543 on Build GRCh38
rs3825251 Chr.12: 108894208-108894208 on Build GRCh38
rs187944229 Chr.12: 108894224-108894224 on Build GRCh38
rs763346176 Chr.12: 108894245-108894245 on Build GRCh38
rs781499915 Chr.12: 108894297-108894297 on Build GRCh38
rs113090452 Chr.12: 108894230-108894230 on Build GRCh38
rs76477430 Chr.12: 108894519-108894519 on Build GRCh38
rs767019180 Chr.12: 108894262-108894262 on Build GRCh38
rs577672767 Chr.12: 108894481-108894481 on Build GRCh38
rs370118391 Chr.12: 108894381-108894381 on Build GRCh38
rs765190914 Chr.12: 108894355-108894355 on Build GRCh38
rs1039137042 Chr.12: 108894601-108894601 on Build GRCh38
rs968495066 Chr.12: 108894475-108894475 on Build GRCh38
rs753523915 Chr.12: 108894470-108894470 on Build GRCh38
rs368892733 Chr.12: 108894252-108894252 on Build GRCh38
rs200850756 Chr.12: 108894351-108894351 on Build GRCh38
rs1027402276 Chr.12: 108894186-108894186 on Build GRCh38
rs774892336 Chr.12: 108894322-108894322 on Build GRCh38
rs760006596 Chr.12: 108894326-108894326 on Build GRCh38
rs1031323045 Chr.12: 108894506-108894506 on Build GRCh38
rs1053465402 Chr.12: 108894529-108894529 on Build GRCh38
rs778738305 Chr.12: 108894289-108894289 on Build GRCh38
rs538560784 Chr.12: 108894486-108894486 on Build GRCh38
rs184376128 Chr.12: 108894421-108894421 on Build GRCh38
rs752336965 Chr.12: 108894589-108894589 on Build GRCh38
rs865789057 Chr.12: 108894578-108894578 on Build GRCh38
rs371113557 Chr.12: 108894283-108894283 on Build GRCh38
rs775218259 Chr.12: 108894337-108894337 on Build GRCh38
rs780181602 Chr.12: 108894212-108894212 on Build GRCh38
rs984546370 Chr.12: 108894435-108894435 on Build GRCh38
rs901104545 Chr.12: 108894443-108894443 on Build GRCh38
rs751758143 Chr.12: 108894293-108894293 on Build GRCh38
rs939977008 Chr.12: 108894579-108894579 on Build GRCh38
rs370764819 Chr.12: 108894319-108894319 on Build GRCh38
rs150760302 Chr.12: 108894327-108894327 on Build GRCh38
rs778140294 Chr.12: 108894317-108894317 on Build GRCh38
rs770153487 Chr.12: 108894227-108894227 on Build GRCh38
rs1007604835 Chr.12: 108894599-108894599 on Build GRCh38
rs146867483 Chr.12: 108894464-108894464 on Build GRCh38
rs909019930 Chr.12: 108894196-108894196 on Build GRCh38
rs763876748 Chr.12: 108894278-108894278 on Build GRCh38
rs752105034 Chr.12: 108894273-108894273 on Build GRCh38
rs768707717 Chr.12: 108894214-108894214 on Build GRCh38
rs542844495 Chr.12: 108894544-108894544 on Build GRCh38
rs955729691 Chr.12: 108894512-108894512 on Build GRCh38
rs753033542 Chr.12: 108894367-108894367 on Build GRCh38
rs1028045834 Chr.12: 108894233-108894234 on Build GRCh38
rs746851660 Chr.12: 108894501-108894502 on Build GRCh38
rs201916428 Chr.12: 108894480-108894480 on Build GRCh38
rs979967663 Chr.12: 108894478-108894478 on Build GRCh38
rs749784465 Chr.12: 108894384-108894384 on Build GRCh38
rs748120086 Chr.12: 108894301-108894301 on Build GRCh38
rs546096622 Chr.12: 108894577-108894577 on Build GRCh38
rs901052124 Chr.12: 108894605-108894605 on Build GRCh38
rs75607844 Chr.12: 108894426-108894426 on Build GRCh38
rs187271060 Chr.12: 108894528-108894528 on Build GRCh38
rs766920526 Chr.12: 108894248-108894248 on Build GRCh38
rs771521944 Chr.12: 108894386-108894386 on Build GRCh38
rs931400479 Chr.12: 108894550-108894550 on Build GRCh38
rs757061637 Chr.12: 108894474-108894475 on Build GRCh38
rs747297378 Chr.12: 108894213-108894213 on Build GRCh38
rs761787902 Chr.12: 108894353-108894353 on Build GRCh38
rs754776936 Chr.12: 108894569-108894569 on Build GRCh38
rs139166976 Chr.12: 108894350-108894350 on Build GRCh38
rs749872143 Chr.12: 108894318-108894318 on Build GRCh38
rs576791724 Chr.12: 108894445-108894445 on Build GRCh38
rs112967246 Chr.12: 108894571-108894571 on Build GRCh38
rs750170054 Chr.12: 108894359-108894359 on Build GRCh38
rs376326395 Chr.12: 108894294-108894294 on Build GRCh38
rs758205328 Chr.12: 108894363-108894363 on Build GRCh38
rs769925183 Chr.12: 108894307-108894307 on Build GRCh38
rs368059507 Chr.12: 108894380-108894380 on Build GRCh38
rs373110648 Chr.12: 108894284-108894284 on Build GRCh38
rs781206127 Chr.12: 108894364-108894364 on Build GRCh38
rs1045217537 Chr.12: 108894345-108894345 on Build GRCh38
rs928461508 Chr.12: 108894500-108894500 on Build GRCh38
rs774137595 Chr.12: 108894251-108894251 on Build GRCh38
rs772487765 Chr.12: 108894411-108894411 on Build GRCh38
rs929030294 Chr.12: 108894192-108894192 on Build GRCh38
rs924312907 Chr.12: 108894282-108894282 on Build GRCh38
rs79406702 Chr.12: 108894195-108894195 on Build GRCh38
rs755669589 Chr.12: 108894277-108894277 on Build GRCh38
rs776614494 Chr.12: 108894222-108894222 on Build GRCh38

View Full Product Details