Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108897019-108897558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAO
Entrez Gene ID
1610
Gene Name
D-amino acid oxidase
Gene Aliases
DAAO, DAMOX, OXDA
UniGene
Hs.113227
Gene Chromosome Location
Chr.12: 108880073-108901043 on Build GRCh38
Transcripts
NM_001917.4 XM_005268692.3 XM_011538004.2 XM_011538005.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6031843 c.680C>T p.P227L Chr.12: 108897073-108897073 on Build GRCh38
5601330 c.691C>T p.P231S Chr.12: 108897084-108897084 on Build GRCh38
934656 c.655C>A p.P219T Chr.12: 108897048-108897048 on Build GRCh38
3954141 c.661A>G p.R221G Chr.12: 108897054-108897054 on Build GRCh38
467744 c.628A>G p.M210V Chr.12: 108897021-108897021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768305609 Chr.12: 108897105-108897105 on Build GRCh38
rs991094671 Chr.12: 108897500-108897500 on Build GRCh38
rs957921333 Chr.12: 108897419-108897419 on Build GRCh38
rs539414902 Chr.12: 108897218-108897218 on Build GRCh38
rs868014293 Chr.12: 108897071-108897071 on Build GRCh38
rs759267411 Chr.12: 108897138-108897138 on Build GRCh38
rs750080350 Chr.12: 108897374-108897374 on Build GRCh38
rs117200825 Chr.12: 108897475-108897475 on Build GRCh38
rs569298712 Chr.12: 108897159-108897159 on Build GRCh38
rs879520577 Chr.12: 108897247-108897247 on Build GRCh38
rs774160299 Chr.12: 108897136-108897136 on Build GRCh38
rs1046111670 Chr.12: 108897359-108897359 on Build GRCh38
rs77011540 Chr.12: 108897364-108897364 on Build GRCh38
rs368493622 Chr.12: 108897095-108897095 on Build GRCh38
rs1029014433 Chr.12: 108897179-108897179 on Build GRCh38
rs143555005 Chr.12: 108897373-108897373 on Build GRCh38
rs200391092 Chr.12: 108897066-108897066 on Build GRCh38
rs76676467 Chr.12: 108897361-108897361 on Build GRCh38
rs928842059 Chr.12: 108897518-108897518 on Build GRCh38
rs919731901 Chr.12: 108897279-108897279 on Build GRCh38
rs894489628 Chr.12: 108897241-108897241 on Build GRCh38
rs971026887 Chr.12: 108897533-108897533 on Build GRCh38
rs990834280 Chr.12: 108897065-108897065 on Build GRCh38
rs955062783 Chr.12: 108897193-108897193 on Build GRCh38
rs76292940 Chr.12: 108897362-108897362 on Build GRCh38
rs773123069 Chr.12: 108897118-108897118 on Build GRCh38
rs556610360 Chr.12: 108897515-108897515 on Build GRCh38
rs891336743 Chr.12: 108897463-108897463 on Build GRCh38
rs1006239566 Chr.12: 108897250-108897250 on Build GRCh38
rs558114821 Chr.12: 108897231-108897231 on Build GRCh38
rs779585168 Chr.12: 108897321-108897321 on Build GRCh38
rs1008340579 Chr.12: 108897194-108897194 on Build GRCh38
rs753916764 Chr.12: 108897067-108897067 on Build GRCh38
rs1054498161 Chr.12: 108897402-108897402 on Build GRCh38
rs755498610 Chr.12: 108897310-108897310 on Build GRCh38
rs76594384 Chr.12: 108897363-108897363 on Build GRCh38
rs140084770 Chr.12: 108897280-108897280 on Build GRCh38
rs762998898 Chr.12: 108897122-108897122 on Build GRCh38
rs757422803 Chr.12: 108897074-108897074 on Build GRCh38
rs372787944 Chr.12: 108897336-108897336 on Build GRCh38
rs950073833 Chr.12: 108897345-108897345 on Build GRCh38
rs767319263 Chr.12: 108897333-108897333 on Build GRCh38
rs896839916 Chr.12: 108897487-108897487 on Build GRCh38
rs753061582 Chr.12: 108897053-108897053 on Build GRCh38
rs778064377 Chr.12: 108897492-108897492 on Build GRCh38
rs753304654 Chr.12: 108897376-108897376 on Build GRCh38
rs932850421 Chr.12: 108897440-108897440 on Build GRCh38
rs562456281 Chr.12: 108897272-108897272 on Build GRCh38
rs1045813791 Chr.12: 108897073-108897073 on Build GRCh38
rs751222640 Chr.12: 108897107-108897107 on Build GRCh38
rs1024817778 Chr.12: 108897507-108897507 on Build GRCh38
rs946190536 Chr.12: 108897083-108897083 on Build GRCh38
rs893287102 Chr.12: 108897433-108897433 on Build GRCh38
rs758786375 Chr.12: 108897102-108897102 on Build GRCh38
rs372956556 Chr.12: 108897108-108897108 on Build GRCh38
rs769363507 Chr.12: 108897110-108897110 on Build GRCh38
rs758676423 Chr.12: 108897090-108897090 on Build GRCh38
rs961078543 Chr.12: 108897223-108897223 on Build GRCh38
rs954476399 Chr.12: 108897328-108897328 on Build GRCh38
rs766992423 Chr.12: 108897124-108897124 on Build GRCh38
rs1015654491 Chr.12: 108897478-108897478 on Build GRCh38
rs1026760735 Chr.12: 108897496-108897496 on Build GRCh38
rs959190194 Chr.12: 108897497-108897497 on Build GRCh38
rs1015266330 Chr.12: 108897209-108897209 on Build GRCh38
rs779493322 Chr.12: 108897078-108897078 on Build GRCh38
rs1037593728 Chr.12: 108897266-108897266 on Build GRCh38
rs1030208302 Chr.12: 108897315-108897315 on Build GRCh38
rs746442296 Chr.12: 108897088-108897088 on Build GRCh38
rs893318343 Chr.12: 108897140-108897140 on Build GRCh38
rs544932023 Chr.12: 108897108-108897108 on Build GRCh38
rs749902245 Chr.12: 108897254-108897254 on Build GRCh38
rs938804911 Chr.12: 108897178-108897178 on Build GRCh38
rs573855130 Chr.12: 108897460-108897460 on Build GRCh38
rs977557269 Chr.12: 108897495-108897495 on Build GRCh38

View Full Product Details