Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110327462-110327971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2A2
Entrez Gene ID
488
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2
Gene Aliases
ATP2B, DAR, DD, SERCA2
UniGene
Hs.506759
Gene Chromosome Location
Chr.12: 110281227-110351093 on Build GRCh38
Transcripts
NM_001681.3 NM_170665.3 XM_005253888.2 XM_011538402.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3416469 c.691G>A p.E231K Chr.12: 110327613-110327613 on Build GRCh38
934938 c.698G>A p.G233D Chr.12: 110327620-110327620 on Build GRCh38
5816083 c.1044C>T p.I348I Chr.12: 110327966-110327966 on Build GRCh38
934939 c.767T>G p.F256C Chr.12: 110327689-110327689 on Build GRCh38
3368644 c.690C>T p.T230T Chr.12: 110327612-110327612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201573336 Chr.12: 110327789-110327789 on Build GRCh38
rs761165682 Chr.12: 110327819-110327819 on Build GRCh38
rs750310602 Chr.12: 110327835-110327835 on Build GRCh38
rs746331231 Chr.12: 110327933-110327933 on Build GRCh38
rs763520238 Chr.12: 110327517-110327517 on Build GRCh38
rs549471747 Chr.12: 110327579-110327579 on Build GRCh38
rs778028514 Chr.12: 110327633-110327633 on Build GRCh38
rs531838109 Chr.12: 110327540-110327540 on Build GRCh38
rs771912488 Chr.12: 110327580-110327580 on Build GRCh38
rs200807356 Chr.12: 110327853-110327853 on Build GRCh38
rs34150151 Chr.12: 110327774-110327775 on Build GRCh38
rs757279201 Chr.12: 110327918-110327918 on Build GRCh38
rs138160424 Chr.12: 110327488-110327488 on Build GRCh38
rs746309999 Chr.12: 110327761-110327761 on Build GRCh38
rs988306010 Chr.12: 110327496-110327496 on Build GRCh38
rs369594808 Chr.12: 110327532-110327532 on Build GRCh38
rs149129262 Chr.12: 110327492-110327492 on Build GRCh38
rs759761421 Chr.12: 110327533-110327533 on Build GRCh38
rs1012731102 Chr.12: 110327716-110327716 on Build GRCh38
rs770031210 Chr.12: 110327631-110327631 on Build GRCh38
rs528936918 Chr.12: 110327905-110327905 on Build GRCh38
rs775164424 Chr.12: 110327777-110327777 on Build GRCh38
rs539141956 Chr.12: 110327693-110327693 on Build GRCh38
rs866239874 Chr.12: 110327932-110327932 on Build GRCh38
rs756426005 Chr.12: 110327903-110327903 on Build GRCh38
rs1803737 Chr.12: 110327938-110327938 on Build GRCh38
rs1043346857 Chr.12: 110327732-110327732 on Build GRCh38
rs751358352 Chr.12: 110327893-110327893 on Build GRCh38
rs143229439 Chr.12: 110327495-110327495 on Build GRCh38
rs772529432 Chr.12: 110327936-110327936 on Build GRCh38
rs373919719 Chr.12: 110327792-110327792 on Build GRCh38
rs121912733 Chr.12: 110327725-110327725 on Build GRCh38
rs1014324433 Chr.12: 110327831-110327831 on Build GRCh38
rs775259638 Chr.12: 110327515-110327515 on Build GRCh38
rs146073302 Chr.12: 110327906-110327906 on Build GRCh38
rs773902750 Chr.12: 110327512-110327512 on Build GRCh38
rs767203045 Chr.12: 110327538-110327538 on Build GRCh38
rs986622422 Chr.12: 110327536-110327536 on Build GRCh38
rs757243926 Chr.12: 110327578-110327578 on Build GRCh38
rs772303786 Chr.12: 110327768-110327768 on Build GRCh38
rs143310856 Chr.12: 110327612-110327612 on Build GRCh38
rs776134017 Chr.12: 110327799-110327799 on Build GRCh38
rs760353108 Chr.12: 110327780-110327780 on Build GRCh38
rs569637288 Chr.12: 110327855-110327855 on Build GRCh38
rs748353664 Chr.12: 110327613-110327613 on Build GRCh38
rs778661160 Chr.12: 110327600-110327600 on Build GRCh38
rs376731879 Chr.12: 110327718-110327718 on Build GRCh38
rs944717624 Chr.12: 110327564-110327564 on Build GRCh38
rs142721178 Chr.12: 110327705-110327705 on Build GRCh38
rs750240717 Chr.12: 110327603-110327603 on Build GRCh38
rs188765221 Chr.12: 110327548-110327548 on Build GRCh38
rs749389852 Chr.12: 110327909-110327909 on Build GRCh38
rs56277015 Chr.12: 110327528-110327528 on Build GRCh38
rs753306264 Chr.12: 110327558-110327558 on Build GRCh38
rs1048975717 Chr.12: 110327666-110327666 on Build GRCh38
rs749276909 Chr.12: 110327644-110327644 on Build GRCh38
rs779524206 Chr.12: 110327930-110327930 on Build GRCh38
rs760228338 Chr.12: 110327541-110327541 on Build GRCh38

View Full Product Details