Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110387506-110388012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC7
Entrez Gene ID
51434
Gene Name
anaphase promoting complex subunit 7
Gene Aliases
APC7
UniGene
Hs.719935
Gene Chromosome Location
Chr.12: 110372900-110403730 on Build GRCh38
Transcripts
NM_001137664.1 NM_016238.2 XM_005253903.3 XM_011538454.2 XM_017019425.1 XM_017019426.1 XM_017019427.1 XM_017019428.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1298906 c.652G>C p.E218Q Chr.12: 110387863-110387863 on Build GRCh38
4760096 c.626A>G p.Y209C Chr.12: 110387787-110387787 on Build GRCh38
1358693 c.725C>T p.A242V Chr.12: 110387790-110387790 on Build GRCh38
3359535 c.627T>C p.Y209Y Chr.12: 110387786-110387786 on Build GRCh38
1358694 c.623C>T p.A208V Chr.12: 110387790-110387790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745758366 Chr.12: 110387676-110387676 on Build GRCh38
rs756804998 Chr.12: 110387705-110387705 on Build GRCh38
rs748942760 Chr.12: 110387957-110387957 on Build GRCh38
rs957868269 Chr.12: 110387587-110387587 on Build GRCh38
rs753234725 Chr.12: 110387870-110387870 on Build GRCh38
rs1007474190 Chr.12: 110387528-110387531 on Build GRCh38
rs372373517 Chr.12: 110387935-110387935 on Build GRCh38
rs371661838 Chr.12: 110387888-110387888 on Build GRCh38
rs1038291785 Chr.12: 110387780-110387780 on Build GRCh38
rs930679243 Chr.12: 110387598-110387598 on Build GRCh38
rs199898262 Chr.12: 110387810-110387810 on Build GRCh38
rs941298883 Chr.12: 110387759-110387759 on Build GRCh38
rs764442525 Chr.12: 110387912-110387915 on Build GRCh38
rs892121688 Chr.12: 110387709-110387709 on Build GRCh38
rs749985145 Chr.12: 110387942-110387942 on Build GRCh38
rs758009768 Chr.12: 110387943-110387943 on Build GRCh38
rs948879290 Chr.12: 110387648-110387648 on Build GRCh38
rs759512682 Chr.12: 110387827-110387827 on Build GRCh38
rs777201239 Chr.12: 110387930-110387930 on Build GRCh38
rs550571187 Chr.12: 110387578-110387578 on Build GRCh38
rs767743380 Chr.12: 110387920-110387923 on Build GRCh38
rs999104309 Chr.12: 110387684-110387684 on Build GRCh38
rs184914089 Chr.12: 110387849-110387849 on Build GRCh38
rs778072322 Chr.12: 110387699-110387699 on Build GRCh38
rs952031061 Chr.12: 110387956-110387956 on Build GRCh38
rs1049122901 Chr.12: 110387621-110387621 on Build GRCh38
rs757004344 Chr.12: 110387931-110387931 on Build GRCh38
rs754086982 Chr.12: 110387904-110387904 on Build GRCh38
rs1018419381 Chr.12: 110387541-110387541 on Build GRCh38
rs543702060 Chr.12: 110387952-110387952 on Build GRCh38
rs766907617 Chr.12: 110387884-110387886 on Build GRCh38
rs368734362 Chr.12: 110387932-110387932 on Build GRCh38
rs150897727 Chr.12: 110387821-110387821 on Build GRCh38
rs890890712 Chr.12: 110387798-110387798 on Build GRCh38
rs201808610 Chr.12: 110387923-110387923 on Build GRCh38
rs139954100 Chr.12: 110387961-110387961 on Build GRCh38
rs536542391 Chr.12: 110387964-110387964 on Build GRCh38
rs971947914 Chr.12: 110387919-110387919 on Build GRCh38
rs746900196 Chr.12: 110387789-110387789 on Build GRCh38
rs1050712353 Chr.12: 110387797-110387797 on Build GRCh38
rs763239910 Chr.12: 110387801-110387801 on Build GRCh38
rs1045914067 Chr.12: 110387649-110387649 on Build GRCh38
rs928730283 Chr.12: 110387663-110387663 on Build GRCh38
rs907416476 Chr.12: 110387652-110387652 on Build GRCh38
rs1020263555 Chr.12: 110387610-110387610 on Build GRCh38
rs201209979 Chr.12: 110387889-110387889 on Build GRCh38
rs867713124 Chr.12: 110387584-110387584 on Build GRCh38
rs375107592 Chr.12: 110387704-110387704 on Build GRCh38
rs766877353 Chr.12: 110387556-110387556 on Build GRCh38
rs916405278 Chr.12: 110387713-110387713 on Build GRCh38
rs752016031 Chr.12: 110387903-110387906 on Build GRCh38
rs532207744 Chr.12: 110387977-110387977 on Build GRCh38
rs986000069 Chr.12: 110387552-110387552 on Build GRCh38
rs776229522 Chr.12: 110387793-110387793 on Build GRCh38
rs373690811 Chr.12: 110387897-110387897 on Build GRCh38
rs61754907 Chr.12: 110387786-110387786 on Build GRCh38
rs916022780 Chr.12: 110387633-110387633 on Build GRCh38
rs367762819 Chr.12: 110387757-110387757 on Build GRCh38
rs771053134 Chr.12: 110387803-110387803 on Build GRCh38
rs919204823 Chr.12: 110387927-110387927 on Build GRCh38
rs768053095 Chr.12: 110387828-110387828 on Build GRCh38
rs756894970 Chr.12: 110387960-110387960 on Build GRCh38
rs768592404 Chr.12: 110387790-110387790 on Build GRCh38
rs542356588 Chr.12: 110387619-110387619 on Build GRCh38
rs911768843 Chr.12: 110387566-110387566 on Build GRCh38
rs778494172 Chr.12: 110387743-110387743 on Build GRCh38
rs772180548 Chr.12: 110387767-110387767 on Build GRCh38
rs191761851 Chr.12: 110387590-110387590 on Build GRCh38

View Full Product Details