Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110436031-110436538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPC3
Entrez Gene ID
10094
Gene Name
actin related protein 2/3 complex subunit 3
Gene Aliases
ARC21, p21-Arc
UniGene
Hs.524741
Gene Chromosome Location
Chr.12: 110434890-110450411 on Build GRCh38
Transcripts
NM_001278556.1 NM_001287222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1947061 c.447C>T p.F149F Chr.12: 110436137-110436137 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs890594174 Chr.12: 110436062-110436062 on Build GRCh38
rs939100349 Chr.12: 110436516-110436516 on Build GRCh38
rs368946645 Chr.12: 110436255-110436255 on Build GRCh38
rs1023814637 Chr.12: 110436228-110436228 on Build GRCh38
rs995046006 Chr.12: 110436430-110436430 on Build GRCh38
rs746040046 Chr.12: 110436176-110436176 on Build GRCh38
rs1009482316 Chr.12: 110436074-110436074 on Build GRCh38
rs751458309 Chr.12: 110436118-110436118 on Build GRCh38
rs776903182 Chr.12: 110436216-110436216 on Build GRCh38
rs776464231 Chr.12: 110436412-110436412 on Build GRCh38
rs752531052 Chr.12: 110436137-110436137 on Build GRCh38
rs888081312 Chr.12: 110436498-110436498 on Build GRCh38
rs893617794 Chr.12: 110436085-110436085 on Build GRCh38
rs779083687 Chr.12: 110436156-110436156 on Build GRCh38
rs754887091 Chr.12: 110436124-110436124 on Build GRCh38
rs923826196 Chr.12: 110436264-110436264 on Build GRCh38
rs1029878722 Chr.12: 110436335-110436335 on Build GRCh38
rs11553917 Chr.12: 110436116-110436116 on Build GRCh38
rs1033343312 Chr.12: 110436249-110436249 on Build GRCh38
rs761649448 Chr.12: 110436431-110436431 on Build GRCh38
rs757510145 Chr.12: 110436143-110436143 on Build GRCh38
rs898093569 Chr.12: 110436123-110436123 on Build GRCh38
rs866290048 Chr.12: 110436232-110436232 on Build GRCh38
rs779704048 Chr.12: 110436232-110436233 on Build GRCh38
rs547541950 Chr.12: 110436317-110436317 on Build GRCh38
rs906294091 Chr.12: 110436232-110436232 on Build GRCh38
rs202063357 Chr.12: 110436214-110436214 on Build GRCh38
rs765822603 Chr.12: 110436242-110436242 on Build GRCh38
rs989384663 Chr.12: 110436406-110436406 on Build GRCh38
rs750005002 Chr.12: 110436081-110436081 on Build GRCh38
rs577557084 Chr.12: 110436463-110436463 on Build GRCh38
rs535586985 Chr.12: 110436300-110436300 on Build GRCh38
rs751602350 Chr.12: 110436239-110436240 on Build GRCh38
rs532068844 Chr.12: 110436285-110436285 on Build GRCh38
rs368898128 Chr.12: 110436215-110436215 on Build GRCh38
rs1015153403 Chr.12: 110436084-110436084 on Build GRCh38
rs761611955 Chr.12: 110436061-110436061 on Build GRCh38
rs570345755 Chr.12: 110436375-110436375 on Build GRCh38
rs982375073 Chr.12: 110436254-110436254 on Build GRCh38
rs762251378 Chr.12: 110436220-110436220 on Build GRCh38
rs564784250 Chr.12: 110436277-110436277 on Build GRCh38
rs542211245 Chr.12: 110436092-110436092 on Build GRCh38
rs773033686 Chr.12: 110436222-110436222 on Build GRCh38
rs559194473 Chr.12: 110436386-110436386 on Build GRCh38
rs772545473 Chr.12: 110436511-110436511 on Build GRCh38
rs758021807 Chr.12: 110436221-110436222 on Build GRCh38
rs548181925 Chr.12: 110436341-110436341 on Build GRCh38
rs371633553 Chr.12: 110436136-110436136 on Build GRCh38
rs746599908 Chr.12: 110436315-110436315 on Build GRCh38
rs750076555 Chr.12: 110436067-110436067 on Build GRCh38
rs371765450 Chr.12: 110436202-110436202 on Build GRCh38
rs76226633 Chr.12: 110436268-110436268 on Build GRCh38
rs945171726 Chr.12: 110436464-110436464 on Build GRCh38
rs1036474217 Chr.12: 110436368-110436368 on Build GRCh38
rs574704287 Chr.12: 110436059-110436059 on Build GRCh38
rs767458524 Chr.12: 110436509-110436509 on Build GRCh38
rs565769880 Chr.12: 110436305-110436305 on Build GRCh38
rs898020033 Chr.12: 110436411-110436411 on Build GRCh38
rs1026588409 Chr.12: 110436186-110436186 on Build GRCh38
rs764957112 Chr.12: 110436070-110436070 on Build GRCh38
rs759459517 Chr.12: 110436508-110436508 on Build GRCh38
rs374547515 Chr.12: 110436233-110436233 on Build GRCh38
rs765949521 Chr.12: 110436117-110436117 on Build GRCh38
rs754880081 Chr.12: 110436516-110436518 on Build GRCh38
rs754642678 Chr.12: 110436119-110436119 on Build GRCh38
rs906409163 Chr.12: 110436501-110436501 on Build GRCh38
rs780112499 Chr.12: 110436207-110436207 on Build GRCh38
rs535842609 Chr.12: 110436322-110436322 on Build GRCh38
rs560822834 Chr.12: 110436221-110436221 on Build GRCh38
rs372335617 Chr.12: 110436465-110436465 on Build GRCh38
rs369206641 Chr.12: 110436338-110436338 on Build GRCh38
rs911207077 Chr.12: 110436353-110436353 on Build GRCh38
rs547135304 Chr.12: 110436302-110436302 on Build GRCh38
rs537766698 Chr.12: 110436382-110436382 on Build GRCh38

View Full Product Details