Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110846776-110847285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC63
Entrez Gene ID
160762
Gene Name
coiled-coil domain containing 63
Gene Aliases
ODA5
UniGene
Hs.437141
Gene Chromosome Location
Chr.12: 110844489-110907535 on Build GRCh38
Transcripts
NM_001286243.1 NM_001286244.1 NM_152591.2 XM_006719262.1 XM_006719263.2 XM_011537999.1 XM_011538001.2

SNP Detail

SNP ID Chromosome Location
rs922839650 Chr.12: 110847073-110847073 on Build GRCh38
rs996624179 Chr.12: 110847197-110847197 on Build GRCh38
rs35841828 Chr.12: 110846833-110846834 on Build GRCh38
rs773034208 Chr.12: 110847132-110847132 on Build GRCh38
rs970242088 Chr.12: 110847246-110847246 on Build GRCh38
rs1033510587 Chr.12: 110847220-110847220 on Build GRCh38
rs375879174 Chr.12: 110847154-110847154 on Build GRCh38
rs116321620 Chr.12: 110846987-110846987 on Build GRCh38
rs998573914 Chr.12: 110847218-110847218 on Build GRCh38
rs540900646 Chr.12: 110846969-110846969 on Build GRCh38
rs1014143539 Chr.12: 110847150-110847150 on Build GRCh38
rs964374649 Chr.12: 110847048-110847048 on Build GRCh38
rs533678170 Chr.12: 110847076-110847076 on Build GRCh38
rs745731598 Chr.12: 110847181-110847181 on Build GRCh38
rs1055473256 Chr.12: 110847142-110847142 on Build GRCh38
rs982906792 Chr.12: 110847099-110847099 on Build GRCh38
rs562903312 Chr.12: 110847242-110847242 on Build GRCh38
rs929356184 Chr.12: 110847125-110847125 on Build GRCh38
rs959702841 Chr.12: 110847237-110847237 on Build GRCh38
rs1031759115 Chr.12: 110847227-110847227 on Build GRCh38
rs1037812893 Chr.12: 110847097-110847097 on Build GRCh38
rs12303008 Chr.12: 110847027-110847027 on Build GRCh38
rs568204053 Chr.12: 110846867-110846867 on Build GRCh38
rs916093671 Chr.12: 110847070-110847070 on Build GRCh38
rs940422219 Chr.12: 110846944-110846944 on Build GRCh38
rs907588903 Chr.12: 110846904-110846904 on Build GRCh38
rs1001237573 Chr.12: 110846978-110846978 on Build GRCh38
rs557334295 Chr.12: 110847046-110847046 on Build GRCh38
rs535637121 Chr.12: 110846889-110846889 on Build GRCh38
rs141086095 Chr.12: 110847014-110847014 on Build GRCh38
rs976059780 Chr.12: 110846831-110846831 on Build GRCh38
rs938900241 Chr.12: 110846808-110846808 on Build GRCh38
rs188864895 Chr.12: 110847193-110847193 on Build GRCh38
rs2339635 Chr.12: 110847068-110847068 on Build GRCh38
rs895685036 Chr.12: 110847149-110847149 on Build GRCh38
rs61944261 Chr.12: 110846875-110846875 on Build GRCh38
rs1019874633 Chr.12: 110847005-110847005 on Build GRCh38
rs2339636 Chr.12: 110847217-110847217 on Build GRCh38
rs960568925 Chr.12: 110847052-110847052 on Build GRCh38
rs1014009279 Chr.12: 110846992-110846992 on Build GRCh38
rs964301375 Chr.12: 110846807-110846807 on Build GRCh38
rs1037442137 Chr.12: 110847117-110847117 on Build GRCh38
rs192127129 Chr.12: 110847238-110847238 on Build GRCh38
rs1047293184 Chr.12: 110846888-110846888 on Build GRCh38
rs967411619 Chr.12: 110847023-110847023 on Build GRCh38
rs908726919 Chr.12: 110847103-110847103 on Build GRCh38
rs374883838 Chr.12: 110846993-110846993 on Build GRCh38
rs936999148 Chr.12: 110847141-110847141 on Build GRCh38
rs898838605 Chr.12: 110846973-110846973 on Build GRCh38
rs899702092 Chr.12: 110847196-110847196 on Build GRCh38
rs1057482401 Chr.12: 110846824-110846824 on Build GRCh38
rs1047003118 Chr.12: 110846981-110846981 on Build GRCh38
rs922903023 Chr.12: 110846845-110846845 on Build GRCh38
rs948788631 Chr.12: 110846926-110846926 on Build GRCh38
rs1056098099 Chr.12: 110846981-110846981 on Build GRCh38
rs1020079013 Chr.12: 110847016-110847016 on Build GRCh38
rs1008576335 Chr.12: 110847192-110847192 on Build GRCh38
rs554440826 Chr.12: 110847116-110847116 on Build GRCh38
rs955657028 Chr.12: 110847092-110847092 on Build GRCh38
rs115581704 Chr.12: 110846798-110846798 on Build GRCh38
rs1037759699 Chr.12: 110846951-110846951 on Build GRCh38
rs755670973 Chr.12: 110846861-110846861 on Build GRCh38
rs1021366291 Chr.12: 110847205-110847205 on Build GRCh38
rs752175696 Chr.12: 110846802-110846802 on Build GRCh38
rs553194123 Chr.12: 110846818-110846818 on Build GRCh38

View Full Product Details